Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC11A2 SIRNA Assays ›
          Gene Symbol:
          SLC11A2
          Gene Name:
          solute carrier family 11 member 2
          Gene Aliases:
          AHMIO1, DCT1, DMT1, NRAMP2
          Chromosome Location:
          Chr.12: 50952263 - 51028335 on Build GRCh38
          UniGene ID:
          Hs.505545
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s9709
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC11A2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000617.2 NP_000608.1 15 1591
          NM_001174125.1 NP_001167596.1 15 1583
          NM_001174126.1 NP_001167597.1 15 1591
          NM_001174127.1 NP_001167598.1 15 1586
          NM_001174128.1 NP_001167599.1 15 1586
          NM_001174129.1 NP_001167600.1 15 1851
          NM_001174130.1 NP_001167601.1 14 1566
          NR_033421.1 15 1511
          NR_033422.1 15 1586
          XM_005268911.3 XP_005268968.1 15 1643
          XM_005268912.4 XP_005268969.1 14 1768
          XM_011538404.2 XP_011536706.1 15 1967
          XM_011538405.2 XP_011536707.1 15 1575
          XM_017019355.1 XP_016874844.1 15 1851
          XM_017019356.1 XP_016874845.1 15 1781
          GenBank mRNA AB004857.1 - 15 1534
          AB062284.1 - 15 1508
          AF046997.1 - 15 1544
          AF064484.1 - 15 1534
          AJ493662.1 - 15 1583
          AK094735.1 - 15 1851
          AK128640.1 - 15 1483
          AK296445.1 - 14 1545
          AK315987.1 - 15 1497
          AK316507.1 - 14 1522
          AL832886.1 - 5 917
          BC002592.2 - 15 1538 3160522
          BC100014.1 - 15 1559 5300266
          KT455015.1 - 15 1547
          L37347.1 - 12 1284

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00167206_m1

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          heme biosynthetic process
          cobalt ion transport
          copper ion transport
          manganese ion transport
          cellular iron ion homeostasis
          learning or memory
          response to iron ion
          ferrous iron transport
          lead ion transport
          transferrin transport
          copper ion transmembrane transport
          dendrite morphogenesis
          erythrocyte development
          multicellular organismal iron ion homeostasis
          cadmium ion transmembrane transport
          ferrous iron import
          manganese ion transmembrane transport
          hydrogen ion transmembrane transport
          ferrous iron transmembrane transport
          copper ion transmembrane transporter activity
          iron ion transmembrane transporter activity
          manganese ion transmembrane transporter activity
          protein binding
          hydrogen ion transmembrane transporter activity
          cadmium ion transmembrane transporter activity
          cobalt ion transmembrane transporter activity
          ferrous iron transmembrane transporter activity
          lead ion transmembrane transporter activity
          solute:proton symporter activity
          ferrous iron uptake transmembrane transporter activity
          transferrin transmembrane transporter activity
          cadmium ion binding
          transition metal ion transmembrane transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0