Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACVR1 SIRNA Assays ›
          Gene Symbol:
          ACVR1
          Gene Name:
          activin A receptor type 1
          Gene Aliases:
          ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI
          Chromosome Location:
          Chr.2: 157736446 - 157875880 on Build GRCh38
          UniGene ID:
          Hs.470316
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s974
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACVR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105.4 NP_001096.1 7 1209
          NM_001111067.2 NP_001104537.1 7 1028
          XM_005246939.3 XP_005246996.1 7 1080
          XM_005246940.3 XP_005246997.1 7 1048
          XM_006712825.3 XP_006712888.1 7 1161
          XM_011512106.2 XP_011510408.1 8 1128
          XM_011512107.2 XP_011510409.1 7 1100
          XM_011512108.2 XP_011510410.1 7 2507
          GenBank mRNA AK297765.1 - 7 827
          AK313630.1 - 7 1004
          BC033867.1 - 7 1209 5209711
          BC036748.1 - 7 1209
          KU177885.1 - 5 779
          KU177886.1 - 5 779
          KU177887.1 - 5 779
          L02911.1 - 8 1119
          Z22534.1 - 6 882

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00153836_m1

          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          patterning of blood vessels
          in utero embryonic development
          gastrulation with mouth forming second
          mesoderm formation
          neural crest cell migration
          acute inflammatory response
          embryonic heart tube morphogenesis
          outflow tract septum morphogenesis
          atrioventricular valve morphogenesis
          mitral valve morphogenesis
          endocardial cushion morphogenesis
          endocardial cushion fusion
          atrial septum primum morphogenesis
          protein phosphorylation
          transforming growth factor beta receptor signaling pathway
          germ cell development
          determination of left/right symmetry
          negative regulation of signal transduction
          positive regulation of pathway-restricted SMAD protein phosphorylation
          peptidyl-threonine phosphorylation
          signal transduction by protein phosphorylation
          regulation of ossification
          positive regulation of cell migration
          positive regulation of bone mineralization
          BMP signaling pathway
          activin receptor signaling pathway
          negative regulation of activin receptor signaling pathway
          positive regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          smooth muscle cell differentiation
          pharyngeal system development
          pathway-restricted SMAD protein phosphorylation
          ventricular septum morphogenesis
          cardiac muscle cell fate commitment
          BMP signaling pathway involved in heart development
          endocardial cushion cell fate commitment
          cellular response to glucocorticoid stimulus
          cellular response to BMP stimulus
          positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
          positive regulation of determination of dorsal identity
          negative regulation of extrinsic apoptotic signaling pathway
          protein kinase activity
          protein serine/threonine kinase activity
          transmembrane receptor protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          transforming growth factor beta receptor activity, type I
          protein binding
          ATP binding
          activin receptor activity, type I
          peptide hormone binding
          protein homodimerization activity
          SMAD binding
          metal ion binding
          activin binding
          transforming growth factor beta binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0