Search
Search

Prkd2| Gene Symbol: Prkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252458.1 | NP_001239387.1 | 6, 7 | 1173 | ||
| NM_178900.4 | NP_849231.1 | 5, 6 | 1458 | |||
| GenBank mRNA | AK037030.1 | - | 5, 6 | 1459 | ||
| AK041207.1 | - | 4, 5 | 678 | |||
| AK154029.1 | - | 5, 6 | 1307 | |||
| AK170236.1 | - | 6, 7 | 1146 | |||
| AK170761.1 | - | 6, 7 | 1137 | |||
| BC095949.1 | - | 6, 7 | 1123 | 4950957 | ||
| BC096444.1 | - | 6, 7 | 1141 | 4216827 | ||