Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: D330045A20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175326.5 | NP_780535.1 | 6 | 1329 | ||
GenBank mRNA | AK052376.1 | - | 6 | 1253 | ||
AK155588.1 | - | 6 | 1326 | |||
AK171459.1 | - | 6 | 1328 | |||
BC112416.1 | - | 5 | 1137 | |||
BC113128.1 | - | 6 | 1229 | 40061080 |