Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TXNDC5 SIRNA Assays ›
Gene Symbol:
Txndc5
Gene Name:
thioredoxin domain containing 5
Gene Aliases:
AL022641, ERp46, PC-TRP
Chromosome Location:
Chr.13: 38500266 - 38528824 on Build GRCm38
UniGene ID:
Mm.28622
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s98419
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Txndc5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289598.1
NP_001276527.1
5
715
NM_001289599.1
NP_001276528.1
5
682
NM_145367.4
NP_663342.3
5
739
GenBank mRNA
AK136046.1
-
5
627
AK142049.1
-
5
725
AK150781.1
-
5
723
AK169680.1
-
5
706
AK185996.1
-
1
55
AK205431.1
-
1
55
AK224911.1
-
4
392
AK224913.1
-
5
726
AK224922.1
-
5
684
AK224923.1
-
5
706
AK224924.1
-
5
714
AK224926.1
-
5
684
AK224927.1
-
5
735
AK224930.1
-
5
687
AK224933.1
-
5
676
AK224934.1
-
5
730
AK224936.1
-
5
729
AK224937.1
-
5
728
AK224938.1
-
5
723
AK224939.1
-
5
716
AK224943.1
-
5
687
AK224945.1
-
5
723
AK224946.1
-
5
726
AK224947.1
-
5
713
AK224948.1
-
5
687
AK224949.1
-
5
676
AK224950.1
-
5
676
AK224951.1
-
5
734
AK224952.1
-
5
702
AK224956.1
-
5
646
AK224957.1
-
5
692
AK224958.1
-
5
692
AK224960.1
-
5
734
AK224962.1
-
5
694
AK224963.1
-
5
735
AK224964.1
-
5
735
AK224965.1
-
5
711
AK224966.1
-
5
711
AK224967.1
-
5
702
AK224968.1
-
3
269
AK224969.1
-
3
269
AK224970.1
-
5
676
AK224971.1
-
5
736
AK224972.1
-
5
726
AK224973.1
-
5
676
AK224974.1
-
5
679
AK224976.1
-
5
709
AK224977.1
-
5
723
AK224978.1
-
5
723
AK224979.1
-
5
710
AK224981.1
-
5
680
AK224982.1
-
5
735
AK224983.1
-
5
724
AK224984.1
-
5
689
AK224985.1
-
5
693
AK224986.1
-
5
737
AK224988.1
-
5
713
AK224989.1
-
5
730
AK224990.1
-
5
725
AK224991.1
-
5
720
AK224992.1
-
5
720
AK224993.1
-
5
720
AK224994.1
-
5
728
AK224995.1
-
5
691
AK224996.1
-
5
662
AK224997.1
-
5
687
AK224998.1
-
5
685
AK224999.1
-
5
707
AK225004.1
-
5
728
AK225005.1
-
5
706
AK225007.1
-
5
728
AK225010.1
-
5
728
AK225011.1
-
5
728
AK225012.1
-
5
497
AK225013.1
-
5
497
AK225014.1
-
5
706
AK225015.1
-
5
497
AK225016.1
-
5
636
AK225017.1
-
5
684
AK225019.1
-
5
716
AK225020.1
-
5
667
AK225021.1
-
5
667
AK225022.1
-
5
515
AK225023.1
-
5
741
AY243534.1
-
5
702
AY548112.1
-
5
663
BC016252.1
-
5
706
4911455
BC024505.1
-
5
651
4983496
BC036155.1
-
4
325
BC046789.1
-
5
665
5716897
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
response to endoplasmic reticulum stress
apoptotic cell clearance
cell redox homeostasis
protein disulfide isomerase activity
isomerase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-7wptj:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0