Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kctd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111028.1 | NP_001104498.1 | 10, 11 | 1162 | ||
NM_001285933.1 | NP_001272862.1 | 10, 11 | 1289 | |||
NM_134073.2 | NP_598834.2 | 9, 10 | 1012 | |||
GenBank mRNA | AK087312.1 | - | 10, 11 | 1036 | ||
AK158922.1 | - | 9, 10 | 1014 | |||
AK166501.1 | - | 9, 10 | 848 | |||
BC011302.1 | - | 10, 11 | 1103 | |||
CK618608.1 | - | 3, 4 | 307 |