Search
Search

OPHN1| Gene Symbol: OPHN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002547.2 | NP_002538.1 | 17 | 1685 | ||
| XM_005262270.1 | XP_005262327.1 | 17 | 1755 | |||
| XM_006724653.2 | XP_006724716.1 | 17 | 1755 | |||
| XM_011530961.1 | XP_011529263.1 | 17 | 1606 | |||
| XM_017029555.1 | XP_016885044.1 | 17 | 1755 | |||
| GenBank mRNA | AB102656.1 | - | 16 | 1391 | ||
| AJ001189.1 | - | 17 | 2029 | |||
| AK295769.1 | - | 17 | 1755 | |||
| BC140763.1 | - | 17 | 1723 | 9021631 | ||