Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ankrd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025572.1 | NP_001020743.1 | 7 | 908 | ||
GenBank mRNA | AK133715.1 | - | 7 | 907 | ||
AK134530.1 | - | 2 | 87 | |||
AK153850.1 | - | 6 | 817 | |||
BC023046.1 | - | 7 | 779 | |||
BC050185.1 | - | 7 | 888 | |||
BC064777.1 | - | 6 | 823 | |||
BC080825.1 | - | 6 | 831 |