Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MACROD1 SIRNA Assays ›
Gene Symbol:
Macrod1
Gene Name:
MACRO domain containing 1
Gene Aliases:
AI604841, AW743046, D930010J01Rik, Lrp16
Chromosome Location:
Chr.19: 7056715 - 7198062 on Build GRCm38
UniGene ID:
Mm.439977
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s98791
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Macrod1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_134147.4
NP_598908.2
2
475
GenBank mRNA
AK052998.1
-
2
434
BC008653.1
-
2
200
3597023
BQ126199.1
-
2
475
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_139337, XM_006230677, AF404762, FQ216840, AF419856.
Gene Ontology Categories:
Function(s)
Process(es)
cellular response to DNA damage stimulus
purine nucleoside metabolic process
protein de-ADP-ribosylation
purine-specific mismatch base pair DNA N-glycosylase activity
histone deacetylase activity
uracil DNA N-glycosylase activity
protein-N-terminal asparagine amidohydrolase activity
UDP-3-O-[3-hydroxymyristoyl] N-acetylglucosamine deacetylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
single-strand selective uracil DNA N-glycosylase activity
iprodione amidohydrolase activity
(3,5-dichlorophenylurea)acetate amidohydrolase activity
DNA N-glycosylase activity
deacetylase activity
4'-(2-hydroxyisopropyl)phenylurea amidohydrolase activity
didemethylisoproturon amidohydrolase activity
N-isopropylacetanilide amidohydrolase activity
N-cyclohexylformamide amidohydrolase activity
isonicotinic acid hydrazide hydrolase activity
cis-aconitamide amidase activity
gamma-N-formylaminovinylacetate hydrolase activity
DNA-3-methylbase glycosylase activity
N2-acetyl-L-lysine deacetylase activity
O-succinylbenzoate synthase activity
indoleacetamide hydrolase activity
N-acetylcitrulline deacetylase activity
N-acetylgalactosamine-6-phosphate deacetylase activity
diacetylchitobiose deacetylase activity
chitooligosaccharide deacetylase activity
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0