Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Il1rl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133193.3 | NP_573456.1 | 3 | 474 | ||
GenBank mRNA | AF284433.1 | - | 3 | 479 | ||
AK054530.1 | - | 1 | 2975 | |||
AK085780.1 | - | 3 | 342 | |||
BC004628.1 | - | 3 | 468 | |||
BC117797.2 | - | 3 | 475 | 40092208 |