Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Pacs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153129.2 | NP_694769.2 | 17 | 2254 | ||
GenBank mRNA | AK154563.1 | - | 2128 | |||
AK164770.1 | - | 18 | 2240 | |||
AK171969.1 | - | 17 | 2237 | |||
AK219829.1 | - | 2 | 161 | |||
BC034670.1 | - | 6 | 620 | |||
BC062886.1 | - | 17 | 2254 | 5706409 |