Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRM7 SIRNA Assays ›
Gene Symbol:
Grm7
Gene Name:
glutamate receptor, metabotropic 7
Gene Aliases:
6330570A01Rik, BB176677, C030018L03, E130018M02Rik, Gpr1g, Gprc1g, mGluR7
Chromosome Location:
Chr.6: 110645458 - 111567230 on Build GRCm38
UniGene ID:
Mm.240881
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s99106
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Grm7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_177328.3
NP_796302.2
8
2314
GenBank mRNA
AK053447.1
-
9
2364
BC080315.1
-
8
2314
6853901
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_031040, XM_008763199, D16817, U06832.
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
conditioned taste aversion
behavioral fear response
signal transduction
G-protein coupled receptor signaling pathway
negative regulation of adenylate cyclase activity
adenylate cyclase-inhibiting G-protein coupled glutamate receptor signaling pathway
chemical synaptic transmission
sensory perception of sound
sensory perception of smell
memory
short-term memory
associative learning
negative regulation of glutamate secretion
transmission of nerve impulse
adult behavior
negative regulation of cAMP biosynthetic process
regulation of cyclase activity
multicellular organismal response to stress
regulation of ion transmembrane transport
neurological system process
response to stimulus
regulation of synaptic transmission, glutamatergic
calcium ion transmembrane transport
regulation of neuron death
adenylate cyclase inhibiting G-protein coupled glutamate receptor activity
group III metabotropic glutamate receptor activity
signal transducer activity
G-protein coupled receptor activity
voltage-gated calcium channel activity
calcium channel regulator activity
calcium ion binding
calmodulin binding
glutamate receptor activity
adenylate cyclase inhibitor activity
glutamate binding
PDZ domain binding
protein homodimerization activity
calcium-dependent protein binding
serine binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0