Search
Search

OVGP1| Gene Symbol: OVGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002557.3 | NP_002548.3 | 11 | 1434 | ||
| GenBank mRNA | AB208783.1 | - | 9 | 1816 | ||
| AK298757.1 | - | 9 | 1177 | |||
| AK301726.1 | - | 7 | 916 | |||
| AK314074.1 | - | 11 | 1392 | |||
| AY189737.1 | - | 11 | 1435 | |||
| BC126177.1 | - | 11 | 1417 | 8991893 | ||
| BC136406.1 | - | 11 | 1417 | 9020393 | ||
| U09550.1 | - | 11 | 1390 | |||