Search
Search

OVGP1| Gene Symbol: OVGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002557.3 | NP_002548.3 | 5 | 514 | ||
| GenBank mRNA | AB208783.1 | - | 3 | 896 | ||
| AK295207.1 | - | 4 | 854 | |||
| AK301726.1 | - | 1 | 37 | |||
| AK314074.1 | - | 5 | 472 | |||
| AY189737.1 | - | 5 | 515 | |||
| BC126177.1 | - | 5 | 497 | 8991893 | ||
| BC136406.1 | - | 5 | 497 | 9020393 | ||
| U09550.1 | - | 5 | 470 | |||