Whole Allergen
f182 Lima bean
f182 Lima bean Scientific Information
Type:
Whole Allergen
Display Name:
Lima bean
Allergen code:
f182
Family:
Fabaceae/Leguminoacea
Species:
Phaseolus lunatus, Phaseolus inamoenus, Phaseolus limensis, Phaseolus lunatus var. lunonanus, Phaseolus tunkinensis, Phaseolus viridis
Route of Exposure:
Ingestion
Latin Name:
Phaseolus lunatus
Other Names:
Butter bean, sieva bean, double bean, Madagascar bean, chad bean, or wax bean
Summary
Lima beans are a type of legume eaten across the world; reports of lima bean allergy are relatively rare with limited information. Studies show that clinical reactions to lima bean typically correlate with cross-reactivity to several other legumes, including soya, peanut, black gram, and mesquite tree pollen.
Allergen
Nature
Lima beans, along with other beans within the genus Phaseolus (kidney, garden, butter, snap sugar, string, navy, pinto and white beans), are classified within the Leguminoacea family (1) and are eaten across the world (2). 
Taxonomic tree of Lima bean {CABI, 2019}
Domain
Eukaryota
Kingdom
Plantae
Phylum
Spermatophyta
Subphylum
Angiospermae
Class
Dicotyledonae
Order
Fabales
Family
Fabaceae
Genus
Phaseolus
Species
Phaseolus lunatus
Route of Exposure
Main
Lima beans are ingested as food across the world (2).
Other topics
There has also been a report of occupational asthma induced by exposure to boiled, minced and dried lima bean exposure (3)
Clinical Relevance
Reports of lima bean allergy are relatively limited. Studies show that clinical reactions to one legume typically correlate with cross-reactivity to several other legumes. Based on this idea, mesquite tree pollen sensitized patients also showed cross-reactivity to lima bean (2).
In a study of patients with legume hypersensitivity in the USA, 87% had positive skin prick tests to peanut (60 of 69 patients), whereas 41% were positive to lima bean (13 of 32 patients). However, none of the patients had positive oral food challenges with lima bean (4). 
Asthma
Two studies have investigated sensitization to lima beans in asthmatic patients. In one study of 470 asthma and rhinitis patients in Delhi, India, 3.4% had positive skin prick tests (2). In another study of 216 bronchial asthmatic patients, 5–6% had positive skin prick tests to lima bean, defined as a wheal of ≥3 mm (5). 
Prevention and Therapy
Avoidance
Despite the potential for cross-reactivity, clinical hypersensitivity to one legume does not warrant dietary elimination of all legumes. Oral food challenges have shown clinically important cross-reactivity to legumes is rare, and due to the lack of clinical relevance, skin prick test results should not be used to determine prolonged food restriction diets (5)
Molecular Aspects
Allergenic molecules
One report identified 23 potentially allergenic proteins isolated from lima beans (6). 
Cross-reactivity
Studies show that clinical reactions to one legume typically correlate with cross-reactivity to several other legumes such as soya, peanut, black gram (2). Studies have identified cross reactivity between lima bean and mesquite, including mesquite tree pollen.
In a study of 20 patients with a positive skin prick test to mesquite (Prosopis juliflora), none demonstrated skin-prick positivity to lima bean. However, 4 patients were positive for IgE to lima bean measured by enzyme-linked immunosorbent assay suggesting the presence of cross-reactive IgE. This study identified five shared protein allergens between mesquite and lima bean of various sizes: 20, 26, 35, 66 and 72 kDa (2). Another report confirmed cross-reactivity between mesquite tree pollen and lima bean (7).
A 35 kDa vicilin-like major allergen isolated from mesquite showed cross-reactivity to a number of legume proteins including lima bean (8).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Michael Thorpe
Last reviewed:January 2022
References
  1. Zacharisen MC, Kurup V. Anaphylaxis to beans. J Allergy Clin Immunol. 1998;101(4 Pt 1):556-7.
  2. Dhyani A, Arora N, Jain VK, Sridhara S, Singh BP. Immunoglobulin E (IgE)-mediated cross-reactivity between mesquite pollen proteins and lima bean, an edible legume. Clin Exp Immunol. 2007;149(3):517-24.
  3. Tonini S, Perfetti L, Pignatti P, Pala G, Moscato G. Occupational asthma induced by exposure to lima bean (Phaseolus lunatus). Ann Allergy Asthma Immunol. 2012;108(1):66-7.
  4. Bernhisel-Broadbent J, Sampson HA. Cross-allergenicity in the legume botanical family in children with food hypersensitivity. J Allergy Clin Immunol. 1989;83(2 Pt 1):435-40.
  5. Kumar R, Singh BP, Srivastava P, Sridhara S, Arora N, Gaur SN. Relevance of serum IgE estimation in allergic bronchial asthma with special reference to food allergy. Asian Pacific journal of allergy and immunology. 2006;24(4):191-9.
  6. Bernhisel-Broadbent J, Taylor S, Sampson HA. Cross-allergenicity in the legume botanical family in children with food hypersensitivity. II. Laboratory correlates. J Allergy Clin Immunol. 1989;84(5 Pt 1):701-9.
  7. Verma AK, Kumar S, Das M, Dwivedi PD. A comprehensive review of legume allergy. Clin Rev Allergy Immunol. 2013;45(1):30-46.
  8. Arora B, Sharma S, Gaur SN, Jain VK, Lavasa S, Arora N. Identification of a vicilin-like major allergen from Prosopis juliflora exhibiting cross- reactivity with legume food allergens. Mol Immunol. 2021;137:84-93