Type:
Whole Allergen
Whole Allergen
Whole Allergen
Trout, Rainbow trout
f204
Salmonidae
Oncorhynchus mykiss
Trout, Rainbow trout
XM64N8
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Atopic dermatitis has been seen in patients with IgE antibodies to trout.
All Pacific salmon belong to the genus Oncorhynchus, the same as the Rainbow trout. King, Coho, Pink and Chub are major species caught outside the North American west coast. Several species
A study found some cod-sensitive children to have IgE antibodies to trout.(1) IgE antibodies to trout were also measured in a group of patients suffering from atopic dermatitis using Pharmacia CAP System™ (2), and in food-allergic children (3)
Species within groups of fish, like Gadiformes (examples: codfish and hake) and Scombroid fishes (examples: mackerel and tuna) seem to share allergenic components. The overlap of allergen specificity between the groups seems to be moderate or even small.
Last reviewed: June 2022
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.