Whole Allergen
f206 Mackerel
f206 Mackerel Scientific Information
Type:
Whole Allergen
Display Name:
Mackerel
Allergen code:
f206
Family:
Scombridae
Latin Name:
Scomber scombrus
Other Names:
Mackerel
WHO/ICD-11 code:
XM95M8
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Route of Exposure
Mackerel is usually eaten cooked, marinated or smoked.
Clinical Relevance
IgE-mediated reactions
Sensitivity to mackerel is quite common.
Mackerel may contain the parasite Anisakis simplex p4, and presumed sensitivity to mackerel  may actually be an allergy to this parasite.
Other reactions
Members of the Scombroidae family easily form histamine at storage, ("scombroid poisoning").
This salt water fish, highly prized for its meat, approaches the shore in shoals in summer to spawn. The range is from the Mediterranean and Black Sea along Europe's west coast to the Arctic and down the American east coast. It occurs in dense schools just beneath the surface of the water. Catches suffer from huge size variations posing a puzzle for marine researchers and economic uncertainties for fishermen. Mackerel and other members of the family rapidly degrade and may, if improperly stored, contain large amounts of histamine associated with the bacterial enzyme histidine decarboxylase, which converts histidine to histamine. Mackerel and tuna reportedly cause intoxications referred to as scombroid poisoning (1).
Sensitivity to mackerel was seen in 20% of a group of cod-sensitive children (2). In a study of fish- and crustacea-allergic adults, the reactivity to mackerel was the second highest (3). The clinical sensitivity and specificity of Pharmacia CAP System™ f206 Mackerel was reported to be 86% and 90% respectively (4).
Molecular Aspects
Cross Reactivity
Species within groups of fish, like Gadiformes (examples: codfish and hake) and Scombroid fishes (examples: mackerel and tuna) seem to share allergenic components. The overlap of allergen specificity between the groups seems to be moderate or even small.
Compiled By
Last reviewed : June 2022
References
  1. Taylor, SL; Stratton, JE; Nordlee, JA. Histamine poisoning (scombroid fish poisoning): An allergy-like intoxication. Clin Toxicol; 1989; 27: 225-240.
  2. de Martino, M; Novembre, E; Galli, L; de Marco, A; Botarelli, P; Marano, E; Vierucci, A. Allergy to different fish species in cod-allergic children: In vivo and in vitro studies. J Allergy Clin Immunol; 1990; 86: 909-914.
  3. McCants, ML; Helbling, A; Schwartz, HJ; Lopez, M; Lehrer, SB. Skin test and RAST reactivity to seafood. J Allergy Clin Immunol; 1992; 89: 194.
  4. Hansen, TK; Abrahamsen, L; Bindslev-Jensen, C; Poulsen, LK. Results of the CAP System and Immunoblotting in clinically fish allergic adults. Allergologie; 1995; 18: 407.