Type:
Whole Allergen
Whole Allergen
Whole Allergen
Clam
f207
Veneridae
Clam
Clam
Cooked, often in sauces
Occupational asthma and food allergy caused by clam have been reported (1, 2).
The Manilla clam, also referred to as littleneck clam or carpet shell, is a hard shell clam cultivated on a very large scale in, for example, Japan, and then frozen whole and exported widely. It is commonly used around the world, for instance, in clam sauce for pasta or just steamed. This clam is closely related to quahogs and cockles and cross-reactivity to other bivalves cannot be excluded.
Occupational asthma and food allergy caused by clam have been reported (1, 2).
Cross-reactivity with other bivalves cannot be excluded. A study by Eriksson et al. demonstrated cross-reactivity between clam, krill and oyster (3).
Last reviewed : June 2022
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.