Whole Allergen
f209 Grapefruit
f209 Grapefruit Scientific Information
Type:
Whole Allergen
Display Name:
Grapefruit
Allergen code:
f209
Family:
Rutaceae
Latin Name:
Citrus paradisi
Other Names:
Grapefruit, Shaddock
WHO/ICD-11 code:
XM5J64
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Route of Exposure
Grapefruit, orange, and lemon are the principal members of the citrus family. Grapefruit is a subtropical citrus tree grown for its fruit. Its origin is not clear, but it may originally have been a hybrid of the pomelo and the sweet orange tree. Tangelo is a hybrid of grapefruit and orange, and has a less bitter taste than grapefruit.
Grapefruit is pinkish- to yellow-skinned and oblate to round, ranging in diameter from 10 to 15 cm. The most popular varieties cultivated are referred to by the colour of the segmented pulp, which can be red, white, or pink. Grapefruit flavour ranges from highly acidic and somewhat bitter to sweet and tart. Unlike other citrus fruits, grapefruit contains mercaptan, a terpene partly composed of sulphur; mercaptan has a strong influence on the taste and odour of the fruit.
Clinical Relevance
IgE-mediated reactions
Citrus fruits such as grapefruit are among the most common causes of atopic dermatitis. (1-4) IgE antibodies to grapefruit were detected in patients with atopic dermatitis, allergic rhinitis, and bronchial asthma. (5) In a study of children with atopic dermatitis, many were shown to have serum-specific IgE directed against grapefruit. (6) Many patients were also found positive in a similar study of adults. (7) An instance of food-dependent exercise-induced anaphylaxis (FDEIA) attributed to grapefruit has been reported. (8)
A study was conducted at 17 clinics in 15 European cities to evaluate the differences between some Northern countries regarding what foods, according to the patients, elicit hypersensitivity symptoms. Questionnaires concerning 86 different foods were administered to food-allergic individuals. The foods most often reported as eliciting symptoms in Russia, Estonia, and Lithuania were citrus fruits, chocolate, honey, apple, hazelnut, strawberry, fish, tomato, egg, and milk; which differed from the situation in Sweden and Denmark, where birch pollen-related foods, such as nuts, apple, pear, kiwi, stone fruits, and carrot were the most common reported culprits. The most common symptoms reported were oral allergy syndrome and urticaria. Birch pollen-related foods dominated as reported allergens in Scandinavia, whereas some mugwort-related foods were apparently of more importance in Russia and the Baltic States. Among 1 139 individuals, Grapefruit was the 18 (th)-most-often reported food allergen, purportedly resulting in adverse effects in 21%. (4)
A 3½-year-old boy is reported to have developed anaphylaxis after eating cashew nut, and later after eating a pectin-containing fruit ‘smoothie’. He was also reported to have a history of generalised pruritus after eating grapefruit. The child had skin-specific IgE for pectin, high levels of serum-specific IgE for cashew nut and pistachio, and low levels for grapefruit. The pectin in the smoothie was confirmed to be of citrus origin. The authors concluded that ingestion and inhalation of pectin can cause hypersensitivity reactions, and that allergy to cashew nut, and possibly pistachio, may be associated with pectin allergy. (9)
Other reactions
Grapefruit contains naringin, bergamottin and dihydroxybergamottin, which inhibit the protein isoform CYP3A4 in the intestine. It is via inhibition of this enzyme that grapefruit increases the effects of a variety of drugs. Therefore, the co-administration of certain drugs with grapefruit juice can markedly elevate drug bioavailability, and can alter the pharmacokinetic and pharmacodynamic parameters of the drugs. A single glass of the juice is usually sufficient to produce this interaction. (10-12)
Molecular Aspects
No allergens from this plant have yet been characterised.
In a patient with anaphylaxis from mandarin (Citrus reticulata), a lipid transfer protein was isolated from mandarin fruit. Analysis of the patient's serum also demonstrated sensitisation to the lipid transfer protein Cit s 3, from orange, as well as to Cit s 1, a germin-like allergen. (13) Whether similar allergens occur in grapefruit, a closely-related family member, was not assessed.
Cross Reactivity
Cross-reactivity within the Rutaceae family (lemon, lime, orange, tangelo) can be expected, but has not been documented to date.
Latex allergy has been reported to be associated with allergy to a number of other foods, including avocado, banana, kiwi, papaya, chestnut, peach and grapefruit. (14) A 34-year-old female with asthma and atopic dermatitis who developed severe anaphylaxis to latex was described. She was shown to have serum-specific IgE directed against latex, banana, kiwi, grapefruit, and avocado. Skin-specific IgE was also detected to banana, grapefruit, avocado, and latex extract. (15)
Compiled By
Last reviewed : June 2022
References
  1. Host A, Halken S. A prospective study of cow milk allergy in Danish infants during the first 3 years of life. Clinical course in relation to clinical and immunological type of hypersensitivity reaction. Allergy 1990;45(8):587-96.
  2. Kajosaari M. Food allergy in Finnish children aged 1 to 6 years. Acta Paediatr Scand 1982;71(5):815-9.
  3. Stogmann W, Kurz H. Atopic dermatitis and food allergy in infancy and childhood. [German] Wien Med Wochenschr 1996;146(15):411-4.
  4. Eriksson NE, Moller C, Werner S, Magnusson J, Bengtsson U, Zolubas M. Self-reported food hypersensitivity in Sweden, Denmark, Estonia, Lithuania, and Russia. J Investig Allergol Clin Immunol 2004;14(1):70-9.
  5. Chiba T et al. Clinical evaluation of Pharmacia CAP System new food allergens [abstract]. Paper presented at Jap Soc Pediatric Allergology Meeting 1992.
  6. Yamada M, Torii S. Clinical evaluation of Pharmacia CAP System new food and inhalant allergens [abstract]. Paper presented at the Jap Soc Pediatric Allergology Meeting 1992.
  7. Konatsu H, Miyagawa K, Ikezawa Z. Study of clinical efficacy of Pharmacia CAP System new allergens in patients with atopic dermatitis [abstract]. Paper presented at Jap Soc Pediatric Allergology Meeting 1992.
  8. Matsumoto R, Ogawa T, Makino T, Okano M. A clinical study of admitted the review of cases of food-dependent exercise-induced anaphylaxis. [Japanese] Arerugi 2009 May;58(5):548-53.
  9. Ferdman RM, Ong PY, Church JA. Pectin anaphylaxis and possible association with cashew allergy. Ann Allergy Asthma Immunol 2006;97(6):759-60.
  10. Dahan A, Altman H. Food-drug interaction: grapefruit juice augments drug bioavailability--mechanism, extent and relevance. Eur J Clin Nutr 2004;58(1):1-9.
  11. Fuhr U. Drug interactions with grapefruit juice. Extent, probable mechanism and clinical relevance. Drug Saf 1998;18(4):251-72.
  12. Molden E, Spigset O. Fruit and berries--interactions with drugs. [Norwegian] Tidsskr Nor Laegeforen 2007;127(24):3218-20.
  13. Ebo DG, Ahrazem O, Lopez-Torrejon G, Bridts CH, Salcedo G, Stevens WJ. Anaphylaxis from mandarin (Citrus reticulata): Identification of potential responsible allergens. Int Arch Allergy Immunol 2007;144(1):39-43.
  14. Machado M, Sant'anna C, Aires V, Rodrigues PP, Pinheiro MF, Teixeira M. Latex and banana allergies in children with myelomeningocele in the city of Rio de Janeiro. [Portuguese] Rev Assoc Med Bras 2004;50(1):83-6.
  15. Fujie S, Yagami A, Suzuki K, Akamatsu H, Matsunaga K. A case of the latex-induced anaphylaxis by contact with barium enema catheter. [Japanese] Arerugi 2004;53(1):38-42.