Whole Allergen
f273 Thyme
f273 Thyme Scientific Information
Type:
Whole Allergen
Display Name:
Thyme
Allergen code:
f273
Family:
Lamiaceae (Labiatae)
Latin Name:
Thymus vulgaris
Other Names:
Thyme, Garden thyme, Common thyme
Route of Exposure
Thymus is a huge genus, containing 300 to 400 species, most of which are aromatic shrubs or perennials. All are native to Asia or Europe, most probably to Southern Europe. Thyme is now cultivated all over the world and has become naturalised in some areas, including the northeastern US.
Thyme is an evergreen perennial shrub with slightly fuzzy gray-green leaves about a 1.5 cm long. The tiny, tubular, lilac flowers are arranged in whorled, compact heads. Thyme grows in a thick, spreading mound 15-30 cm high and up to twice as wide. As with most mints, the stem is square in cross-section, and the leaves are arranged in pairs opposite each other. Leaves are highly aromatic (reaching their peak just before plants flower) and are frequently used fresh or dried as a seasoning.
There is a great variety of cultivars. Aureus, for example, has golden leaves; Orange blossom has leaves that smell like oranges; the leaves of Silver posie have white margins.
Thyme is one of the classic herbs, often used in soups, stews, sauces, and meat and fish dishes. Cajun and Creole dishes are especially likely to contain Thyme. Thyme is also used to make herbal teas.
The main essential oil in Thyme, thymol, is active against salmonella and staphylococcus bacteria. Thyme and Thyme oil are ingredients in fumigants, antiseptics, disinfectants, and mouthwashes. Thymol is one of the active ingredients in Listerine® mouthwash and provides the "medicated" properties of many consumer products. Oil of Thyme also functions as a carminative and counterirritant. It features in a number of homeopathic and folk remedies (1).
Clinical Relevance
Allergen description
No allergens from this plant have yet been characterised.
Potential Cross Reactivity
An extensive cross-reactivity among the different individual species of the Lamiaceae (Labiatae) family, which comprises plants such as hyssop (Hyssopus officinalis), Basil (Ocimum basilicum), Marjoram (Origanum majorana), Mint (Mentha piperita), Sage (Salvia officinalis) and Lavender (Lavandula officinalis), could be expected (2-3).
Molecular Aspects
Clinical Experience
IgE-mediated reactions
Thyme may uncommonly induce symptoms of allergy in sensitised individuals. Although reactions may be uncommon or rare, they may also often be overlooked.
A 45-year-old man reported 3 reactions to food, as a result of Oregano on a single occasion, and twice to Thyme. He developed pruritis and swelling of the lips and tongue, dysphagia, dysphonia, and progressive upper respiratory difficulty as well as intense facial and palpebral oedema. On 2 occasions the reactions were severe, resulting in hypotension, vomiting, and nausea. His first adverse event occurred within minutes of the ingestion of pizza containing Oregano. The 2 adverse reactions to Thyme occurred after eating meat seasoned with Thyme, and Snails seasoned with Thyme. Skin-specific IgE was detected to a number of plants of the Labiatae family when the skin prick technique was used; when the prick-to-prick method was used, specific IgE was detected to members of the family with the exception of Basil and Lavender. Serum-specific IgE was detected to all herbs tested. The authors concluded that plants belonging to the Labiatae family show cross-sensitivity on the basis of clinical history and in vitro and in vivo test results (3).
Occupational asthma caused by working with a number of aromatic herbs, including Thyme, has been reported. The diagnosis was confirmed by inhalation challenges, and skin- and serum-specific IgE (4).
Other reactions
Similarly, among 46 farmers studied during the threshing of dried Thyme, 4 developed contact dermatitis after 5 to 30 minutes of exposure to Thyme dust. Thyme-specific IgE was found in 1 person with work-related symptoms, but also in 2 asymptomatic farmers. The authors proposed that an IgE mechanism may be questionable in eczema related to Thyme dust, that the aetiology of Thyme-related skin symptoms was obscure, and that an irritant mechanism seems probable (5).
A number of reports describe allergic contact dermatitis to Thyme oil or thymol. In a group of 100 patients with ulcus cruris, the incidence of contact allergies was determined with the use of patch tests for 18 products that might have come into contact with the skin. Balsam of Peru (14%), wool-wax alcohols (10%), para-aminobenzoate component (7%), neomycin (5%) and Thyme oil (5%) were the allergens most frequently encountered (6). Photoaggravated allergic contact dermatitis to Rosemary and Thyme was described in a 62-year-old woman who presented with several episodes of itchy hand, forearm and face dermatitis after picking Rosemary on sunny days. Skin-specific IgE testing with Rosemary leaves was initially negative, but a reaction was detected at the prick site 2 days later.
The patient was shown to be allergic to Thyme as well (7).
Allergic contact dermatitis due to thymol (a main component of Thyme oil) was reported in a 70-year-old woman 6 weeks after initiating treatment of 4% thymol once daily to a chronic paronychia (8); and similarly, in a report, pruritic contact dermatitis was noted to a topical application of the combination antiseptic solution Listerine to a chronic parenchyma of the toe. Patch testing with the individual ingredients revealed selective allergic hypersensitivity to thymol (9). Although massage with essential oils, including those manufactured from Thyme, may alleviate symptoms of eczema, individuals sensitised to Thyme may experience exacerbation of their eczema (10).
In a group of 47 Thyme farmers examined while threshing Thyme and exposed to organic dusts, 63.8% reported occurrence of work-related symptoms during this activity. The most common complaints were blocking of the nose (38.3%), dry cough (29.8%) and general weakness (27.7%). Lung function tests were normal and did not show a significant post-work decline. Positive skin hyperactivity to microbial antigens and positive precipitin reactions was demonstrated. The study concluded that Thyme farmers engaged in threshing of Thyme are at a greater risk for work-related symptoms as a result of associated organic dust (11-12). A number of molds has been associated with Thyme cleaning (13).
Allergic alveolitis was described in a female farmer as a result of massive exposure to organic dust contaminated with microorganisms during threshing of Thyme (14).
Compiled By
Last reviewed: June 2022.
References
  1. Basch E, Ulbricht C, Hammerness P, Bevins A, Sollars D. Thyme (Thymus vulgaris L.), thymol. J Herb Pharmcother 2004;4(1):49-67.
  2. Yman L. Botanical relations and immunological cross-reactions in pollen allergy. 2nd ed. Pharmacia Diagnostics AB. Uppsala. Sweden. 1982: ISBN 91-970475-09
  3. Benito M, Jorro G, Morales C, Pelaez A, Fernandez A. Labiatae allergy: systemic reactions due to ingestion of oregano and thyme. Ann Allergy 1996;76(5):416-8
  4. Lemiere C, Cartier A, Lehrer SB, Malo JL. Occupational asthma caused by aromatic herbs. Allergy 1996;51(9):647-9
  5. Spiewak R, Skorska C, Dutkiewicz J. Occupational airborne contact dermatitis caused by thyme dust. Contact Dermatitis 2001;44(4):235-9
  6. Le Roy R, Grosshans E, Foussereau J. Investigation of contact allergies in 100 cases of ulcus cruris (author's transl). [French] Derm Beruf Umwelt 1981;29(6):168-70
  7. Armisen M, Rodriguez V, Vidal C. Photoaggravated allergic contact dermatitis due to Rosmarinus officinalis cross-reactive with Thymus vulgaris. Contact Dermatitis 2003;48(1):52-3
  8. Lorenzi S, Placucci F, Vincenzi C, Bardazzi F, Tosti A. Allergic contact dermatitis due to thymol. Contact Dermatitis 1995 Dec;33(6):439-40
  9. Fisher AA. Allergic contact dermatitis due to thymol in Listerine for treatment of paronychia. Cutis 1989 Jun;43(6):531-2
  10. Anderson C, Lis-Balchin M, Kirk-Smith M. Evaluation of massage with essential oils on childhood atopic eczema. Phytother Res 2000;14(6):452-6
  11. Golec M, Skorska C, Mackiewicz B, Dutkiewicz J. Health effects of exposure to thyme dust in a group of thyme growing farmers. Ann Univ Mariae Curie Sklodowska [Med] 2003;58(1):195-203
  12. Golec M, Skorska C, Mackiewicz B, Dutkiewicz J. Immunologic reactivity to work-related airborne allergens in people occupationally exposed to dust from herbs. Ann Agric Environ Med 2004;11(1):121-7
  13. Krysinska-Traczyk E. Microflora of the farming work environment as an occupational risk factor. [Polish] Med Pr 2000;51(4):351-5
  14. Mackiewicz B, Skorska C, Dutkiewicz J, et al. Allergic alveolitis due to herb dust exposure. Ann Agric Environ Med 1999;6(2):167-70