Type:
Whole Allergen
Whole Allergen
Whole Allergen
Langust (spiny lobster)
f304
Palinuridae
Palinurus spp.
Langust (spiny lobster)
IgE antibodies to spiny lobster have been measured in both atopic and non-atopic patients
When searching for possible shared antigens in assessing crustacea sensitivity, IgE antibodies to spiny lobster were measured in both atopic and non-atopic patients (1). In a study of adult shrimp-hypersensitive patients, a common major allergen of 36kd was identified in shrimp (f24), crayfish, crab (f23), and spiny lobster, which may explain the clinical histories of patients with immediate hypersensitivity reactions to the crustacea family (2).
Langust has a major allergen of 36 kD in common with shrimp (f24), crayfish (Rf320) and crab (f23).Crossreactivity with other Crustaceans can also be expected.(2)
Last Reviewed- April 2022
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.