Type:
Whole Allergen
Whole Allergen
Whole Allergen
Hake
f307
Merlucciidae
Merluccius merluccius
Hake
XM95M8
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Hake is one of the most frequently consumed fish in Spain, and is closely related to cod. See also f3
In a study of Spanish children hake was shown to provoke the strongest IgE responses.
Hake is closely related to codfishes, belonging to the order Gadiformes, but has a special systematic position because of its distribution. The family has only one genus. It is distributed in the northeast Atlantic and the southwest coast of Europe. It lives in deep water and thus can travel back and forth between the hemispheres in the cooler waters. There are some species in the Pacific. Hake can be over 1 meter long but there are small and medium-sized species as well. Cannibalism is quite common, and a favorite food is squid.
Hake is one of the most frequently consumed fish in Spain. In a study in Spanish children, hake was shown to provoke the strongest IgE responses (1). When RAST® inhibition was performed with several fish species, it was shown that hake had allergens in common with other species (1). Gad c 1 was not an important allergen in hake despite the close relation to cod.
Species within groups of fish, like Gadiformes (examples: codfish and hake) and Scombroid fishes (examples: mackerel and tuna) seem to share allergenic components. The overlap of allergen specificity between the groups seems to be moderate or even small.
Last Reviewed : June 2022
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.