Type:
Whole Allergen
Whole Allergen
Whole Allergen
Sheep's milk
f325
Ovis spp.
Sheep's milk
XM8RU4
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Sheep's cheese, and other dairy products.
Sometimes included in infant formulas.
Sheep's cheese, and other dairy products
Sometimes included in infant formulas.
Sheep’s milk is sometimes recommended as a substitute for cow’s milk in atopic children. However, 15/16 cows´s milk allergic patients had IgE antibodies against sheep’s milk. Crossreactivity with cow’s milk was confirmed in RAST® inhibition (1). Serum albumin and caseins show homology to the corresponding proteins of cow and sheep (2). Milking sheep can cause IgE mediated respiratory problems in milk allergic individuals (3).
Crossreactivity between sheep's milk and cow's milk and whey might be expected.
Milk from other related animals.
Last Reviewed : June 2022
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.