Type:
Whole Allergen
Whole Allergen
Whole Allergen
Mutton
f88
Ovis spp.
Mutton
XE25P
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Mutton is regarded as a weak allergen and preparations of lamb meat have been suggested as an alternative to hydrolyzed cow's milk formulas
>Sheep and goats belong to Caprines of the order Artiodactyla (even-toed hoofed animals). The true sheep which resembles the domesticated animal is Ovis. Many breeds exist and a characteristic feature is the animal’s climbing ability, making it appropriate for grazing in mountainous areas. Lamb is less than one year. Mutton is the meat from animals more than one year old. Beef and mutton share allergens and it cannot be excluded that mutton has some allergens in common with sheep wool. Mutton is regarded as a weak allergen and preparations of lamb meat have been suggested as an alternative to hydrolyzed cow’s milk formulas (1). The allergenicity of the meat is decreased by heating and pepsin treatment (2, 3).
Last Reviewed- April 2022
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.