Whole Allergen
f94 Pear
f94 Pear Scientific Information
Type:
Whole Allergen
Display Name:
Pear
Allergen code:
f94
Family:
Rosaceae
Species:
communis/pyrifolia
Route of Exposure:
Ingestion, contact
Latin Name:
Pyrus communis; Pyrus pyrifolia /P. sinensis
Other Names:
European pear; Asian/Oriental pear
WHO/ICD-11 code:
XM4GG6
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Pear (Pyrus communis, P. pyrifolia/sinensis) is a fruit of the Rosaceae family and grown in many countries. There are many different cultivars, which vary in appearance. Peel and flesh of the fruit are the main sources of hypersensitivity and allergic reactions, though pollen may also be implicated in allergies. Several pear allergens have been described.
Pollen-food allergy syndrome (PFAS) and one of its common presentations, oral allergy syndrome (OAS) is a common clinical presentation of pear allergy, with symptoms ranging from localized to systemic, including anaphylaxis. Contact dermatitis and seasonal allergy may also be a clinical presentation of pear hypersensitivity. Cross reactivity with several other plant food and pollen allergens may occur.
Allergen
Nature
Pear is the fruit of the pear tree (European pear Pyrus communis; Oriental or Asian pear Pyrus pyrifolia/sinensis), which can be found worldwide (1-3). All species of Pyrus are able to hybridize (4).
The leaves are simple, glossy green or silvery and hairy depending on the species. The tree produces a variable number of flowers, usually white in color, grouped in corymbs, a type of flower cluster (5). The pear fruit is actually a pseudo-fruit, composed of the greatly enlarged calyx which encloses the true fruit (normally called “core”) where the seeds are found (5). Due to the many cultivars and the ability to hybridize, the shape, size, flavor and texture of the edible fruit varies (5). The flesh is firm prior to ripening, becoming soft as it ripens (6), maintaining a degree of grittiness (due to the presence of stone cells – sclerenchyma cells) (7).
The pear plant and fruit are versatile; the fruit is commonly consumed fresh or preserved, canned as slices or in other forms (2, 3, 7).
Taxonomy
Taxonomic tree of pear Pyrus communis (2, 3)
Domain
Eukaryota
Kingdom
Plantae
Phylum
Spermatophyta
Subphylum
Angiospermae
Class
Dicotyledonae
Order
Rosales
Family
Rosaceae
Genus
Pyrus
Tissue 
The main tissues capable of causing allergic reactions are the peel and flesh of the fruit (8, 9)
Epidemiology
Worldwide distribution 
Pear allergy has been reported in a number of countries, including Japan (10), The Netherlands (8), China (11), and Spain (9).
Risk factors 
Certain occupations handling pears may pose risk of contact dermatitis (12).
Consumption of unpeeled pear may be a trigger for allergic reactions (9).
For patients suffering from pollen-food allergy syndrome (PFAS), the season of pollination of the plants they are allergic to is more likely to trigger food allergy symptoms (13). In specialized orchards, where pollination is carried out by operators, exposure to pear pollen (in this case, P. pyrifolia) is able to cause sensitization and symptoms of seasonal allergies (10).
Environmental Characteristics
Worldwide distribution 
Pear trees have worldwide distribution, preferring temperate climates (4, 14).
Route of Exposure
Main
Ingestion of the fruit (8), peel (9), contact with pear proteins (12).
Clinical Relevance
Allergic rhinitis
Chronic rhinosinusitis with nasal polyps (CRSwNP) could be associated with food allergies; both asthma and allergic rhinitis are often comorbidities in people with food allergies, due to cross-reactivity between aeroallergens. A study showed that in patients with CRSwNP, IgG to certain types of foods (including pears and apples) were significantly reduced, irrespective of consumption. The authors suggested this might be due to IgG responses deviated towards other allergens (15).
Atopic Dermatitis
Contact dermatitis from pear proteins, at times also causing urticaria (12).
Other diseases
OAS (considered a manifestation of pollen-food allergy syndrome), consisting of IgE-mediated allergy symptoms limited to the oral cavity, is another presentation of pear allergy. The symptoms can start within minutes of consuming the sensitizing food. Clinical presentation is variable; normally limited to oral cavity with itching/burning sensation of lips, mouth, ear and throat, perioral erythema, urticaria, anaphylaxis. There may be swelling in the face, a sense of suffocation, hypotension and digestive symptoms (13). Another study using two varieties of pear in a single-blind oral challenge study reported symptoms of itchy mouth, itchy nose and ears, nausea, wheezing, skin and laryngeal symptoms (8).
In patients that have no history of pollen allergies, anaphylaxis is a severe presentation of food allergy to Rosacea fruits, including pear; 82% of patients reported systemic symptoms, with anaphylactic shock recorded in 36% of patients (16).
Diagnostics Sensitization
Testing for food allergies involves measuring serum specific IgE responses, skin prick tests (SPTs) and oral challenges (13, 17, 18). A Multiple Allergen Simultaneous Test-Chemiluminescent Assay (MAST-CLA), measuring specific IgE, may also be used to diagnose hypersensitivity to Rosacea plant foods, although its usefulness regarding pear hypersensitivity is lower than for apple (19).
In a study of 36 patients with symptoms of OAS following ingestion of Rosacea fruits, half were split into the birch pollen-allergic group, and the other half to non-allergic. It was shown that for the birch pollen-allergic patients, SPTs were a reliable method to diagnose fruit hypersensitivity. However, for those patients not allergic to birch pollen the SPTs are best carried out with fruit peel extracts (17).
In another study which involved patients with known pollen allergies, pear was the third most common Rosacea fruit causing hypersensitivity, with 26.6% of positive SPTs (19). A similar study on 129 patients, 83 of which with PFAS and the remaining suffering from seasonal pollen allergies without food component, showed that pear was again the third most common fruit reported as cause of hypersensitivity reactions (11).
A blind oral challenge was carried out in 15 birch pollen-allergic patients with a known history of pear allergy; in this study, two different pear varieties were used. All patients were sensitized to one or both pear varieties, as shown by specific IgE testing, SPT, prick-to-prick (PTP) tests and the oral challenge. Analyzing the pattern of responses to the two varieties, the study concluded that for some patients with pear hypersensitivity it may be possible to consume certain varieties outside of the birch pollination season (8).
Molecular Aspects
Allergenic molecules
Table adapted from Allergome.org (20).
Allergen
Type
Mass (kDa)
Pyr c 1 [fruit]
Ribonuclease
17.5
Pyr c 3 [fruit]
Lipid transfer protein
9.5
Pyr c 4 [fruit]
Profilin; actin binding protein
14
Pyr c 5 [fruit]
Isoflavone reductase
34
Pyr py 2 [fruit]
Thaumatin
26
A number of pear allergenic antigens have been identified. Pyr c 1 is a homolog to Bet v 1 (Jong 2021), a PR-10 type of allergen (21, 22). Pyr c 3 is a pear LTP, and it was shown to be mostly confined to the peel (22, 23). IgE tests showed significant binding to this allergen (Ramazzina 2012). Lipid transfer proteins (LTPs) are considered major Rosacea allergens in patients not allergic to birch pollen (17). The pear profilin Pyr c 4 (22) was recognized by 88% of IgE samples specific for birch pollen profilin (24). An isoflavone reductase, named Pyr c 5, showed allergenic activity in some patients allergic to pears (22).
The antigens Ole e 3 and Ole e 7 from Olea europaea (olive) showed significant association with Rosacea fruits in patients with known positive SPTs to olive allergens (18).
Biomarkers of severity
A study using radioallergosorbent test (RAST) suggested that due to variable allergens concentrations within the fruits, and because the allergens localized mostly in the peel, pear was the least allergenic of the 3 Rosacea fruits studied (apple, peach and pear) (16). In patients with pear and apple allergy, 40% were able to tolerate ingestion of pulp alone, with adverse reactions developing when the whole fruit was consumed. The severity of adverse reactions increased with consumption of the whole fruit, compared to pulp alone (9).
Cross-reactivity
Cross-reactivity between members of the Rosaceae family has been reported. Apple, peach and pear share cross-allergenic components (16). Birch pollen contains allergens cross reactive with Rosacea plants (25); the allergen Bet v 1 is related to the pear Pyr c 1 (8, 21). Pyr c 1 is also related to Mal d 1 from apple (21). Pyr c 4 (profilin) showed a strong cross-reactivity with birch pollen antigen Bet v 2 (24) and with mango profilin (26). The peach skin LTP allergen Pru p 3 may cause sensitization to the pear homolog (22).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer 
Last reviewed:May 2022
References
  1. Ghazouani T, Talbi W, Sassi CB, Fattouch S. Chapter 41 - Pears. In: Jaiswal AK, editor. Nutritional Composition and Antioxidant Properties of Fruits and Vegetables: Academic Press; 2020. p. 671-80.
  2. CABI. Pyrus communis (European pear) 2019 [cited 2022 11.01.22]. Available from: https://www.cabi.org/isc/datasheet/46190.
  3. CABI. Pyrus pyrifolia (Oriental pear tree) 2019 [cited 2022 11.01.22]. Available from: https://www.cabi.org/isc/datasheet/46199.
  4. Li X, Li X, Wang T, Gao W. Chapter 24 - Nutritional Composition of Pear Cultivars (Pyrus spp.). In: Simmonds MSJ, Preedy VR, editors. Nutritional Composition of Fruit Cultivars. San Diego: Academic Press; 2016. p. 573-608.
  5. Quinet M, Wesel J-P. Botany and Taxonomy of Pear. 2019. p. 1-33.
  6. Kojima T, Fujita S, Tanaka M, Sirisomboon P. 11 - Plant compounds and fruit texture: the case of pear. In: Kilcast D, editor. Texture in Food: Woodhead Publishing; 2004. p. 259-94.
  7. 2 - Canning of fruit. In: Featherstone S, editor. A Complete Course in Canning and Related Processes (Fourteenth Edition): Woodhead Publishing; 2016. p. 85-134.
  8. de Jong NW, Terlouw S, van Boven FE, van Maaren MS, Schreurs MWJ, van den Berg-Somhorst DBPM, et al. Birch Pollen Related Pear Allergy: A Single-Blind Oral Challenge TRIAL with 2 Pear Cultivars. Nutrients. 2021;13(4):1355.
  9. Fernández-Rivas, Cuevas. Peels of Rosaceae fruits have a higher allergenicity than pulps. Clinical & Experimental Allergy. 1999;29(9):1239-47.
  10. Teranishi H, Uchida M, Hayashi S, Yamada N. Allergenic pollens and spores in the working environment of Japanese pear farmers. Int J Immunopathol Pharmacol. 2007;20(2 Suppl 2):65-7.
  11. Ma S, Wang R, Nie L, Yin J. Pollen-food allergy syndrome in China. Food and Agricultural Immunology. 2018;29(1):281-93.
  12. Amaro C, Goossens A. Immunological occupational contact urticaria and contact dermatitis from proteins: a review. Contact Dermatitis. 2008;58(2):67-75.
  13. Kelava N, Lugovic-Mihic L, Duvančić T, Romic R, Situm M. Oral allergy syndrome--the need of a multidisciplinary approach. Acta clinica Croatica. 2014;53:210-9.
  14. Mir H, Patel VB. Chapter 5 - Genetic Engineering of Temperate Fruit Crops. In: Rout GR, Peter KV, editors. Genetic Engineering of Horticultural Crops: Academic Press; 2018. p. 89-119.
  15. Veloso-Teles R, Cerejeira R, Rodrigues D, Roque-Farinha R, von Buchwald C. Food-Specific IgE and IgG Antibodies in Patients With Chronic Rhinosinusitis With Nasal Polyps: A Case-Control Study. Ear Nose Throat J. 2021;100(3):177-84.
  16. Fernández-Rivas M, van Ree R, Cuevas M. Allergy to Rosaceae fruits without related pollinosis. J Allergy Clin Immunol. 1997;100(6 Pt 1):728-33.
  17. Asero R, Mistrello G, Roncarolo D, Casarini M, Falagiani P. Allergy to nonspecific lipid transfer proteins in Rosaceae: a comparative study of different in vivo diagnostic methods. Annals of Allergy, Asthma & Immunology. 2001;87(1):68-71.
  18. Florido Lopez JF, Quiralte Enriquez J, Arias de Saavedra Alías JM, Saenz de San Pedro B, Martin Casañez E. An allergen from Olea europaea pollen (Ole e 7) is associated with plant-derived food anaphylaxis. Allergy. 2002;57(s71):53-9.
  19. Barajas M. 431 Frequency and Characterization of Oral Allergy Syndrome in Mexican Adults with Nasal Pollinosis. World Allergy Organization Journal. 2012;5:S137.
  20. Allergome.org. Pear 2021 [cited 2022 11.01.22]. Available from: http://www.allergome.org/script/search_step2.php.
  21. Karamloo F, Scheurer S, Wangorsch A, May S, Haustein D, Vieths S. Pyr c 1, the major allergen from pear (Pyrus communis), is a new member of the Bet v 1 allergen family. Journal of Chromatography B: Biomedical Sciences and Applications. 2001;756(1):281-93.
  22. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, et al. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016;27 Suppl 23:1-250.
  23. Ramazzina I, Amato S, Passera E, Sforza S, Mistrello G, Berni R, et al. Isoform identification, recombinant production and characterization of the allergen lipid transfer protein 1 from pear (Pyr c 3). Gene. 2012;491(2):173-81.
  24. Scheurer S, Wangorsch A, Nerkamp J, Skov PS, Ballmer-Weber B, Wüthrich B, et al. Cross-reactivity within the profilin panallergen family investigated by comparison of recombinant profilins from pear (Pyr c 4), cherry (Pru av 4) and celery (Api g 4) with birch pollen profilin Bet v 2. J Chromatogr B Biomed Sci Appl. 2001;756(1-2):315-25.
  25. Asero R. Fennel, cucumber, and melon allergy successfully treated with pollen-specific injection immunotherapy. Annals of Allergy, Asthma & Immunology. 2000;84(4):460-2.
  26. Ukleja-Sokołowska N, Gawrońska-Ukleja E, Lis K, Żbikowska-Gotz M, Sokołowski Ł, Bartuzi Z. Anaphylactic reaction in patient allergic to mango. Allergy, Asthma & Clinical Immunology. 2018;14(1):78.