Type:
Whole Allergen
Whole Allergen
Whole Allergen
Cultivated wheat
g15
Poaceae
Triticum aestivum
Inhalation (1)
Triticum aestivum
Bread wheat (1)
Triticum aestivum is a member of the Poaceae family that produces wheat aeroallergens responsible for pollen allergy and occupational respiratory allergy to cereal flour, known as ‘Baker’s asthma’. Sensitization to cultivated wheat causes rhinitis and asthma and is prevalent among patients with respiratory allergies who are multi-sensitized to aeroallergens.
T. aestivum is a self-pollinating (1) cultivated wheat species that is grown in spring and winter for cereal grain (2). It is the main cereal used to produce human and animal food (2).
Cultivated cereals are widely grown throughout the world (3). They mainly flower in spring and produce large amounts of pollen, approximately 40–122 μm in size (1). However, although weather can affect pollen dispersion, the large size of the pollen prohibits dispersion more than 0.5km (1). Exposure to T. aestivum allergens is, thus, generally limited to occupational allergy or to the immediate vicinity of T. aestivum cultivation (1).
Taxonomic tree of Triticum aestivum (3) |
|
Domain |
Eukaryota |
Kingdom |
Plantae |
Phylum |
Spermatophyta |
Subphylum |
Angiospermae |
Class |
Monocotyledonae |
Order |
Cyperales |
Family |
Poaceae |
Genus |
Triticum |
Pollen and cereal flour (1).
Sensitization to cultivated wheat is common in people with respiratory allergies who are multi-sensitized to aeroallergens (1). Sensitization is thought to affect males more than females (Moradi et al, 2018) and a wide range of sensitization frequency (17.7–60.8%) has been reported in studies; cross-reactivity with timothy grass may account for the high percentage of sensitization levels seen (1).
Baker's asthma, an occupational respiratory allergy to cereal flour, affects up to 9% of bakers; it is the leading cause of occupational asthma in France, and the second highest in the UK (4). Among bakers with occupational asthma, 47% are sensitized to the T. aestivum allergen 25 (Tri a 25) (5).
Patients with baker's asthma usually ingest wheat products without issue, although in a study of children with grass pollen allergy, up to 65% had false positive results for wheat extract (6).
Living or working close to T. aestivum cultivation (1).
Occupations that involve working with cereal flours, such as bakers, farmers (1), confectioners, flour millers, and food processors (4).
T. aestivum can be found worldwide (3) and are well adapted to temperate regions (1).
Inhalation (1).
Contact (6).
Symptoms of cultivated wheat sensitization include rhinitis, conjunctivitis, urticaria, asthma and, rarely, systemic anaphylaxis (4).
A number of wheat allergens have been identified; some are involved in the development of respiratory allergies and others in the development of wheat food allergy (7).
In a study that investigated the IgE reactivity of different wheat recombinant allergens using serum from 22 patients that suffered from wheat respiratory allergy, the results showed that 91% of sera recognized wheat CAP. The six individual wheat recombinant allergens (thioredoxin H, serine proteinase inhibitor, glutathione transferase, profilin, dehydrin and 1-cis-peroxyredoxine) were recognized between 4.5 and 32% of serum samples (7). Profilin is a common cross-reactive allergen that is present in wheat seeds, wheat pollen and other grass pollens including timothy (7). The recombinant versions of the timothy grass allergens Phl p 1 and Phl p 5 were identified as marker allergens specific for grass pollen allergy (7).
This table, adapted from Allergome.org (8), shows allergens identified in wheat pollen.
Allergen |
Type |
Mass (kDa) |
Tri a 1 |
β-expansin |
30 |
Tri a 2 |
Pollen allergen II |
13.3 |
Tri a 3 |
Unknown |
13 |
Tri a 4 |
57.5 |
|
Tri a 5 |
24 |
|
Tri a 7 |
n/a |
|
Tri a 12 |
Profilin |
14 |
Tri a 13 |
- |
Cross-reactivity has been reported between wild grass and cereal pollen; cultivated maize, rice, wheat, rye, barley and oats, can induce symptoms in individuals sensitized to grass pollen (1). Cross-reactivity with common grass species (Anthoxanthum odoratum, Dactylis glomerata, Lolium perenne, Poapratensis and Phleum pratense) has been shown (9).
The Zea mays allergen 25 (Zea m 25) exhibits high IgE cross-reactivity with the T. aestivum allergen 25 (Tri a 25) (Weichel et al, 2006) while rice pollen shows weak cross-reactivity with wheat (1).
Profilin is recognized as a cross-reactive allergen in patients with baker’s asthma, food and pollen allergy (7).
Author: RubyDuke Communications
Reviewer: Dr.Christian Fischer
Last reviewed: June 2022