Table of Contents
Component
g205 Phl p 1
g205 Phl p 1 Scientific Information
Type:
Component
Name; WHO/IUIS:
Phl p 1
Biological function:
b-Expansin
Allergen code:
g205
Native Recombinant:
r Phl p 1 is a cross-reactive carbohydrate determinants (CCD)-free recombinant protein
Molecular Weight:
30 kDa
Route of Exposure:
Airway
Summary
Phl p 1 allergen derived from timothy grass (Phelum pratanse) pollen is one of the distinguished group 1 grass pollen allergens (1, 2). Group 1-grass pollen allergens are glycosylated proteins with a molecular weight of ~30 kDa that appear as cross-reactive antigens in majority of the grasses and corn species (3). Group 1 grass pollen allergen belongs to the subclass beta-expansin (EXP-B) of the expansin protein superfamily, which is involved in cell wall loosening and extension in the plants (4).
Phl p 1, the major allergen of timothy grass is identified by more than 90% of grass pollen-allergic patients. This allergen is highly cross-reactive among all the grass species of temperate and tropical regions (1, 5) and shows 90% sequence identity with allergens of other members of the Pooideae subfamily (5). A major IgE-reactive segment of Phl p 1 allergen reveals a substantial sequence identity of about 40% with the family of immunoglobulin domain-like group 2/3 grass pollen allergens (3).
Phl p 1 initiates sensitization in majority of the pediatric allergic rhinitis (AR) patients, followed by sensitization of these patients to other timothy grass pollen allergens (such as Phl p 5, Phl p 4, Phl p 2, and Phl p 6, and Phl p12) (6).
Recombinant pollen allergens aid in lowering the number of allergens required for appropriate diagnosis of grass pollen allergy.  Only a limited number of recombinant timothy grass pollen allergens results in the determination of a high percentage of patients with grass pollen-specific IgE. This  indicates the benefits of recombinant allergens not only for in vitro diagnosis but also to evaluate the specific percentage of IgE produced by individuals against some allergens (7). 
Epidemiology
Worldwide distribution
Phl p 1 allergen is found in grass species of both temperate and tropical regions of the world (2). A study using sera from European, Asian, and American subjects assessed the percentage of IgE directed to pollen extracts from nine different monocot species including rPhl p 1, rPhl p 2, rPhlp 5 of timothy grass and rBet v 2 of birch pollen. The study results showed that IgE antibodies to these four recombinant pollen allergens accounted for a mean of 59% (8). A study from China detected that out of all timothy grass serum(s) IgE positive patients only 17.1% were positive to the antigen Phl p 1 and 100% were positive to Phl p 4 (9). A longitudinal birth cohort study in Germany investigated the evolution of IgE sensitization to P. pratense allergenic molecules in children suffering from seasonal AR due to grass pollens. The study detected Phl p1 initiated sensitization in more than 75% of these patients followed by sensitization to other allergens. Moreover, Phl p 1, Phl p 5, and Phl p 4 were the most frequently identified molecules in the preclinical sensitization phase (6).
Environmental Characteristics
Source and tissue
All grass species are pollinated by wind and during the peak flowering season they release allergenic pollen grains (5). The allergenic glycoprotein Phl p 1 has been found to be localized in the cytoplasm and the outer layer (exine) of mature pollen grains of grasses (10, 11) and these allergens are released from the pollen grain on hydration (5). 
Figure 1: Timothy grass (P. pratense) major pollen allergen Phl p 1 (12)
Figure 2: Timothy grass (P. pratense) within pollen grains by fluorescence microscopy and after immunogold silver staining at the light microscopic. (12)          
Risk factors
Röschmann et al. (2011) studied the response of airway epithelium to Phl p 1, with an aim to obtain an insight into the immunostimulatory capacity of Phl p 1. They observed that Phl p 1 triggers the release of a wide range of mediators (IL-6, IL-8, TGF-β), which in turn attract and influence the immunocompetent cells’ activities. Hence, Phl p 1 is a powerful inducer of the immune system and linked with allergic respiratory diseases, especially AR (13) .
Clinical Relevance
Specific molecules
Recombinant Phl p 1 (rPhl p 1) is a non-glycosylated protein produced in an Escherichia coli (E. coli) strain that carry a cloned cDNA encoding P. pratense allergen Phl p 1. rPhl p 1 closely resembles the natural Phl p 1 and binds to IgE in majority of the patients with grass pollen allergy, indicating that rPhl p 1 shares many of the IgE epitopes with natural group 1 grass pollen allergens (10). rPhl p 1 is considered to be one of the most important “species specific allergens” of timothy grass followed by rPhl p 5 (14). Scaparrotta and colleagues (2013) detected rPhl p1 to be the predominant sensitizing allergen at all ages of pediatric population and at all levels of timothy grass pollen specific IgE molecules (14). Another study assessing specific IgE levels to the clinically relevant timothy grass pollen allergens found the level of Phl p 1 to be significantly higher (p=0.02) in children with moderate AR than in children with mild AR indicating the ability of Phl p 1 to induce allergic symptoms in AR patients (15).
Douladaris et al. (2019) constructed a recombinant hybrid molecule with four major timothy grass pollen allergens, Phl p 1, Phl p 2, Phl p 3, and Phl p 6 for in vitro serological testing and in vivo diagnosis of grass pollen allergy in adult patients. The success of this hybrid molecule suggested that all grass pollen allergen patients showed IgE reactivity and also positive skin reaction to this molecule (16).
Cross-reactive molecules
IgE antibodies from grass pollen allergic patients and also group 1 specific antibodies are found to cross-react with natural group 1 grass pollen allergens of different grass species (10). rPhl p 1 has been shown to inhibit IgE antibody binding to most of the group 1 isoallergens from 7 to 8 grass species indicating wide-spread cross reactivity between different grass species (7, 10). Hence, a single recombinant group 1 allergen contains many of the IgE epitopes of group 1 isoallergenic variants from several different grass species (10). The major timothy grass pollen allergen Phl p 1 shows cross-reactivity with most grass, corn, and monocot-derived group 1 allergens (17).
Molecular Aspects
Biochemistry
Recombinant Phl p1 protein of timothy grass has a molecular weight of 27 kDa (18). Unlike the native Phl p 1, which has N-glycosylation and contains glycans (11), the recombinant allergen is unglycosylated. Phl p 1 contains 7 cysteine residues (3). Phl p 1 are 31-35 kDa glycoproteins, belongs to EXPB protein family, and help in pollen tube growth (4).
Isoforms, epitopes, and antibodies
Phl p1 allergen has two isoforms of molecular weights of 37 kDa and 38 kDa (19). Each isoform of Phl p 1 has two isoallergens (Phl p 1.0101 and Phl p 1.0102) (14).
Phl p 1 consists of the majority of group 1-specific T- cell epitopes (1). The analysis of Phl p 1 IgE epitopes using rPhl p1 fragments revealed the presence of multiple continuous IgE epitopes. The prominent one being present in the C-terminal end of Phl p 1 and this intact folded Phl p1 domain indicates the primary targets of patients’ IgE antibodies (3). A study by Madritsch et al. (2015) determined three independently accessible IgE epitopes available on the surface of Phl p 1 (1).
Phl p 1 contains most of the group 1-specific IgE. More than 90% of the grass pollen allergic patients  show IgE reactivity to group 1 allergen and elicit most of the allergic symptoms (1, 16). 
Cross reactivity
Phl p 1 allergens are extremely cross-reactive allergens in majority of the grass species of temperate and tropical regions (1).
Diagnostic Relevance
AIT Prescription
Allergic Immunotherapy (AIT)
The use of allergen immunotherapy (AIT) have increased owing to rise in the number of patients with respiratory allergic disease and have shown long-term benefits (20). The development of recombinant allergens (such as rPhl p1) helps to develop patient-tailored immunotherapy (7).
Exposure
The major grass pollen allergens, such as Phl p 1 are usually present within the pollen grain in a typical pattern and at the time of activation, allergen moves from the exine of the pollen grain to the surface in order to be released (5, 11, 12). Activation of the pollen grain leads to allergen release (normally under humid condition) at the mucosal surface of the nose and triggers allergic reactions (12).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Christian Fischer
Last reviewed: October  2020
References
  1. Madritsch C., Gadermaier E., Roder U.W., al. e. High-Density IgE Recognition of the Major Grass Pollen Allergen Phl p 1 Revealed with Single-Chain IgE Antibody Fragments Obtained by Combinatorial Cloning. J Immunol. 2015;194(5):2069-78.
  2. Ghunaim N., Grçnlund H., Kronqvist M., al. e. Antibody profiles and self-reported symptoms to pollen-related food allergens in grass pollen-allergic patients from northern Europe. Allergy. 2005;60:185-91.
  3. Ball T., Edstorm W., Mauch L., al. e. Gain of structure and IgE epitopes by eukaryotic expression of the major Timothy grass pollen allergen, Phl p 1. FEBS. 2004;272(1).
  4. Aud-in S., Somkid K., W. S. Group-1 Grass Pollen Allergens with Near-Identical Sequences Identified in Species of Subtropical Grasses Commonly Found in Southeast Asia. Medicina. 2019;55(193):1-10.
  5. Gangi K., Niederberger V., R. V. Multiple grass mixes as opposed to single grasses for allergen immunotherapy in allergic rhinitis. Clin Exp Allergy. 2013;43(11):1202-16.
  6. Hatzler L., Panetta V., Lau S., al. e. Molecular spreading and predictive value of preclinical IgE response to Phleum pratense in children with hay fever. J Allergy Clin Immunol. 2012;130:894-901.
  7. Laffer S., Vrtala S., Duchene M., al. e. IgE-binding capacity of recombinant timothy grass (Phleum pretense) pollen allergens. J Allergy Clin Immunol. 1994;94:88-94.
  8. Niederberger V., Laffer S., Froschi R., al. e. IgE antibodies to recombinant pollen allergens (Phl p 1, Phl p 2, Phl p 5, and Bet v 2) account for a high percentage of grass pollen–specific IgE. J Allergy Clin Immunol. 1998;101:258-64.
  9. Luo W., Huang H., Zheng P., al. e. Major grass pollen allergens and components detected in a southernChinese cohort of patients with allergic rhinitis and/or asthmaWenting. Mol Immunol. 2016;78:105-12.
  10. Laffer S., Duchene M., Reimitzer I., al. e. COMMON IgE-EPITOPES OF RECOMBINANT Phl p I, THE MAJOR TIMOTHY GRASS POLLEN ALLERGEN AND NATURAL GROUP I GRASS POLLEN ISOALLERGENS. Mol Immunol. 1996;33(4/5):417-26.
  11. Andersson K., J. L. Characteristics and Immunobiology of Grass Pollen Allergens. Int Arch Allergy Immunol. 2003;130:87-107.
  12. Behrendt H., Tomczok J., Silwa-Tomczok W., al. e. Timothy Grass (Phleum pratense L.) Pollen as Allergen Carriers and Initiators of an Allergic Response. Int Arch Allergy Immunol. 1999;118:414-8.
  13. Röschmann KIL., van Kuijen AM., Jonker MJ, al. e. Purified Timothy grass pollen major allergen Phl p 1 may contribute to the modulation of allergic responses through a pleiotropic induction of cytokines and chemokines from airway epithelial cellscei_4522 413..421. Clin Exp Immunol. 2011;167:413–21.
  14. Scaparrotta A., Verini M., N.P. C, al. e. Sensitization to timothy grass pollen allergenic molecules in children. Resp Med. 2013;8(17):1-8.
  15. Douladiris N., Garob V., Piskou K., al. e. Molecular allergy diagnosis: A potential tool for the assessment of severity of grass pollen‐induced rhinitis in children. 2019.
  16. Douladiris N., Garib V., Focke-tejkl M., al. e. Detection of genuine grass pollen sensitization in children by skin testing with a recombinant grass pollen hybrid. Pediatr Allergy Immunol. 2019;30:59-65.
  17. Focke M., Mahler V., Ball T., al. e. Nonanaphylactic synthetic peptides derived from B cell epitopes of the major grass pollen allergen, Phl p 1, for allergy vaccination. FASEB J. 2001;15(11).
  18. Sub-Committee WIAN. Allergen Nomeclature. Available at: http://www.allergen.org/viewallergen.php?aid=503. Accessed on 10th August 2020.
  19. Becker W.M., Bufe A., A. P. Molecular characterization Molecular characterization of timothy grass pollen group V allergens. Int Arch Allergy Immunol. 1995;107:242-4.
  20. Scaparrotta A., Attanasi M., Petrosino M.I., al. e. Critical appraisal of Timothy grass pollen extract GRAZAX® in the management of allergic rhinitis. Drug Design, Devel Ther. 2015;9:5897-909.