Whole Allergen
i3 Common wasp (Yellow jacket)
i3 Common wasp (Yellow jacket) Scientific Information
Type:
Whole Allergen
Display Name:
Common wasp (Yellow jacket)
Allergen code:
i3
Family:
Vespidae
Species:
V. vulgaris
Route of Exposure:
Injection (sting)
Latin Name:
Vespula spp.
Other Names:
Common wasp, Yellow jacket
WHO/ICD-11 code:
XM6D92
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Hymenoptera of the Vespula genus are social wasps native to the Northern hemisphere, secondarily established in Australia, New Zealand, and South Africa.
Hymenoptera stings cause 48% of severe anaphylactic reactions occurring in European adults, and 20% of those occurring in children. In the United States, the prevalence of Hymenoptera-induced anaphylaxis is estimated at 3% in adults and 1% in children, with 40 to 100 Hymenoptera sting-induced fatalities being documented annually. Anaphylaxis is more common in adults than in children. Systemic reactions usually occur within minutes of being stung. The risk of repeated anaphylaxis is 30% to 70%. An estimated 9 to 42% of the general population is sensitized to Hymenoptera venom. Beekeepers, greenhouse workers, and rural populations are at higher risk of developing bee sting allergy. Mast cell disorders including hereditary α-tryptasemia, elevated baseline serum tryptase, or a family history of honeybee allergy are associated with an increased risk of occurrence and severity of Hymenoptera sting-induced reactions. A history of Hymenoptera-induced anaphylaxis is a red flag for an underlying clonal mast cell disorder.
Six molecular allergens have been characterized so far in the Vespula spp venom, of which two, Ves v 1 and Ves v 5, are available for in vitro diagnosis. Ves v 1 and Ves v 5 are marker allergens for genuine sensitization to Vespid venom.
Allergen
Nature
Vespula spp venom consists of a complex mixture of allergenic and non-allergenic molecules contained in the venom sac at the distal extremity of the insect’s abdomen. Similar to most Hymenoptera, Vespula spp can extract their stringer from the victim and are thus able to sting multiple times during their lifetime.
Taxonomy
Taxonomic tree of Vespula spp.
Domain Eukaryota
Kingdom Metazoa (Animalia)
Phylum Arthropoda
Subphylum Hexapoda
Class Insecta
Order Hymenoptera
Suborder Apocrita
Family Vespidae
Genus Vespula
Species V. vulgaris, V. germanica, V. pensylvanica, V. maculifrons, V. flavopilosa
The order of Hymenoptera comprises the families Vespids (wasps and hornets), Apids (bees and bumblebees) and Formicids (stinging ants) (1). The two Vespid subfamilies are Vespinae, with genera Vespula, Dolichovespula, and Vespa, and Polistinae, with Polistes and Polybia. Vespula (known as wasps in Europe and yellow jackets in the USA) are considered to be the most important species in Europe (2).
Tissue
Wasp venoms contain three major molecule groups: proteins such as allergens and enzymes; small peptides with neuroactive and antimicrobial activities; and substances of low molecular weight including bioactive amines (2). Peptides and proteins in wasp venom can be grouped according to their activity, for example mastoparans, chemotactic peptides, wasp kinins and enzymes (3). 
Epidemiology
Worldwide distribution
Hymenoptera stings cause 48% of severe anaphylactic reactions occurring in European adults, and 20% of those occurring in children (2).
It is estimated that the worldwide annual incidence of immunologic reactions to Hymenoptera stings ranges from 0.3–3.0% which equates to almost 100 million cases per year. Severity ranges from  local wheal-and-flare reactions to death from anaphylactic shock (4).
In the US, Hymenoptera-induced systemic reactions are estimated to occur in 3% of adults and 1% of children, and approximately 40 to 100 fatalities are reported each year, although the figure is likely to be higher (5). An Australian study reported honeybee to be the main cause of fatal insect venom anaphylaxis, however, there is a relatively high prevalence of honeybee allergy in Australia, and UK and European studies found that wasp was the most common cause of fatal and nonfatal venom anaphylaxis, respectively (6). A consistent finding across studies in Australia, Canada, UK and US was that fatal insect venom anaphylaxis occurs at an approximate rate of 0.1 cases per million population (6). In Europe, the prevalence of systemic reactions to Hymenoptera stings is 0.3% to 7.5% (7, 8).[JV1]
Risk factors
Identifying patients at risk for severe reactions to Hymenoptera venom requires a careful record of clinical history and a stepwise procedure in the use of diagnostic tests  (9). The severity of past reactions to Hymenoptera stings is the best predictor of the severity of recurrent reactions, while the most significant risk factor for severe reactions is an underlying mast cell disorder (9).  
The prevalence and severity of Hymenoptera venom reactions are increased in patients with mast cell disorders including hereditary α-tryptasemia, with or without an elevated baseline serum tryptase concentration  (10-13). Hymenoptera venom allergy was observed in 50% of patients with systemic mastocytosis without hereditary α-tryptasemia and in 82% of those with concurrent hereditary α-tryptasemia (11).
Cardiovascular risk factors, male gender and older age have also been associated with an increased risk of severe reactions to Hymenoptera venom (14). On the other hand, sensitization to Hymenoptera venom is frequent, estimated at 9.2% to 42% of the adult population, and comprises a majority of asymptomatic individuals (2).
Pediatric issues
The prevalence of systemic reactions to Hymenoptera venoms is estimated at 3.4% in children (15). In children younger than 16 years experiencing a cutaneous reaction to Hymenoptera venom, the chance of anaphylaxis if re-stung is lower than 3% (5).
Environmental Characteristics
Living environment
The nests of Vespula species are usually found in hidden locations, for example in building frames, shutters and wall cracks, and consist of paper-like or wood pulp material (5, 16). Vespids are known to be aggressive in defense of their nests, with the common wasp being particularly aggressive and easily provoked (5, 16).
Worldwide distribution
V. vulgaris and V. germanica are native Eurasian members of the Vespula genus, while V. pensylvanica originates from North America, however, Vespula species have been established worldwide, in South America, Australia and New Zealand (17). 
Route of Exposure
Main
Exposure to Vespula spp venom occurs through a sting when the insect’s stinger becomes embedded in the flesh and the venom is injected from the venom sac. Wasps, hornets and yellow jackets are able to stings several times without dying, as they can extract their stinger from the victim (2). 
Clinical Relevance
Five types of reactions to Hymenoptera stings are recognized: normal local reactions, large local reactions (LLR), systemic anaphylactic reactions, systemic toxic reactions, and unusual reactions. Of these, LLRs and systemic anaphylactic reactions are the most common (18).
Anaphylaxis
Systemic reactions limited to cutaneous signs only carry a risk of anaphylaxis to a future sting below 3%. Conversely, a history of systemic sting-induced reaction and detectable venom IgE put the risk of a second systemic reaction at 60% (19). The patient’s prior sting history – the severity and pattern of reactions, baseline tryptase level, age and concurrent medications all influence future risk (1). Hymenoptera sting-induced anaphylaxis must prompt investigations for an underlying mast cell disorder including hereditary α-tryptasemia (10-13, 20).
Local reactions
In the general population, the reported prevalence of LLR ranges between 2.4% and 26.4%. If local inflammation is contiguous with the sting site, it may be considered and managed as a local reaction (1). LLR are not dangerous unless they cause compression, and compartment syndrome develops, or if a patient is stung in the oropharynx, when airway obstruction becomes a risk (21), or in the context of an underlying condition (15)
LLR patients exhibit a 10% risk of systemic reactions and a 3% risk of severe anaphylaxis if re-stung  (1, 21).
Diagnostics Sensitization
A convincing clinical history and proven Vespula venom sensitization are required for the diagnosis of Vespula venom allergy (2, 9). As venom sensitization is identified in approximately 10–30% of history-negative persons, only those with a history of a previous systemic reaction are usually eligible for diagnostic testing (2, 9).
In vitro tests to whole venom extracts are negative in approximately 20% of patients with positive skin tests, and  positive in an estimated 10% of patients with negative skin tests, therefore the European guidelines recommend sequential skin and venom specific IgE testing as a standard protocol in all patients with a history of systemic reactions, ensuring a high diagnostic sensitivity of 94% (2, 9, 15). Recent data confirmed that in vitro and skin tests with Vespid venom extracts yielded complementary rather than overlapping results, and suggested that in vitro diagnosis might suffice, or might be performed as the first-line test  (19, 22).
As Hymenoptera venom IgE persist for extended periods, in vitro and skin testing can be done even a long time after the reported clinical reaction, however, it is recommended to observe a 2-week interval after the reaction before performing skin tests (2, 19). If, based on clinical history, the index of suspicion for anaphylactic reaction is high, but in vitro and skin tests are negative, testing should be repeated after one to six months (5, 9).
In vitro diagnostics
In vitro testing is devoid of clinical risk of adverse reactions to applied venom and is less labor-intensive than skin testing (22).
In a prospective diagnostic study, the positive predictive value of ImmunoCAP in vitro testing of Hymenoptera extracts was 77% and the negative predictive value 59%, while intradermal skin tests yielded 87% and 55% respectively (19).  The diagnostic sensitivity of the whole allergen extract of Vespula spp venom is 83-91%, with improved performance since its spiking with Ves v 5 (2, 23)
Besides allergen-extract specific IgE, in vitro investigation of Hymenoptera sting-induced reactions comprises allergen component IgE and CCD IgE determination to assess genuine versus cross-reactive sensitization, as double positivity to bee and wasp venom extracts during in vitro testing occurs in up to 50% of venom-allergic patients (2, 24, 25).
A combination of the Vespula spp recombinant allergen components rVes v 1 and rVes v 5 has been reported to yield a sensitivity as high as 92–98% for the identification of genuine sensitization to Vespula spp venom (2). Conversely, a patient who tests negative for specific IgE to both rVes v 1 and rVes v 5 is unlikely to have genuine wasp venom sensitization, regardless of IgE to other wasp components (26).
Total IgE could be useful, particularly in cases with low levels of specific IgE, for calculating the specific-to-total IgE ratio, a proposed indicator for clinically relevant sensitization (2).
Diagnostic investigation of Hymenoptera sting-induced systemic reactions also requires determination of baseline tryptase in search for a mast cell disorder, with levels at 8 µg/L or higher suggesting hereditary α-tryptasemia (2, 13). Further investigations such as testing for the D816V c-kit mutation in peripheral blood may be considered (12).
In the approximately 5% of Hymenoptera venom-allergic patients with elevated baseline tryptase levels and/or mastocytosis, diagnostic sensitivity is enhanced by using molecular components and a cut-off level of 0.1 kUA/L for specific IgE positivity (27).
Skin tests
Skin tests with Vespid venom extracts can be performed as skin prick tests or intradermal tests (9). Their diagnostic sensitivity is estimated at 64% (2).
The skin test reaction to venom extracts is not correlated to the severity of past or recurrent reactions to a future sting (2, 9).  The sensitivity of the skin prick test is lower than that of the intradermal test, which is used to confirm a negative result (9). Diagnostic intradermal testing is usually carried out using venom extracts of concentrations of between 0.001 µg/mL and 1.0 µg/mL. The accuracy of the test is subject to proportionate representation of the relevant allergens in the extract, as false-negatives may be the result of underrepresented components and, conversely, irritant compounds may lead to false-positives (1).
Challenge tests
Live sting challenges are not a standard procedure in clinical practice (1, 2, 15).
Other topics
Prevention and Therapy
Allergen immunotherapy
Venom immunotherapy (VIT) is the only treatment that can prevent future sting-induced anaphylaxis in Hymenoptera venom-allergic patients, effectively inducing tolerance in 91–99% of Vespid venom allergic patients (2). During successful Vespid VIT, venom-specific IgE decrease while venom-specific IgG and IgG4 increase (22).
VIT is most successful when the treatment is selected based on specific IgE to venom allergens, i.e., bee and/or wasp molecular allergens (2). Candidates for VIT must have a documented history of a systemic reaction to a sting and evidence of IgE reactivity to a specific venom. Treatment with the incorrect venom, or with more than one venom without evidence of sensitization, can induce de novo sensitization, increase the likelihood of adverse effects, risk insufficient protection, and increase costs (28).
The usual duration of VIT is 3 to 5 years, although more prolonged or even lifelong VIT should be considered in patients with mast cell disorders, due to an increased risk of severe reactions and a lower efficacy of VIT in these patients (2, 15).
An elevated baseline serum tryptase in a venom-allergic patient, even without a formal diagnosis of systemic mastocytosis, may be associated with severe anaphylactic reactions (29) and may indicate the need for lifelong VIT (30).
Prevention strategies
Sting avoidance is difficult to achieve as it requires caution during outdoor activities (2).
Other topics
An emergency kit comprising autoinjectable epinephrin should be carried by Hymenoptera venom-allergic patients having experienced systemic reactions, including those having completed a successful VIT (2).
Molecular Aspects
Allergenic molecules
Among the six characterized allergens from the Vespula spp venom, most are enzymes, such as the phosholipase A1 Ves v 1, while Ves v 5 is of as-yet unknown function and vitellogenin Ves v 6 contributes to embryo growth (31, 32). Vespula allergens are glycosylated, implying that IgE to cross-reactive carbohydrates (CCD) should also be assayed during in vitro diagnosis of Vespid sensitization. The biochemical properties, molecular mass, and sensitization prevalence for Vespula vulgaris allergens are summarized below (2, 31).
Table 2: Biochemical properties, molecular mass, and sensitization rate for Vespula vulgaris allergens (2, 31, 32).
Allergen Biochemical property/ alternative name Molecular weight (SDS-PAGE, kDa) Prevalence of sensitization in wasp-allergic patients (%)
Ves v 1 Phospholipase A1B 34 33 - 66
Ves v 2 Hyaluronidase 38 5 -25
Ves v 3 Dipeptidyl peptidase (DPP) IV 100 30 - 57
Ves v 5 Antigen 5 (unknown function) 23 82 – 98
Ves v 6 Vitellogenin 200 39
Cross-reactivity
Sensitization to Vespula spp venom is frequently associated to sensitization to other venoms, such as honeybee venom, Polistes spp venom, hornet venoms, or others. Double sensitization, whether resulting from genuine double sensitization or cross-reactivity, may be clinically relevant and put the patients at risk of severe reactions to stings from various Hymenoptera species, especially in subjects with underlying mast cell disorders (2, 9, 33).
In vitro diagnosis with marker molecular allergens allows the identification of the genuine sensitizers in cases of Vespid/honeybee venom double positivity, however, there is no currently available diagnostic marker to distinguish between Vespula spp and Polistes spp sensitization, nor between Vespula spp and hornet sensitization (2).  Double positivity to bee and wasp venom during in vitro testing, seen in 40% to 50% of venom-allergic patients, is solved by the use of marker allergens (Ves v 1 and Ves v 5 or Pol d 5 for Vespids, and Api m 1, Api m 3, Api m 4 and Api m 10 for honeybee venom) and CCD, allowing distinction between genuine bee-wasp cosensitization and sensitization to only bee or wasp with cross-reactivity through shared allergens or CCD (2).
Other topics
Compiled By
Author: Prof. Joana Vitte
Reviewer: Dr. Merima Mehic Chaveton
References
  1. Tracy JM, Golden DBK. Hymenoptera Venom Extracts in Clinical Practice. J Allergy Clin Immunol Pract. 2018;6(6):1856-62.
  2. EAACI. MAUG 2.0 2022 [Available from: https://hub.eaaci.org/resources_guidelines/molecular-allergology-users-guide-2-0/.
  3. dos Santos-Pinto JRA, Perez-Riverol A, Lasa AM, Palma MS. Diversity of peptidic and proteinaceous toxins from social Hymenoptera venoms. Toxicon. 2018;148:172-96.
  4. Diaz JH. Recognition, management, and prevention of hymenopteran stings and allergic reactions in travelers. J Travel Med. 2009;16(5):357-64.
  5. Philipp A, Ferdman RM, Tam JS. Evaluation of venom allergy. Ann Allergy Asthma Immunol. 2016;117(4):344-7.
  6. Turner PJ, Jerschow E, Umasunthar T, Lin R, Campbell DE, Boyle RJ. Fatal Anaphylaxis: Mortality Rate and Risk Factors. J Allergy Clin Immunol Pract. 2017;5(5):1169-78.
  7. Zahirovic A, Luzar J, Molek P, Kruljec N, Lunder M. Bee Venom Immunotherapy: Current Status and Future Directions. Clin Rev Allergy Immunol. 2020;58(3):326-41.
  8. Bilo MB, Bonifazi F. The natural history and epidemiology of insect venom allergy: clinical implications. Clin Exp Allergy. 2009;39(10):1467-76.
  9. Bonadonna P, Korosec P, Nalin F, Golden DB. Venom anaphylaxis: Decision points for a more aggressive workup. J Allergy Clin Immunol Pract. 2023.
  10. Schuler CFt, Volertas S, Khokhar D, Yuce H, Chen L, Baser O, et al. Prevalence of mastocytosis and Hymenoptera venom allergy in the United States. J Allergy Clin Immunol. 2021;148(5):1316-23.
  11. Greiner G, Sprinzl B, Gorska A, Ratzinger F, Gurbisz M, Witzeneder N, et al. Hereditary alpha tryptasemia is a valid genetic biomarker for severe mediator-related symptoms in mastocytosis. Blood. 2021;137(2):238-47.
  12. Selb J, Rijavec M, Erzen R, Zidarn M, Kopac P, Skerget M, et al. Routine KIT p.D816V screening identifies clonal mast cell disease in patients with Hymenoptera allergy regularly missed using baseline tryptase levels alone. J Allergy Clin Immunol. 2021;148(2):621-6 e7.
  13. Selb J, Rijavec M, Kopac P, Lyons JJ, Korosec P. HalphaT is associated with increased risk for severe Hymenoptera venom-triggered anaphylaxis. J Allergy Clin Immunol. 2023;151(3):804-5.
  14. Pastorello EA, Borgonovo L, Preziosi D, Schroeder JW, Pravettoni V, Aversano MG, et al. Basal Tryptase High Levels Associated with a History of Arterial Hypertension and Hypercholesterolemia Represent Risk Factors for Severe Anaphylaxis in Hymenoptera Venom-Allergic Subjects over 50 Years Old. Int Arch Allergy Immunol. 2021;182(2):146-52.
  15. Sturm GJ, Varga EM, Roberts G, Mosbech H, Bilo MB, Akdis CA, et al. EAACI guidelines on allergen immunotherapy: Hymenoptera venom allergy. Allergy. 2018;73(4):744-64.
  16. Khurana T, Bridgewater JL, Rabin RL. Allergenic extracts to diagnose and treat sensitivity to insect venoms and inhaled allergens. Ann Allergy Asthma Immunol. 2017;118(5):531-6.
  17. Lester PJ, Beggs JR. Invasion Success and Management Strategies for Social Vespula Wasps. Annu Rev Entomol. 2019;64:51-71.
  18. Elieh Ali Komi D, Shafaghat F, Zwiener RD. Immunology of Bee Venom. Clin Rev Allergy Immunol. 2018;54(3):386-96.
  19. Park HJ, Brooks DI, Chavarria CS, Wu RL, Mikita CP, Beakes DE. Combining Discordant Serum IgE and Skin Testing Improves Diagnostic and Therapeutic Accuracy for Hymenoptera Venom Hypersensitivity Immunotherapy. J Allergy Clin Immunol Pract. 2022;10(3):837-43 e3.
  20. Kacar M, Rijavec M, Selb J, Korosec P. Clonal mast cell disorders and hereditary alpha-tryptasemia as risk factors for anaphylaxis. Clin Exp Allergy. 2023.
  21. Golden DB. Large local reactions to insect stings. J Allergy Clin Immunol Pract. 2015;3(3):331-4.
  22. Hollstein MM, Matzke SS, Lorbeer L, Traidl S, Forkel S, Mobs C, et al. Skin Tests versus Serology: Specific IgE May Suffice for Diagnosis of Vespid Venom Allergy and Follow-Up of Allergen Immunotherapy. Int Arch Allergy Immunol. 2023:1-10.
  23. Vos B, Kohler J, Muller S, Stretz E, Rueff F, Jakob T. Spiking venom with rVes v 5 improves sensitivity of IgE detection in patients with allergy to Vespula venom. J Allergy Clin Immunol. 2013;131(4):1225-7, 7 e1.
  24. Muller U, Schmid-Grendelmeier P, Hausmann O, Helbling A. IgE to recombinant allergens Api m 1, Ves v 1, and Ves v 5 distinguish double sensitization from crossreaction in venom allergy. Allergy. 2012;67(8):1069-73.
  25. Spillner E, Blank S, Jakob T. Hymenoptera allergens: from venom to "venome". Front Immunol. 2014;5:77.
  26. Müller U, Schmid-Grendelmeier P, Hausmann O, Helbling A. IgE to recombinant allergens Api m 1, Ves v 1, and Ves v 5 distinguish double sensitization from crossreaction in venom allergy. Allergy. 2012;67(8):1069-73.
  27. Michel J, Brockow K, Darsow U, Ring J, Schmidt-Weber CB, Grunwald T, et al. Added sensitivity of component-resolved diagnosis in hymenoptera venom-allergic patients with elevated serum tryptase and/or mastocytosis. Allergy. 2016;71(5):651-60.
  28. Ollert M, Blank S. Anaphylaxis to insect venom allergens: role of molecular diagnostics. Curr Allergy Asthma Rep. 2015;15(5):26.
  29. Bilo BM, Rueff F, Mosbech H, Bonifazi F, Oude-Elberink JN, Hypersensitivity EIGoIV. Diagnosis of Hymenoptera venom allergy. Allergy. 2005;60(11):1339-49.
  30. Simons FE, Ardusso LR, Bilo MB, Cardona V, Ebisawa M, El-Gamal YM, et al. International consensus on (ICON) anaphylaxis. World Allergy Organ J. 2014;7(1):9.
  31. IUIS/WHO. Vespula vulgaris 2023 [Available from: http://allergen.org/search.php?allergenname=&allergensource=Vespula+vulgaris&TaxSource=&TaxOrder=&foodallerg=all&bioname=.
  32. Blank S, Seismann H, McIntyre M, Ollert M, Wolf S, Bantleon FI, et al. Vitellogenins are new high molecular weight components and allergens (Api m 12 and Ves v 6) of Apis mellifera and Vespula vulgaris venom. PLoS One. 2013;8(4):e62009.
  33. Blank S, Pehlivanli S, Methe H, Schmidt-Weber CB, Biedermann T, Horny HP, et al. Fatal anaphylaxis following a hornet sting in a yellow jacket venom-sensitized patient with undetected monoclonal mast cell activation syndrome and without previous history of a systemic sting reaction. J Allergy Clin Immunol Pract. 2020;8(1):401-3 e2.