Whole Allergen
k80 Formaldehyde/Formalin
k80 Formaldehyde/Formalin Scientific Information
Type:
Whole Allergen
Display Name:
Formaldehyde/Formalin
Allergen code:
k80
Route of Exposure:
Skin Contact
WHO/ICD-11 code:
XM0TV9
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Formaldehyde is a chemical agent found in virtually all homes and workplaces. In addition to acting as a contact allergen, inhalation of formaldehyde can exacerbate asthma.
Allergen
Nature
Formaldehyde is a low-molecular weight organic chemical (1), used in industrial and commercial settings as a solution, disinfectant, preservative, and resin (2), and in household settings as foam insulation, in cleaning and personal care products, textiles, dishwashing liquids and in pressed wood products (2-4). It is present in virtually all homes and workplaces (Lam et al, 2021). As such, formaldehyde is a common contact allergen (3) that, in addition to being an irritant, is also an allergen acting as a hapten (1).
Some products that do not contain formaldehyde still contain substances that release formaldehyde as they break down (4). These are known as formaldehyde releasers and can independently cause contact allergic dermatitis (4).
Epidemiology
Worldwide distribution 
Formaldehyde exposure is widespread in homes, communities, and workplaces (2). In Denmark over a ten-year period (2007–2016), the prevalence of contact allergy to formaldehyde 1% was found to be 1.5%, while contact allergy to formaldehyde 2% was found to be 2.4% (5). Among patients referred for allergic contact dermatitis to an American medical center, 8.1% were allergic to formaldehyde, 7.1% were allergic to Bioban™ (a cleaning chemical), and 9.5% were allergic to other formaldehyde releasers (6).
In Finland, formaldehyde and formaldehyde releasers caused 5.5% of cases of occupational allergic contact dermatitis during the period from 2005–2016 (7).
Risk factors 
Occupational contact allergic dermatitis is seen in many professions, including manual labor and professional occupations (8).
Low-income communities are disproportionately exposed to formaldehyde; the use of low-cost building materials, including pressed wood products, puts inhabitants at an increased risk of formaldehyde exposure (2).
Route of Exposure
Main 
Contact (3).
Secondary
Inhalation (2).
Clinical Relevance
Allergic contact dermatitis in formaldehyde-sensitized patients is often chronic due to the ubiquitous nature of formaldehyde, and frequently presents on the hands and face where contact is most likely (4).
In addition to contact allergies, inhalation of formaldehyde is a potential cause of asthma, where exposure occurs primarily through inhalation, but also occurs by direct contact irritation of the respiratory tract (2).
Molecular Aspects
Allergenic molecules
Main formaldehyde Releasers (adapted from (4)):
Cosmetics
Industrial Products
Quaternium -15
Biobans
Imidazolidinyl urea
Diazolidinyl urea
DMDM hydantoin
Grotan BK (tris(N-hydroxyethyl) hexahydrotriazine)
Bronopol (2-bromo-2-nitropropane-1,3-diol)
Textiles
Methenamine (hexamethylenetetramine)
Formaldehyde urea resin
Benzylhemiformal
Formaldehyde melamine
resin
Cross-reactivity
Formaldehyde-releasing substances induce positive reactions in formaldehyde-allergic patients (7); In a study, 80% of formaldehyde-allergic patients had a positive patch test reaction to at least one formaldehyde releaser, most commonly benzylhemiformal (7). In patients sensitized to bronopol and quaternium-15, 25% and over 50% respectively also react to formaldehyde (4). The strongest association of sensitization is between formaldehyde and quaternium-15 (4).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer 
Last reviewed: April 2022
References
  1. Jang JH, Park SH, Jang HJ, Lee SG, Park JH, Jeong JW, et al. A Case of Recurrent Urticaria Due to Formaldehyde Release from Root-Canal Disinfectant. Yonsei Med J. 2017;58(1):252-4.
  2. Lam J, Koustas E, Sutton P, Padula AM, Cabana MD, Vesterinen H, et al. Exposure to formaldehyde and asthma outcomes: A systematic review, meta-analysis, and economic assessment. PLoS One. 2021;16(3):e0248258.
  3. Flyvholm MA, Menné T. Allergic contact dermatitis from formaldehyde. A case study focussing on sources of formaldehyde exposure. Contact Dermatitis. 1992;27(1):27-36.
  4. Latorre N, Silvestre JF, Monteagudo AF. [Allergic contact dermatitis caused by formaldehyde and formaldehyde releasers]. Actas Dermosifiliogr. 2011;102(2):86-97.
  5. Fasth IM, Ulrich NH, Johansen JD. Ten-year trends in contact allergy to formaldehyde and formaldehyde-releasers. Contact Dermatitis. 2018;79(5):263-9.
  6. Anderson BE, Tan TC, Marks JG, Jr. Patch-test reactions to formaldehydes, bioban, and other formaldehyde releasers. Dermatitis. 2007;18(2):92-5.
  7. Aalto-Korte K, Pesonen M. Patterns of positive patch test reactions to formaldehyde and formaldehyde releasers at the Finnish Institute of Occupational Health from 2007 to 2020. Contact Dermatitis. 2021;85(4):429-34.
  8. Tagka A, Lambrou GI, Matsopoulos GK, Fytili D, Mirkopoulou D, Katsarou A, et al. Analysis of Prevalence and Risk Factors of Contact Sensitization with respect to the Occupational Profiles in a Greek Patient Cohort: A Retrospective Analysis of a Greek Referral Centre and Future Perspectives. Biomed Res Int. 2021;2021:6672506.