Type:
Whole Allergen
Whole Allergen
Whole Allergen
Tilletia tritici/Ustilago
m201
Ustilaginaceae
tritici/ nuda, tritici, segetum
Inhalation
Tilletia tritici / Ustilago nuda/tritici/maydis
Tilletia caries-Wheat bunt/ Ustilago nuda – U. tritici - U. segetum-loose wheat smut
Tilletia spp. (smuts) and Ustilago spp. (bunts) are common molds of the Class Ustilaginomycetes, mostly found in cereal crops and soil. They are well known plant pathogens which may have a role in the sensitization of allergic patients. The particles causing sensitization are spores found in the environment.
Smuts may be involved in the development of asthma and hypersensitivity pneumonitis. Cross reactivity with other molds within the Phylum Basidiomycota may occur.
The molds Tilletia tritici (T. caries) and Ustilago spp. are members of the Phylum Basidiomycota. Members of the subphylum Ustilaginomycotina are able to grow within, or on the surface of the host plants, including stems, leaves, flower parts, bulbs and seeds; teliospores are released in characteristic dark masses (1).
Fungi of the genus Tilletia are usually called “bunts” and those of the genus Ustilago are often collectively named “smuts” (2, 3). Many of these molds are plant pathogens, usually attacking cereal crops and causing different symptoms in the plants, which can appear blackened (4). Tilletia spp. can be found in infected seed and soil; these molds produce teliospores (a type of spore) within the plant ovaries. The spores can have a rough or smooth surface (4). Ustilago spp. produce thick walled teliospores that readily germinate into a cylindrical basidium (a reproductive structure); the teliospores can also survive dormant for years (1, 5, 6).
Smut spores from Ustilago species could be detected in several countries at different times of the year, with peaks in February, May, August – or the rainy season in Nigeria (7-12). Moderate to high winds and temperatures in November increased the presence of Ustilago spores in the air, whereas in spring the peak was due to lawn mowing and disturbances of wheat crops (8). High concentrations of smut spores positively correlated with higher temperatures and negatively correlated with humidity (10). Ustilago spores were also frequently detected in the indoor setting of a hospital outpatient ward (13).
Taxonomic tree of Tilletia genus (2, 14) |
|
Domain |
Eukaryota |
Kingdom |
Fungi |
Phylum |
Basidiomycota |
Subphylum |
Ustilaginomycotina |
Class |
Ustilaginomycetes |
Order |
Tilletiales |
Family |
Tilletiaceae |
Genus |
Tilletia |
Taxonomic tree of Ustilago genus (3, 15) |
|
Domain |
Eukaryota |
Kingdom |
Fungi |
Phylum |
Basidiomycota |
Subphylum |
Ustilaginomycotina |
Class |
Ustilaginomycetes |
Order |
Ustilaginales |
Family |
Ustilaginaceae |
Genus |
Ustilago |
Tilletia/Ustilago allergens are spread by airborne spores. Those of T. caries were ejected and able to achieve longer distances than the other fungal species studied (16). Smut spores ranged between 1.6–25.6% of the total detected in different studies (8-11, 17). In a study where spore sampling was carried out at altitude and at ground level, Ustilago spores had higher concentrations at ground level and in urban environments (17).
There is a distinct lack of specific reports of allergic disease attributed to Tilletia spp. and Ustilago spp.; however, their involvement in developing symptoms of asthma was demonstrated in Australian studies (18, 19). Smut species were also the cause of allergic disease in Japan and Northern America (20, 21).
Some occupations (e.g. farmers) may be at higher risk of allergic respiratory conditions when exposed to smuts (21), given the prevalence of these species (Tilletia/Ustilago) as cereal pathogens (2, 3).
Tilletia tritici (T. caries) can be found worldwide (2). Ustilago nuda (U. tritici/segetum) can be found in many countries (3).
Inhalation of spores (22).
Burkard Volumetric Spore Trap (7, 18, 19).
The concentration of Ustilago spores detected daily was normally less than 1,000 per m3, though a peak of nearly 6,000 per m3 was also recorded (7).
In a study of 2,098 children who received treatment in hospital for asthma, it was shown that in the age group 14–18 there was a significant association between the presence of environmental Ustilago spores and hospitalization (18). People with asthma could show higher inflammation and lower lung function when exposed to Ustilago smuts (19).
Ustilago esculenta has been reported as a cause of hypersensitivity pneumonitis (HP) in a 40-year old patient with symptoms of cough, fever and dyspnea (20). In another study into the development of HP in farmers who experienced respiratory symptoms, 11.2% of positive results (SPTs or RAST assays) were associated with grain smuts extracts (21).
The presence of allergen-specific antibodies is usually determined by skin prick tests (SPTs) and serology for IgE (22). In a study investigating skin test reactions of allergic patients to lesser reported fungi, 56% showed a positive dermal reactivity to fungal extracts, including Ustilago maydis (23). In another study of U. maydis extracts in 200 patients with either asthma or allergic rhinitis, asthmatic patients were more likely to have immediate skin reactions on SPT, whereas the patients with rhinitis were more likely to have late phase sensitivity (24).
Immunotherapy is currently not recommended for patients allergic to molds, due to complexities of the allergens and patient co-allergies (22).
Avoidance is difficult to achieve (22), due to the wide range of environments in which Tilletia spp. and Ustilago spp. can be found (2, 3).
An extract of Ustilago maydis showed cross-reactivity with Cryptococcus neoformans, suggesting that some Basidiomycetes may share antigens (25).
Author: RubyDuke Communications
Reviewer: Dr. Christian Fischer
Last reviewed:January 2022