Table of Contents
Component
m218 Asp f 1
m218 Asp f 1 Scientific Information
Type:
Component
Name; WHO/IUIS:
Asp f 1
Biological function:
unknown
Allergen code:
m218
Molecular Weight:
18 kDa
Source Material:
Recombinant, CCD-free protein
Other Names :
Ribonuclease mitogillin
Summary
Asp f 1, the ribonuclease mitogillin, a member of the ribotoxin family, is a major allergen of and marker of genuine sensitization to the ubiquitous environmental mold Aspergillus fumigatus (A. fumigatus), associated to allergic fungal airway diseases (AFAD) affecting upper and lower airways, such as allergic fungal rhinosinusitis (AFRS), severe asthma with fungal sensitization (SAFS), and allergic bronchopulmonary aspergillosis (ABPA). Asp f 1, a secreted protein, exerts cytotoxic and proinflammatory effects. 
Epidemiology
Worldwide distribution
A. fumigatus is a cosmopolitan, thermotolerant airborne fungus affecting human health through multiple pathways: sensitization and allergy, infection, direct toxicity [1].
The prevalence of Asp f 1 sensitization is highly variable depending on the study population in terms of geography, underlying lung condition (asthma, CF, or COPD), stage of AFAD, method of assessment (skin prick test, blood tests) and cut-off values (reviewed in [2, 3]). Despite wide variations, Asp f 1 sensitization presents with higher prevalence and higher levels in ABPA as compared to control populations. For example, Asp f 1 sensitization in ABPA was found at 60% and 67% in two Japanese cohorts [4, 5], at 83% in British patients [6] and at 100% in French patients [7]. In asthmatic patients with A. fumigatus sensitization, detectable IgE to Asp f 1 was reported in 18% of Japanese patients [4] and in 46% of German and Polish patients [8].
In an unselected population of 23,077 consecutive Italian subjects with a suspicion of airborne or food allergy, 0.43% displayed detectable IgE sensitization to Asp f 1, ranking 64 among 75 assayed allergenic molecules, and in 10% Asp f 1 presented as apparent monosensitization [9]
Environmental Characteristics
Source and tissue
Asp f 1 is a secreted ribonuclease [10]. 
Risk factors
Sensitization and allergy to A. fumigatus occur mainly in subjects with pre-existent lung conditions, usually asthma or cystic fibrosis (CF), although chronic obstructive pulmonary disease (COPD) is increasingly recognized as another predisposing condition [1].
Clinical Relevance
Detailed information regarding A. fumigatus is available in the whole allergen section. The demonstration of Asp f 1 IgE confirms genuine sensitization to A. fumigatus [10].  
Clinical relevance of Asp f 1 IgE in ABPA
The diagnosis of ABPA is often complicated by symptoms due to underlying conditions and a complex pathophysiology combining IgE and IgG responses, sputum and systemic eosinophilia, and much debated fungal colonization [3, 11]. The levels, rather than the prevalence, of Asp f 1 sensitization have been proposed as a diagnostic criterion for ABPA [2, 7, 12]. Median levels of IgE to Asp f 1 in ABPA patients may vary from 2.5 kUA/L to 17 kUA/L among different cohorts [5-7, 12], supporting the observation that statistically determined cut-off levels in a given population perform better than fixed cut-offs [12].  Using population-based cut-offs, the reported area under the receiver operating curve of Asp f 1 was close to 0.9 in different populations [7, 12]. A meta-analysis showed a pooled sensitivity of 80% and pooled specificity of 70.1% for Asp f 1 as a diagnostic tool for ABPA in asthmatic patients, and 78.9% and 81.9% respectively for the diagnosis of ABPA in CF patients [2]. Diagnostic performance of Asp f 1 is improved when used in conjunction with other A. fumigatus molecular allergens [12-14]  .
Disease severity and prediction
Increasing levels of Asp f 1 IgE in asthmatic or CF patients are statistically associated to an increased risk of ABPA, as explained above. Outside ABPA, it has long been known that sensitization to A. fumigatus is associated with lung function deterioration in adult and pediatric asthmatic patients [15, 16]. Using molecular allergen diagnostics in a COPD cohort, it was shown that Asp f 1 sensitization, rather than A. fumigatus sensitization, was associated with bronchiectasis, an irreversible complication leading to poorer lung function and mortality [17]. In this study, Asp f 1 sensitization was detectable in 12% of COPD patients and in 2% of controls (p = 0.02) and was associated with an odds ratio of 2.8 for having bronchiectasis.
Cross-reactive molecules
Asp f 1 is a member of the ribotoxin family and shares a sequence identity of 80% or with homologues from a small number of other Aspergillus species, mainly restrictocin from A. restrictus, and α-sarcin from A. giganteus [18-20].
Molecular Aspects
Biochemistry
Asp f 1 is a basic protein of 18 kDa, stabilized by four disulfide bonds, secreted by A. fumigatus into the environment. Similar to other members of the ribotoxin family, it acts as a ribonuclease. Asp f 1 targets the ribosome and thus blocks protein synthesis. Its biological function, although not yet identified, might relate to fungal defense and parasitism [19, 20]. The presentation of Asp f 1 is restricted to HLA DR2 and DR5  [21].
Isoforms, epitopes, antibodies
As of November 7th, 2021, Asp f 1 comprises only one isoallergen officially published by the World Health Organization (WHO) and the International Union of Immunological Societies (IUIS) Allergen Nomenclature: Asp f1.0101 [22].
Cross-reactivity due to structural similarity
Asp f 1 cross-reactivity at the IgE binding level has been reported with ribotoxins from a limited number of other Aspergillus species [18, 20].
Diagnostic Relevance
Disease Severity
Asp f 1 sensitization, a marker of genuine sensitization to A. fumigatus, is associated with ABPA [2]and lung function deterioration in COPD patients [17]
Cross-Reactivity
Asp f 1 displays clinically relevant cross-reactivity with a small number of related allergens from other Aspergillus species.
Exposure
Asp f 1 sensitization is believed to occur mainly through inhalation upon exposure to A. fumigatus [10]
Compiled By
Author: Joana Vitte
Reviewer: Dr. Christian  Fischer
Last reviewed:January 2022
References
  1. Wardlaw, A.J., et al., New Perspectives in the Diagnosis and Management of Allergic Fungal Airway Disease. J Asthma Allergy, 2021. 14: p. 557-573.
  2. Muthu, V., et al., Utility of recombinant Aspergillus fumigatus antigens in the diagnosis of allergic bronchopulmonary aspergillosis: A systematic review and diagnostic test accuracy meta-analysis. Clin Exp Allergy, 2018. 48(9): p. 1107-1136.
  3. Carsin, A., et al., Aspergillus fumigatus in cystic fibrosis: An update on immune interactions and molecular diagnostics in allergic bronchopulmonary aspergillosis. Allergy, 2017. 72(11): p. 1632-1642.
  4. Tanimoto, H., et al., Molecular-based allergy diagnosis of allergic bronchopulmonary aspergillosis in Aspergillus fumigatus-sensitized Japanese patients. Clin Exp Allergy, 2015. 45(12): p. 1790-800.
  5. Kuwabara, K., et al., Serological analysis of sensitization in allergic bronchopulmonary aspergillosis: a study on allergen components and interspecies relationships. J Asthma, 2020. 57(6): p. 610-617.
  6. Alghamdi, N.S., et al., Serum IgE and IgG reactivity to Aspergillus recombinant antigens in patients with cystic fibrosis. J Med Microbiol, 2019. 68(6): p. 924-929.
  7. Vitte, J., et al., Aspergillus fumigatus components distinguish IgE but not IgG4 profiles between fungal sensitization and allergic broncho-pulmonary aspergillosis. Allergy, 2016. 71(11): p. 1640-1643.
  8. Kespohl, S. and M. Raulf, Mold Sensitization in Asthmatic and Non-asthmatic Subjects Diagnosed with Extract-Based Versus Component-Based Allergens. Adv Exp Med Biol, 2019. 1153: p. 79-89.
  9. Scala, E., et al., Cross-sectional survey on immunoglobulin E reactivity in 23,077 subjects using an allergenic molecule-based microarray detection system. Clin Exp Allergy, 2010. 40(6): p. 911-21.
  10. Matricardi, P.M., et al., EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol, 2016. 27 Suppl 23: p. 1-250.
  11. Asano, K., et al., New clinical diagnostic criteria for allergic bronchopulmonary aspergillosis/mycosis and its validation. J Allergy Clin Immunol, 2021. 147(4): p. 1261-1268 e5.
  12. Muthu, V., et al., Diagnostic Cutoffs and Clinical Utility of Recombinant Aspergillus fumigatus Antigens in the Diagnosis of Allergic Bronchopulmonary Aspergillosis. J Allergy Clin Immunol Pract, 2020. 8(2): p. 579-587.
  13. Vitte, J., et al., Multivariate Analysis As a Support for Diagnostic Flowcharts in Allergic Bronchopulmonary Aspergillosis: A Proof-of-Concept Study. Frontiers in Immunology, 2017. 8.
  14. Lukaszewicz, R., et al., Medical algorithm: Aspergillus fumigatus components in the diagnosis of Allergic Bronchopulmonary Aspergillosis. Allergy, 2021.
  15. Woolnough, K.F., et al., The relationship between biomarkers of fungal allergy and lung damage in asthma. Clin Exp Allergy, 2017. 47(1): p. 48-56.
  16. Welsh, K.G., et al., Fungal sensitization and positive fungal culture from sputum in children with asthma are associated with reduced lung function and acute asthma attacks respectively. Clin Exp Allergy, 2021. 51(6): p. 790-800.
  17. Everaerts, S., et al., Sensitization to Aspergillus fumigatus as a risk factor for bronchiectasis in COPD. Int J Chron Obstruct Pulmon Dis, 2017. 12: p. 2629-2638.
  18. Olombrada, M., et al., Fungal Ribotoxins: A Review of Potential Biotechnological Applications. Toxins (Basel), 2017. 9(2).
  19. Arruda, L.K., et al., Aspergillus fumigatus allergen I, a major IgE-binding protein, is a member of the mitogillin family of cytotoxins. J Exp Med, 1990. 172(5): p. 1529-32.
  20. UniProt, B. BLAST UniProtKB - P67875 (RNMG_ASPFU) Asp f 1.  November 7 2021]; Available from: https://www.uniprot.org/blast/uniprot/B20211107A084FC58F6BBA219896F365D15F2EB4401C12BI.
  21. Patel, G. and P.A. Greenberger, Allergic bronchopulmonary aspergillosis. Allergy Asthma Proc, 2019. 40(6): p. 421-424.
  22. IUIS/WHO. IUIS/WHO Aspergillus fumigatus.  November 7 2021]; Available from: http://allergen.org/search.php?allergenname=&allergensource=Aspergillus+fumigatus&TaxSource=&TaxOrder=&foodallerg=all&bioname=.