Table of Contents
Component
o217 Can s 3
o217 Can s 3 Scientific Information
Type:
Component
Name; WHO/IUIS:
Can s 3
Biological function:
Defense and pathogenesis
Allergen code:
o217
Molecular Weight:
9 kDa
Other Names :
Nonspecific lipid transfer protein type 1 (nsLTP 1)
Summary
Despite the widespread use of cannabis, descriptions of genuine IgE-dependent allergic reactions are relatively rare. However, several European surveys have suggested that Can s 3, a nonspecific lipid transfer protein of Cannabis sativa, may be an important cannabis allergen. As such, is the best studied and characterized cannabis protein in the context of allergic reactions. Passive inhalation of smoke from C. sativa without any previous active exposure can lead to sensitization to Can s 3 and other nsLTP allergens. Additionally, both the severe phenotype and the extensive cross-reactivity associated with cannabis allergy can be attributed to the physiochemical properties of Can s 3. Recent studies have suggested clinically-relevant sensitization also occurs in North America and Northern Europe, with broadly similar nsLTP sensitization profiles to those seen in Mediterranean areas..
Epidemiology
Worldwide distribution
Several European surveys have suggested that Can s 3, a nonspecific lipid transfer protein (nsLTP) of Cannabis sativa, may be an important cannabis allergen (1-3).
A case-control study in Belgium reported that 11/12 patients with cannabis allergy (median age 29 years, range 5–41; 6 males) were sensitized to nsLTP (1). In a subsequent study in Belgium, up to 72% of 120 patients with cannabis allergy reporting likely-anaphylaxis were sensitized to Can s 3 (median age 29.2 years; 48% male) (2). In this study, patients were categorized “likely-anaphylactic” if they presented with generalized symptoms to cannabis in two or more organ systems (e.g. respiratory, gastrointestinal, cardiovascular and/or cutaneous) (2).
In Spain, a survey of the sensitization profiles of patients diagnosed with primary cannabis allergy (mean age 26 ± 9 years; 97 men) found that 95.3% (124/130) of sensitized patients showed a response to cannabis LTP (3). These patients were diagnosed with cannabis allergy following positive skin prick test (SPT), specific IgE and bronchial challenge (3).
However, cannabis-sensitive individuals may not necessarily be sensitized to Can s 3 (4). A study in the United States reported on the sequencing of allergens from 23 patients with a positive SPT to a crude extract of C. sativa, none of whom were sensitized to Can s 3 (5). 
Environmental Characteristics
Source and tissue
Discovered and described 40 years ago, nsLTPs are ubiquitous in the plant kingdom and play an important role in plant defense against biotic and abiotic stress in most plant tissues (4). Assessments of Can s 3 sensitivity have variously used crude extracts from C. sativa buds (6) and/or leaves and flowers (3, 5, 7, 8).
Clinical Relevance
Disease severity
Despite the widespread use of cannabis (9), descriptions of genuine IgE-dependent allergic reactions are relatively rare (2, 10, 11), with under-reporting of symptoms likely to be a result of the illegal status of cannabis in most jurisdictions worldwide (10, 12-14). However, reports indicate that the prevalence of IgE-mediated cannabis allergy is increasing, and will continue to increase in the future, due to decriminalization and increasing social, medical and occupational exposure to C. sativa (1, 8, 10, 12, 15).
Cannabis allergy can significantly impact quality of life and manifest severe and generalized symptoms with extensive cross-reactions to other plant-derived foods and substances containing nsLTPs (4, 15). Allergic diseases associated with C. sativa include allergic rhinitis, asthma, allergic conjunctivitis, eczema, food allergy, drug eruption, contact urticaria and anaphylaxis (16).
Passive inhalation of smoke from C. sativa without any previous active exposure can lead to sensitization to Can s 3 and other nsLTP allergens (17). Additionally, both the severe phenotype and the extensive cross-reactivity associated with cannabis allergy can be attributed to the physiochemical properties of Can s 3 (2, 15).
Molecular Aspects
Biochemistry
Can s 3 is the best studied and characterized cannabis protein in the context of allergic reactions (15). Using a crude marijuana extract, Gamboa et al. (2007) were the first to purify and identify Can s 3, a heat- and acid-stable, IgE-binding nsLTP which is responsible in planta for dissolving, binding and shuttling of monomeric lipids between cell membranes (6, 15). Can s 3 showed a single band on SDS-PAGE and was identified as a lipid transfer protein according to its N-terminal amino acid sequence (XITCGQVASS), molecular size (9,226 d as determined by matrix-assisted laser desorption/ionization analysis) and recognition by Pru p 3 antibodies – the nsLTP from peach (6). In vitro tests (sIgE determination, ELISA inhibition and basophil activation test [BAT]) further supported the relevance of Can s 3 as a C. sativa allergen (6).
Cross-reactivity due to structural similarity
Sensitization to nsLTP and consequent reactions to plant foods has become an increasing concern over the last 20 years (4). Most nsLTP allergens have a highly-conserved structure stabilized by four disulfide bridges and have been shown to be very cross-reactive (4). The “cannabis-fruit/vegetable syndrome”, mainly reported in Europe, describes secondary cross-allergies with nsLTP homologues which appear to be triggered by sensitization to cannabis allergens such as Can s 3 (10). However, more recent studies have suggested clinically-relevant sensitization also occurs in North America and Northern Europe, with broadly similar nsLTP sensitization profiles to those seen in Mediterranean areas (4)
Sequence homologues (not exhaustive) for Can s 3 have been shown to demonstrate cross-reactivity to Pru p 3 (peach), Mal d 3 (apple), Cit s 3 (citrus), Mus a 3 (banana), Sol d 3 (tomato), Nic t LTP1 (tobacco), Vit v 1 (grape), Cor a 8 (hazelnut), Hev b 12 (latex), Ara h 9 (peanut), Tri a 14 (wheat) and Jug r 3 (walnut) (reviewed in: (10, 15)).
Diagnostic Relevance
In vitro diagnosis
Multiple reports have described the use of specific IgE (sIgE) quantifications for cannabis using either crude cannabis/hemp extracts or purified/recombinant components (reviewed in: (15)). Diagnostics based on crude cannabis products or extracts can yield clinically irrelevant results, irrespective of the methodology used, with relatively low specificity and positive test results in a significant number of cannabis tolerant individuals, especially in multi-sensitized individuals (15). On the other hand, the use of component-based diagnostics entails a risk of false-negative results as it is unlikely that all cannabis allergic individuals are sensitized to a single component of cannabis (15).
An assessment of the performance of five cannabis diagnostic tests (e.g. sIgE hemp, sIgE and BAT with a recombinant Can s 3 protein from C. sativa (rCan s3), BAT with a crude cannabis extract, and a skin prick test (SPT) with an nCan s 3-rich cannabis extract), concluded that the most effective and practical tests to confirm cannabis allergy are the SPT with an nCan s 3-rich extract and the sIgE rCan s 3 (2). These authors also noted that a negative test result with the sIgE hemp assay could serve as a suitable diagnostic in central Europe to exclude the risk of cannabis allergy (only 18% of individuals with cannabis allergy had negative sIgE hemp results) (2).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Fabio Lachetti
Last reviewed: October  2021
References
  1. Ebo DG, Swerts S, Sabato V, Hagendorens MM, Bridts CH, Jorens PG, et al. New food allergies in a European non-Mediterranean region: is Cannabis sativa to blame? Int Arch Allergy Immunol. 2013;161(3):220-8.
  2. Decuyper, II, Van Gasse AL, Faber MA, Elst J, Mertens C, Rihs HP, et al. Exploring the Diagnosis and Profile of Cannabis Allergy. J Allergy Clin Immunol Pract. 2019;7(3):983-9.e5.
  3. Armentia A, Herrero M, Martín-Armentia B, Rihs HP, Postigo I, Martínez-Quesada J. Molecular diagnosis in cannabis allergy. J Allergy Clin Immunol Pract. 2014;2(3):351-2.
  4. Skypala IJ, Asero R, Barber D, Cecchi L, Diaz Perales A, Hoffmann-Sommergruber K, et al. Non-specific lipid-transfer proteins: Allergen structure and function, cross-reactivity, sensitization, and epidemiology. Clin Transl Allergy. 2021;11(3):e12010.
  5. Nayak AP, Green BJ, Sussman G, Berlin N, Lata H, Chandra S, et al. Characterization of Cannabis sativa allergens. Ann Allergy Asthma Immunol. 2013;111(1):32-7.
  6. Gamboa P, Sanchez-Monge R, Sanz ML, Palacín A, Salcedo G, Diaz-Perales A. Sensitization to Cannabis sativa caused by a novel allergenic lipid transfer protein, Can s 3. J Allergy Clin Immunol. 2007;120(6):1459-60.
  7. Larramendi CH, López-Matas M, Ferrer A, Huertas AJ, Pagán JA, Navarro L, et al. Prevalence of sensitization to Cannabis sativa. Lipid-transfer and thaumatin-like proteins are relevant allergens. Int Arch Allergy Immunol. 2013;162(2):115-22.
  8. Rihs HP, Armentia A, Sander I, Brüning T, Raulf M, Varga R. IgE-binding properties of a recombinant lipid transfer protein from Cannabis sativa. Ann Allergy Asthma Immunol. 2014;113(2):233-4.
  9. UN. World Drug Report 2020. 2: Drug Use and Health Consequences 2020 [updated June 2020; cited 2021 June]. Available from: https://wdr.unodc.org/wdr2020/en/drug-use-health.html.
  10. Decuyper I, Ryckebosch H, Van Gasse AL, Sabato V, Faber M, Bridts CH, et al. Cannabis Allergy: What do We Know Anno 2015. Arch Immunol Ther Exp (Warsz). 2015;63(5):327-32.
  11. Chatkin JM, Zani-Silva L, Ferreira I, Zamel N. Cannabis-Associated Asthma and Allergies. Clin Rev Allergy Immunol. 2019;56(2):196-206.
  12. Bahji A, Stephenson C. International Perspectives on the Implications of Cannabis Legalization: A Systematic Review & Thematic Analysis. Int J Environ Res Public Health. 2019;16(17).
  13. Davidson M, Reed S, Oosthuizen J, O'Donnell G, Gaur P, Cross M, et al. Occupational health and safety in cannabis production: an Australian perspective. Int J Occup Environ Health. 2018;24(3-4):75-85.
  14. Bridgeman MB, Abazia DT. Medicinal Cannabis: History, Pharmacology, And Implications for the Acute Care Setting. P t. 2017;42(3):180-8.
  15. Decuyper, II, Rihs HP, Van Gasse AL, Elst J, De Puysseleyr L, Faber MA, et al. Cannabis allergy: what the clinician needs to know in 2019. Expert Rev Clin Immunol. 2019;15(6):599-606.
  16. Ocampo TL, Rans TS. Cannabis sativa: the unconventional "weed" allergen. Ann Allergy Asthma Immunol. 2015;114(3):187-92.
  17. Decuyper, II, Faber MA, Sabato V, Bridts CH, Hagendorens MM, Rihs HP, et al. Where there's smoke, there's fire: cannabis allergy through passive exposure. J Allergy Clin Immunol Pract. 2017;5(3):864-5.