Whole Allergen
w17 Firebush (Kochia)
w17 Firebush (Kochia) Scientific Information
Type:
Whole Allergen
Display Name:
Firebush (Kochia)
Allergen code:
w17
Family:
Amaranthaceae (Chenopodiaceae)
Latin Name:
Kochia scoparia (Synonyms: Bassia scoparia, Chenopodium scoparia)
Other Names:
Firebush, Kochia, Common Kochia
WHO/ICD-11 code:
XM3EW6
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Clinical Relevance
IgE mediated reactions
Anecdotal evidence suggests that asthma, allergic rhinitis and allergic conjunctivitis are common following exposure to pollen from Kochia; however, few specific studies have been reported to date. (1-3)
In one study in Thailand, Kochia was shown to be the second most important weed aeroallergen, with 14% of 100 patients with allergic rhinitis sensitised to it. (1)
In a study in the Midwestern USA, evaluating the frequency of sensitisation to cannabis pollen, found that 61% were skin prick positive for cannabis and all subjects were also skin test positive to weeds pollinating during the same period: Ragweed, Pigweed, Cocklebur, Russian thistle, Marsh elder, and Kochia. (4)
Among 1,159 patients attending an allergy clinic in Saudi Arabia, 51% of Saudi Arab patients and 28% of North American expatriates living in the area were sensitised to Kochia. This weed’s pollen was the 2 (nd) and 7 (th) most prevalent allergen sensitising the respective groups.(3)
Kochia pollen is also common in Tehran, Iran, (5) and a common cause of sensitisation in St. Louis, Missouri, USA. (6)
Of 327 adult patients with respiratory, dermatologic and ophthalmologic diseases of suspected allergic origin who attended a Hospital based in the United Arab Emirates, skin prick tests found that 244 patients (74.6%) were sensitised to at least one allergen. The twelve most common allergens were: Mesquite (45.5%), Grass Mix (40.7%), Cottonwood (33.1%), Bermuda grass (31.3%), Kochia (25.8%), Acacia (25.6%), Alfalfa (22.9%), Chenopodium (19.6%), Date palm (13.8%), Cockroach (14.7%), house dust (11.9%) and dust mite (9.5%). (2)
Molecular Aspects
No allergens from this plant have yet been characterised.
In a study of sera from 20 US donors with a positive clinical history and strong skin prick test to Salp pollen extract (Salsola pestifer/ Russian thistle / saltwort), Koc s contained one major and four minor allergens of which proteins of 35 and 55 kDa were suggested to be important allergens. (7)
In a study of Iranian allergic patients sensitised to Amaranthus retroflexus (Common pigweed) pollen, several allergenic components (85, 45, 39, 18, 15, and 10 kDa) of the A. retroflexus pollen extract were recognised in patients' sera and the IgE reactivity of the pollen extract was partially inhibited by Chenopodium albumK. scoparia, and Salsola kali. Three proteins (39, 45, and 66 kDa) were suggested as the common allergenic components among the pollens. (8)
Cross Reactivity
An extensive cross-reactivity among the different individual species of the genus could be expected, as well as to a certain degree among members of the families Chenopodiaceae and Amaranthaceae. (9,10)
Compiled By
Last reviewed: May 2022
References
  1. Pumhirun P, Towiwat P, Mahakit P. Aeroallergen sensitivity of Thai patients with allergic rhinitis. Asian Pac J Allergy Immunol 1997;15(4):183-5.
  2. Bener A, Safa W, Abdulhalik S, Lestringant GG. An analysis of skin prick test reactions in asthmatics in a hot climate and desert environment. Allerg Immunol (Paris) 2002;34(8):281-6.
  3. Suliaman FA, Holmes WF, Kwick S, Khouri F, Ratard R. Pattern of immediate type hypersensitivity reactions in the Eastern Province, Saudi Arabia. Ann Allergy Asthma Immunol  1997;78(4):415-8.
  4. Stokes JR, Hartel R, Ford LB, Casale TB. Cannabis (hemp) positive skin tests and respiratory symptoms. Ann Allergy Asthma Immunol 2000;85(3):238-40.
  5. Shafiee A. Atmospheric pollen counts in Tehran, Iran, 1974. Pahlavi Med J 1976;7(3):344-51.
  6. Lewis WH, Imber WE. Allergy epidemiology in the St. Louis, Missouri, area. IV. weeds. Ann Allergy 1975;35(3):180-7.
  7. Wurtzen PA, Nelson HS, Lowenstein H, Ipsen H. Characterization of Chenopodiales (Amaranthus retroflexus, Chenopodium album, Kochia scoparia, Salsola pestifer) pollen allergens. Allergy 1995;50(6):489-97.
  8. Tehrani M, Sankian M, Assarehzadegan MA, Falak R, Jabbari F, Varasteh A. Immunochemical characterization of amaranthus retroflexus pollen extract: extensive cross-reactive allergenic components among the four species of amaranthaceae/chenopodiaceae. Iran J Allergy Asthma Immunol 2010;9(2):87-95.
  9. Lombardero M, Duffort O, Selles JG, Hernandez J, Carreira J. Cross-reactivity among Chenopodiaceae and Amaranthaceae. Ann Allergy 1985;54(5):430-6.
  10. Yman L. Botanical relations and immunological cross-reactions in pollen allergy. 2nd ed. Pharmacia Diagnostics AB. Uppsala. Sweden. 1982: ISBN 91-970475-09.