Does your lab need to increase capacity but have little space to spare? The Phadia™ 200 instrument is made for you.
It has the smallest footprint of all the Phadia Laboratory Systems yet has access to our comprehensive portfolio of up to 650 ImmunoCAP™ and EliA™ tests. It runs on your benchtop and is designed to fit in compact spaces while delivering full automation and quantitative results.
The Phadia 200 instrument also fits into any workflow, enabling you to smoothly expand your test capacity, run specialty tests, and conduct research without affecting the lab’s routine workflow.
*Cluster setup allows several Phadia instruments networked together to be operated with one Phadia Prime computer.
**96 ImmunoCAP test results per batch or 48 EliA tests results per batch.
2 x 96 results per day
2 x 48 results per day