Contact Us
Phadia™ Laboratory Systems
Reliable results to aid in the diagnosis of allergy and autoimmune diseases
Phadia Laboratory Systems are fully automated and designed to optimize workflow efficiency regardless of the size and test volume of your lab. They adapt to your specific needs and allow you to scale seamlessly as your lab operations grow. 
Made to fit
The Phadia family of instruments features a large test menu along with shared system reagents and calibration curves that are stable for 28 days. Combined, the features help minimize time and reagent waste.
All Phadia instruments can run the latest Phadia™ Prime software that allows you to optimize laboratory and information workflows. Regardless of your throughput requirements, space restrictions, and resources, there is a Phadia instrument that meets your needs.
Phadia instruments help your laboratory by:
Gears Scheduled Maintenance
Providing full automation with hands-on time of 15 minutes, enabling your lab to increase productivity with fewer resources.
Step-by-Step OR Workflow
Optimizing workflow efficiency. All calibration curves are stable for one month and cover multiple tests, minimizing time and reagent waste.
Clipboard Checkmark 2
Enabling you to produce accurate, reproducible test results every shift without unnecessary downtime.
Which Phadia instrument is right for your laboratory?
Regardless of your throughput requirements, space restrictions, and resources, there is a Phadia instrument that addresses your needs.
Phadia 200 Instrument
The Phadia™ 200 instrument is a benchtop instrument for EliA™ tests and ImmunoCAP™ tests
Phadia 250 Instrument
Reliability meets versatility on the Phadia™ 250, which runs EliA™ tests and ImmunoCAP™ tests.
Thermo Fisher Phadia 2016. Photo: Markus Marcetic
Phadia 1000 Instrument
The Phadia™ 1000 instrument is a dedicated ImmunoCAP™ test system.
Thermo Fisher Phadia 2016. Photo: Markus Marcetic
Phadia 2500+ Instrument
The Phadia™ 2500+ series meets the demand for high throughput.
Thermo Fisher Phadia 2016. Photo: Markus Marcetic
Phadia 5000+ Instrument
The Phadia™ 5000+ series doubles the capacity of the Phadia 2500+ series.
Cluster setup allows several Phadia instruments networked together to be operated with one Phadia Prime computer.
Find an instrument to run ImmunoCAP™ tests.
Find an instrument to run EliA™ tests.
Software & Services
Phadia™ Prime Software
Gain control over your lab testing environment while ensuring traceability to support compliance.
Phadia™ Stock Manager software module
Simplified inventory management for Phadia Laboratory Systems.
Phadia™ Workflow Advisory Service
The only lab workflow optimization service designed specifically for allergy and autoimmune diagnostics.
Phadia™ LabCommunity
Maximize the productivity of your lab with quick, expert remote support and troubleshooting, and automatic reporting of approved Quality Club results.
Laboratory Automation Systems
Increase workflow efficiency with a connection to a Laboratory Automation System.
Quality Club™
Confirm and benchmark your testing quality.
Service Level Agreement
Invest in peace of mind with a service level agreement.
Phadia™ Xplain ICAP software module
Support result interpretation while improving service to clinicians.
Special Allergen Service
Get allergens designated for research use only.
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability. WEEE Producer Registration Number (Republic of Ireland):  IE002099WB