Phadia Laboratory Systems
From a world leader in allergy and autoimmunity diagnostics.
Take on a higher test volume with fewer resources, while still delivering reliable, accurate results.
No matter the lab size or test volume, unlock your lab's full potential with Phadia Laboratory Systems.
Automated and flexible systems made to fit your specific lab.
Compare instruments
*The Phadia 2500+ series comprises the Phadia™ 2500, Phadia™ 2500E, and Phadia™ 2500EE instruments.
Want to embody efficiency? Need more automation? Desire to maximize patient outcomes?
Our workflow and laboratory experts are here to help you transform into your ideal lab.
Let's talk
Smoother than your most flawless blood smear,
Phadia systems’ workflow makes efficiency seem effortless.
Boot up and head out
With low hands-on time during start up and automatic shutdown and rinse capabilities, you're able to walk away until it’s time to review results.1-3
28-day calibration curve1-3
All Phadia™ Laboratory Systems utilize master calibration curves by isotype, allowing more time for testing with less upkeep and waste.
Allergy and autoimmunity testing capabilities1-3
Consolidate your testing with Phadia Systems’ ability to run both ImmunoCAP allergy solutions and EliA autoimmune solutions.*
*Does not include the Phadia™ 1000 system, which runs only ImmunoCAP Allergy solutions.
Boot up and head out
With low hands-on time during start up and automatic shutdown and rinse capabilities, you're able to walk away until it’s time to review results.1-3
28-day calibration curve1-3
All Phadia™ Laboratory Systems utilize master calibration curves by isotype, allowing more time for testing with less upkeep and waste.
Allergy and autoimmunity testing capabilities1-3
Consolidate your testing with Phadia Systems’ ability to run both ImmunoCAP allergy solutions and EliA autoimmune solutions.*
*Does not include the Phadia™ 1000 system, which runs only ImmunoCAP Allergy solutions.
Streamlining autoimmune workflow with the help of Helios Automated IFA Systems.
Helios® Automated IFA Systems
Automated and climate-controlled IFA processing for ANA, ANCA, and dsDNA allowing labs of any size to access efficient and straightforward IFA testing.
Phadia Laboratory Systems & EliA autoimmune diagnostics
Workflow-optimized systems and quality assays that offer labs the opportunity to optimize their autoimmune disease specificity by identifying specific antibodies in individual patient samples.
  Thermo Fisher Scientific is the official US distributor of Helios systems and AESKUSLIDES®
Every lab is different.
Which Phadia instrument is the catalyst for your lab’s excellence?
Phadia 1000
Name: Phadia 1000
ImmunoCAP: Allergy
Sample Types: Serum, Plasma
EliA:
System Description:
Fully-automated closed immunoassay system
Processing: Continuous
Maximum Throughput: Up to 240 results/hour
Learn More Path: /content/phadia/us/en/our-solutions/phadia-laboratory-systems/phadia-1000
Phadia 5000
Name: Phadia 5000+ series
ImmunoCAP: Allergy
Sample Types: Serum, Plasma
EliA: Autoimmunity
System Description:
Fully-automated closed immunoassay system
Processing: Continuous
Maximum Throughput: Up to 960 results/hour
Learn More Path: /content/phadia/us/en/our-solutions/phadia-laboratory-systems/phadia-5000
Phadia 250
Name: Phadia 250
ImmunoCAP: Allergy
Sample Types: Serum, plasma
EliA: Autoimmunity
System Description:
Fully-automated closed immunoassay system 
Processing: Continuous
Maximum Throughput: Up to 60 results/hour
Learn More Path: /content/phadia/us/en/our-solutions/phadia-laboratory-systems/phadia-250
Phadia 2500
Name: Phadia 2500+ series
ImmunoCAP: Allergy
Sample Types: Serum, Plasma
EliA: Autoimmunity
System Description:
Fully-automated closed immunoassay system
Processing: Continuous
Maximum Throughput: Up to 480 results/hour
Learn More Path: /content/phadia/us/en/our-solutions/phadia-laboratory-systems/phadia-2500
Phadia 1000
Name: Phadia 1000
ImmunoCAP: Allergy
Sample Types: Serum, Plasma
EliA:
System Description:
Fully-automated closed immunoassay system
Processing: Continuous
Maximum Throughput: Up to 240 results/hour
Learn More Path: /content/phadia/us/en/our-solutions/phadia-laboratory-systems/phadia-1000
Phadia 5000
Name: Phadia 5000+ series
ImmunoCAP: Allergy
Sample Types: Serum, Plasma
EliA: Autoimmunity
System Description:
Fully-automated closed immunoassay system
Processing: Continuous
Maximum Throughput: Up to 960 results/hour
Learn More Path: /content/phadia/us/en/our-solutions/phadia-laboratory-systems/phadia-5000
Phadia 250
Name: Phadia 250
ImmunoCAP: Allergy
Sample Types: Serum, plasma
EliA: Autoimmunity
System Description:
Fully-automated closed immunoassay system 
Processing: Continuous
Maximum Throughput: Up to 60 results/hour
Learn More Path: /content/phadia/us/en/our-solutions/phadia-laboratory-systems/phadia-250
Phadia 2500
Name: Phadia 2500+ series
ImmunoCAP: Allergy
Sample Types: Serum, Plasma
EliA: Autoimmunity
System Description:
Fully-automated closed immunoassay system
Processing: Continuous
Maximum Throughput: Up to 480 results/hour
Learn More Path: /content/phadia/us/en/our-solutions/phadia-laboratory-systems/phadia-2500
Unlock your lab’s potential with Phadia instruments today.
References
  1. Phadia® 250 User Manual 12-3904-01/EN Edition 2.2. published 2017 Sep.
  2. Phadia® 1000 User Manual 12-3803-01/EN Edition 2.6. published 2020 July
  3. Phadia® 2500 and Phadia 5000® User Manual EN Edition 1.1. published 2012 March