Talk with Us
Hidden Button for comparing instruments
Phadia 1000 instrument
Compare Instruments
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
The Phadia™ 1000 instrument is dedicated to aid in the diagnosis of allergic diseases.
It provides consistent and reproducible test results with clinical relevance, ideal for high throughput laboratories delivering 2,000 to 10,000 test results per week.
Check out the features that make the Phadia 1000 instrument right for your laboratory
Instrument Details
Back
Sample loading area
High-capacity sample loading area and continuous sample loading without interruptions to allow operator flexibility.
QC positions and ImmunoCAP carrier storage
Run three methods in parallel with dedicated quality controls, enabling more than 300 different tests to be performed in one assay run with all reagents on board.
ImmunoCAP carrier storage
Ensures all necessary tests are onboard to avoid interruptions during operation.
Conjugate storage compartment
Assay runs with three different methods in parallel (e.g., tIgE, sIgE, and tryptase) for up to 3,200 tests.
Development solution compartment
Hold enough development solution onboard for 2,400 to 4,400 test doses.
Processing chambers are the core engine of Phadia Laboratory Systems
Climate-controlled reaction chambers enable access to the gold standard in allergy testing and IgE sensibilization tests in allergy diagnostics.
Canister storage shelf and fixed pipe/drain connections
Efficient and ergonomic fluid-handling options, adaptable to the laboratory’s infrastructure and workflow needs.
Barcode scanning of sample racks, samples, and ImmunoCAP carriers and reagents
Full traceability to ensure patient safety and minimize risk of manual errors.
System operation
Less than 15 minutes’ operator time need to setup the system and start the run to produce up to 240 tests per hour.
Instrument touchscreen, Phadia Prime Software, and status indicator light
Ease of use for the operator.
Stop solution compartment
Hold enough stop solution onboard for 2,400 determinations.
Hidden button for Modal Video
Phadia 1000 testing instrument left view
Phadia 1000 testing instrument right view
Flexibility
  • Large allergen test menu covering more than 550 whole allergens and mixes and 100 allergen components.
  • Automatic start-up and shut-down functionality.
  • User-configurable reflex testing functionality.
  • LIS connectivity via Phadia™ Prime software.
Capability
  • Throughput up to 240 results/hour.
  • Continuous random access.
  • Refrigerated on-board reagent storage.
  • 28-day stored calibration curve.
  • Connectable to Laboratory Automation Systems (LAS).*
  • Network multiple Phadia™ instruments for clustered** setup for efficient handling.
Reliability
  • Excellent consistency over time and between countries, systems, laboratories, and users.
  • Support and troubleshooting enhanced by the secure Phadia LabCommunity connectivity.
  • Quality Club: One of the largest global quality assessment programs.
*Contact your local Thermo Fisher Scientific representative for available LAS connectivity.
**Cluster setup allows several Phadia instruments networked together to be operated with one Phadia Prime computer.
Phadia 1000 instrument capability
Phadia 1000 testing instrument walk away time diagram
Explore the complete line of Phadia Laboratory Systems.
View all systems
See all tests that can be run on the Phadia 1000.
View tests
View all available tests and instruments.
See Catalog