Talk with Us
EliA assays for Rheumatoid Arthritis (RA)
Improve the confidence of RA diagnoses with EliATM rheumatoid arthritis assays in your lab.
A close-up of a shoe and ankle in mid-step with an overlay graphic of the anatomical bone structures.
Why you should add EliA assays for RA to your lab.
RA is the most common form of inflammatory arthritis (IA) and early management can help slow or prevent irreversible joint destruction. Laboratories can help close the care gap by offering the full profile of tests to help improve diagnostic outcomes. Providing comprehensive testing solutions for healthcare providers helps provide patients with diagnostic answers faster.
Steps to an accurate diagnosis 1-5
clipboard-icon
Patient history and symptoms
chevron icon pointing right
Pre-test probability
test-tube-icon
RA specific serology testing (e.g., CCP, RF IgM, RF IgA)
chevron icon pointing right
Post-test probability to narrow differential
magnifying glass icon
If appropriate, referral to rheumatology for further evaluation
chevron icon pointing right
Further evaluation
checkmark icon
Accurate diagnosis
Patient history and symptoms
Pretest probability
RA specific serology testing (e.g., CCP, RF IgM, RF IgA)
Post-test probability to narrow differential
If appropriate, referral to rheumatology for further evaluation
Further evaluation
Accurate diagnosis
What assays are part of the EliA rheumatoid arthritis portfolio?
The EliA™ rheumatoid arthritis portfolio contains assays that detect ACPA (anti-citrullinated peptide antibodies) using CCP (cyclic citrullinated peptide) antigens as well as isotype-specific assays for rheumatoid factors (RF) IgM and IgA, all of which are recommended for a well-informed patient diagnosis. 3,6-9
When CCP, RF IgM and IgA are combined they reach nearly 100% posivite predictive vaue (PPV). 5
Rheumatoid arthrisits probability in patients is best measured with IgM, IgA, and CCP testing. Increased positivity in all 3 tests equals increased risk.
Let's break that down:
EliA CCP assay 
CCP is a clinically specific marker to support in the diagnosis of RA, and its specificity for RA reduces false positive results.7
Alert icon
75% of initial RA testing in primary care does not include CCP. 11
Use of CCP assays helps increase the postive predictive value for testing. 5
A close-up of a bent knee with an overlay graphic of the anatomical bone structures.
EliA RF IgM and RF IgA assays
Combining EliATM RF IgM and EliATM RF IgA in laboratory analysis is vital in helping identify and quickly manage RA.
Less than 5% of non-RA patients are positive for both RF IgM and IgA.12
These are isotype-specific, which provides added value compared to total RF assays based on agglutination (e.g. nephelometric or turbidimetric RF assays). 12,13
Combining assays to increase RA diagnostic confidence 
Testing with EliATM CCP, EliA RF IgM, and EliA RF IgA assays in parallel allows you to identify patients with positivity to two or more analytes. This significantly increases the positive likelihood ratio (LR+) and the positive predictive value (PPV) of test results, as well as diagnostic confidence. 5,14,15
 Become a trusted source for RA diagnotics today.
 Talk with us to learn more.
Chat with us
Gain the benefits of Phadia Laboratory Systems
EliA rheumatoid arthritis assays are run on PhadiaTM Laboratory Systems, including the PhadiaTM 250 instrument.
Phadia 250 System runs tests for rheumatoid arthritis (RA)
 Let the Phadia 250 instrument be the backbone in maximizing your lab's workflow.
Balance of specificity and sensitivity
Antigens used in EliA™ assays are manufactured using innovative recombinant protein technology, which optimizes accessibility and purity, aiding clinicians in making impactful and appropriate decisions.16-18
High quality assays, aiding in antibody identification16-18
Even for difficult-to-detect antibodies highquality EliA™ antigens can help identify important antibodies associated with RA.5
Less false positives
High specificity due to the selection and use of recombinant antigens in this portfolio means laboratories can help reduce the number of false positives, aiding clinicians in prescribing appropriate therapies.16
Unlock your lab’s potential with EliA autoimmune assays and Phadia Laboratory Systems today.
References
  1. Chauhan K, Jandu JS, Brent LH, et al. Rheumatoid Arthritis. [Updated 2023 May 225]. In: StatPearls [Internet]. Treasure Island (FL): StatPearls Publishing; 2024 Jan-. Available from: https://www.ncbi.nlm.nih.gov/books/NBK441999/.
  2. Baker, J. (2024, March 21). Diagnosis and differential diagnosis of rheumatoid arthritis. UpToDate. https://www.uptodate.com/contents/diagnosis-and-differential-diagnosis-of-rheumatoid-arthritis?topicRef=17052&source=see_link .
  3. Littlejohn EA, Monrad SU. Early Diagnosis and Treatment of Rheumatoid Arthritis. Prim Care. 2018 Jun;45(2):237-255 *5
  4. Jonsson T et al. Elevation of only one rheumatoid factor isotype is not associated with increased prevalence of rheumatoid arthritis: a population based study. Scand J Rheumatol. 2000;29:190–191.
  5. Jaskowski TD, Hill HR, Russo, KL, et al. Relationship Between Rheumatoid Factor Isotypes and IgG Anti-Cyclic Citrullinated Peptide Antibodies. J Rheumatol. 2010;37:1582–1588. *11
  6. Aletaha D, Neogi T, et al. 2010 Rheumatoid arthritis classification criteria: an American College of Rheumatology/European League Against Rheumatism collaborative initiative. Arthritis Rheum. 2010 Sep;62(9):2569-81. doi: 10.1002/art.27584. PMID: 20872595.
  7. van Venrooij, Walther J et al. “Anti-CCP antibodies: the past, the present and the future.” Nature reviews. Rheumatology vol. 7,7 391-8. 7 Jun. 2011, doi:10.1038/nrrheum.2011.76.
  8. de Brito Rocha, S., Baldo, D.C. & Andrade, L.E.C. Clinical and pathophysiologic relevance of autoantibodies in rheumatoid arthritis. Adv Rheumatol 59, 2 (2019).
  9. Bizzaro, Nicola. “Antibodies to citrullinated peptides: a significant step forward in the early diagnosis of rheumatoid arthritis.” Clinical chemistry and laboratory medicine vol. 45,2 (2007): 150-7. doi:10.1515/CCLM.2007.027.
  10. Taylor P, Gartemann J, Hsieh J., et al. A Systematic Review of Serum Biomarkers Anti-Cyclic Citrullinated Peptide and Rheumatoid Factor as Tests for Rheumatoid Arthritis. Autoimmune Diseases, 2011, Article ID 815038, 18 pages.
  11. Joy V. et al (2022); Leveraging Real World Data to Identify Addressable Diagnostic Care Gaps in Rheumatoid Arthritis, In: International Congress on Autoimmunity; June 10 – 13; Athens, Greece.
  12. Jónsson, T et al. “Combined elevation of IgM and IgA rheumatoid factor has high diagnostic specificity for rheumatoid arthritis.” Rheumatology international vol. 18,3 (1998): 119-22. doi:10.1007/s002960050069.
  13. Bas, S et al. “Comparative study of different enzyme immunoassays for measurement of IgM and IgA rheumatoid factors.” Annals of the rheumatic diseases vol. 61,6 (2002): 505-10. doi:10.1136/ard.61.6.505
  14. Sieghart, Daniela et al. “Determination of Autoantibody Isotypes Increases the Sensitivity of Serodiagnostics in Rheumatoid Arthritis.” Frontiers in immunology vol. 9 876. 24 Apr. 2018, doi:10.3389/fimmu.2018.00876
  15. Internal Study. Data on File
  16. Orme ME, Andalucia C, Sjolander S, Bossuyt X. A comparison of a fluorescence enzyme immunoassay versus indirect immunofluorescence for initial screening of connective tissue diseases: Systematic literature review and meta-analysis of diagnostic test accuracy studies. Best Pract Res Clin Rheumatol. 2018;32(4):521-34.
  17. van der Pol P, Bakker-Jonges LE, Kuijpers J, Schreurs MWJ. Analytical and clinical comparison of two fully automated immunoassay systems for the detection of autoantibodies to extractable nuclear antigens. Clin Chim Acta. 2018;476:154-9.
  18. Alsaed OS, Alamlih LI, Al-Radideh O, Chandra P, Alemadi S, Al-Allaf AW. Clinical utility of ANA-ELISA vs ANA-immunofluorescence in connective tissue diseases. Sci Rep. 2021;11(1):8229.