Clarity in diagnosis. Confidence in patient care.
Early identification and testing are essential in managing food allergies. Food allergies are becoming increasingly prevalent, now impacting up to 1 in 10 children and adults.1
The use of ImmunoCAP™ specific IgE tests can provide accurate and reliable results that aid in the timely diagnosis and effective management of food allergies.
Food anaphylaxis is increasing worldwide, especially affecting children under the age of 5 years who are most at risk of requiring hospitalization.2 It’s never been more important for healthcare providers to be able to help patients determine which allergens are dangerous to them and to develop a personalized management plan to help them avoid those dangers.
42%
of children with food allergies have experienced a severe reaction.3
51%
of adults with food allergies have experienced a severe reaction.3
Patients with suspected food allergy should receive appropriate testing to support a proper diagnosis and to ensure that quality of life is not unnecessarily impaired by food restrictions.5 Of the 35% of patients who self-report having a food allergy, only 3.5% have a true clinical allergy.1
You play an essential role in early detection, initial testing for allergen triggers, guideline-recommended management and referral to specialists when necessary.
Early diagnosis confirmed by ImmunoCAP Specific IgE tests can guide appropriate intervention for food allergies.
ImmunoCAP Specific IgE tests are easily ordered in primary care, whereas skin prick testing (SPT) usually requires a referral to a specialist. The ImmunoCAP Specific IgE test portfolio, including whole allergens and allergen components, provides reproducible, quantitative results that can strengthen your judgment and sharpen your treatment strategies.
Specific IgE testing is one of the first-line diagnostic recommended by international guidelines.6
Supports patient management
ImmunoCAP allergen component tests aid in patient management by helping to identify which molecules a patient may be reacting to in whole allergens, such as peanuts, tree nuts, milk, eggs, pets and more.7
Safe and accessible
Irrespective of age, skin condition, antihistamine use or pregnancy, ImmunoCAP Specific IgE tests can be used for a variety of patient types and, unlike skin-prick testing, carries no risk of anaphylaxis.8,9
Guideline-driven
Testing for sensitization to single food allergens is endorsed by international guidelines for targeted, patient-centered care.
Determining if symptoms are related to IgE-mediated reactions is fundamental for developing an effective management plan. Sensitization alone is not sufficient for a diagnosis of food allergy, but must be consistent with the patient’s clinical history and symptoms.
IgE-mediated food allergy
Onset: Usually rapid, within minutes to two hours after eating the trigger food
Symptoms:
Examples: Peanut, tree nut and seafood allergies
Non-IgE-mediated food allergy
Onset: Usually delayed, appearing hours or days after ingesting the food
Symptoms:
Examples: Food protein-induced enterocolitis (FPIES), food protein-induced allergic proctocolitis (FPIAP) and food protein-induced enteropathy (FPE)
Key differences
Timing:
IgE reactions are immediate; non-IgE reactions are delayed.
Occurrence:
IgE-mediated responses are reproducible; non-IgE responses are inconsistent.
Immune mechanism:
IgE reactions are mediated by immunoglobulin E antibodies, while non-IgE reactions involve other parts of the immune system.
Severity & outcome:
IgE-mediated reactions can be severe and fatal (anaphylaxis), whereas non-IgE reactions are typically less acute, though they can significantly impact quality of life and growth.
Diagnosis:
IgE allergies can often be diagnosed with the help of skin prick or blood tests. Non-IgE allergies are harder to diagnose and often require an elimination diet and challenge to identify the culprit food.
Consult the Allergen Encyclopedia for more information on the “Big 9” food allergens.
Management of food allergy has evolved from a “wait and see” approach to more active interventions.11
You can provide dietary guidance and an initial treatment plan with the help of specific IgE whole allergen or allergen component test results. The molecular profile supports decisions about the safety of re-introducing a food, conducting an oral food challenge and prescribing immunomodulatory treatments.
Getting a thorough patient history and a list of symptoms is key in taking the next step and ordering a test.
If a history and symptoms indicate a potential food allergy, you can order a test to measure the sensitization to the specific foods suspected.
When specific foods aren't identified from history, it's important to test only for the most common food allergen types. Large, indiscriminate profiles that include many unnecessary food allergens are never appropriate.12
If available, the other important selection is to choose a profile with allergen component reflex in the name.
Allergen component reflex tests provide more detailed information than whole allergen tests alone. If a whole allergen comes back positive, the components of that allergen are automatically tested to get a more granular picture of the patient’s sensitization. This means:
You have the power to make a difference.
Learn more about allergen components.
Find ImmunoCAP test codes for a specific laboratory and order using your preferred method.
The laboratory will process the ImmunoCAP test on Phadia™ Laboratory Systems.
Assess and interpret ImmunoCAP test results to personalize allergen management.
Use these food allergy result interpretation guides for concise information on how to interpret ImmunoCAP Specific IgE test results as well as potential clinical implications to aid in your food allergy diagnosis and patient management plan.
Tools to aid in your diagnosis and patient management