Talk with Us
Hidden Button for comparing instuments
Phadia 2500+ series Made to maximize
Phadia 2500 instrument
Compare Instruments
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
When your laboratory has a growing demand for 10,000 or more tests per week, the Phadia™ 2500+ series* is made for you.
It maximizes your productivity with options to run both EliA™ and ImmunoCAP™ tests in parallel on a single lab instrument. Run any combination of tests on each sample. Continuous random access allows you to add samples, reagents, and consumables during the run without disrupting your high-throughput workflows. With the Phadia 2500+ series, rapidly increasing demands won’t slow you down.
The Phadia 2500+ series comprises three configuration options, giving you the flexibility to order your instrument to fit your testing needs whether for ImmunoCAP™ only (Phadia 2500), EliA™ only (Phadia 2500EE), or a combination of both (Phadia 2500E).
Phadia 2500 testing instrument closeup
Technician using Phadia 2500 testing instrument
Efficiency
  • Throughput up to 480 ImmunoCAP results/hour or up to 300 EliA results/hour.
  • Continuous random access.
  • Refrigerated on-board reagent storage.
  • 28-day stored calibration curve.
  • Onboard dilution (EliA).
  • Network multiple Phadia™ instrument for clustered** setup for efficient handling.
Flexibility
  • Large allergen test menu covering more than 500 whole allergens and mixes and 100 allergen components.
  • Broad, clinically relevant autoimmunity menu for evaluation of more than 50 of the most common autoimmune disease markers.
  • User-configurable reflex testing functionality.
  • LIS connectivity via Phadia™ Prime software.
Confidence
  • Excellent consistency over time and between countries, systems, laboratories, and users.
  • Support and troubleshooting enhanced by the secure Phadia LabCommunity connectivity.
  • Quality Club: One of the largest global quality assessment programs.
*The Phadia 2500+ series comprises the Phadia 2500, Phadia 2500E, and Phadia 2500EE instruments.
**Cluster setup allows several Phadia instruments networked together to be operated with one Phadia Prime computer.
Phadia 2500+ series capability
Phadia 2500 testing instrument walk away time diagram
Throughput up to 480 ImmunoCAP test results/hour or up to 300 EliA test results/hour.
Explore the complete line of Phadia Laboratory Systems.
View all systems
See all tests that can be run on the Phadia 2500+ series.
View tests
View all available tests and instruments.
See Catalog