| Phadia 200 | Phadia 250 | Phadia 1000 | Phadia 2500+ series | Phadia 5000+ series | |
| ImmunoCAP | Allergy | Allergy | Allergy | Allergy | Allergy |
| EliA | Autoimmunity |
Autoimmunity | N/A | Autoimmunity | Autoimmunity |
| System Description | Fully-automated closed immunoassay system | Fully-automated closed immunoassay system | Fully-automated closed immunoassay system | Fully-automated closed immunoassay system | Fully-automated closed immunoassay system |
| Processing | Batch | Continuous | Continuous | Continuous | Continuous |
| Maximum Throughput | Up to 96 test results per batch | Up to 60 results/hour | Up to 240 results/hour | Up to 480 results/hour | Up to 960 results/hour |
| Sample Types | Serum, Plasma | Serum, plasma | Serum, plasma | Serum, plasma | Serum, plasma |
| Sample Tubes for Sample Rack w/ 10 positions |
Height 42-100mm Diam. 10-16mm |
Height 50-110mm Diam. 10-17mm |
Height 50-105mm Diam. 10-17mm |
N/A | N/A |
| Sample Tubes for Sample Rack w/ 5 positions |
N/A | N/A | N/A | Height 70-105mm Diam. 10-17mm |
Height 70-105mm Diam. 10-17mm |
| Sample Barcode Types | Code 39, Code 93, Code 128, Codabar (NW-7), COOP 2 of 5*, EAN-8, Industrial 2 of 5*, Interleave 2–5 *Not for Phadia 200 and Phadia 1000 Use. Phadia 200 not Validated with EAN-8 either |
Code 39, Code 93, Code 128, Codabar (NW-7), COOP 2 of 5*, EAN-8, Industrial 2 of 5*, Interleave 2–5 *Not for Phadia 200 and Phadia 1000 Use. Phadia 200 not Validated with EAN-8 either |
Code 39, Code 93, Code 128, Codabar (NW-7), COOP 2 of 5*, EAN-8, Industrial 2 of 5*, Interleave 2–5 *Not for Phadia 200 and Phadia 1000 Use. Phadia 200 not Validated with EAN-8 either |
Code 39, Code 93, Code 128, Codabar (NW-7), COOP 2 of 5*, EAN-8, Industrial 2 of 5*, Interleave 2–5 *Not for Phadia 200 and Phadia 1000 Use. Phadia 200 not Validated with EAN-8 either |
Code 39, Code 93, Code 128, Codabar (NW-7), COOP 2 of 5*, EAN-8, Industrial 2 of 5*, Interleave 2–5 *Not for Phadia 200 and Phadia 1000 Use. Phadia 200 not Validated with EAN-8 either |
| Sample Dilution | Allergy Autoimmunity |
Allergy Autoimmunity |
N/A | Autoimmunity | Autoimmunity |
| Level and Clot Detection | Yes | Yes | Yes | Yes | Yes |
| Reflex Testing | Yes | Yes | Yes | Yes | Yes |
| STAT Samples | No | Yes | No | Yes | Yes |
| Reagents | General ImmunoCAP and EliA reagents available on board. Loaded batch wise with ImmunoCAP or EliA reagents | General ImmunoCAP and EliA reagents available on board are shared | General ImmunoCAP and EliA reagents available on board are shared | General ImmunoCAP and EliA reagents available on board are shared | General ImmunoCAP and EliA reagents available on board are shared |
| Type of calibration | One Isotype master curve per assay group (IgG, IgE, IgM, IgA) | One Isotype master curve per assay group (IgG, IgE, IgM, IgA) |
One Isotype master curve per assay group (IgG, IgE, IgM, IgA) |
One Isotype master curve per assay group (IgG, IgE, IgM, IgA) |
One Isotype master curve per assay group (IgG, IgE, IgM, IgA) |
| Calibration stability | 28 days | 28 days | 28 days | 28 days | 28 days |
| Water Requirements for Rinse Solution | Clinical laboratory reagent water (CLRW, may be used in place of Type I and Type II water)* | Clinical laboratory reagent water (CLRW, may be used in place of Type I and Type II water)* | Clinical laboratory reagent water (CLRW, may be used in place of Type I and Type II water)* | Clinical laboratory reagent water (CLRW, may be used in place of Type I and Type II water)* | Clinical laboratory reagent water (CLRW, may be used in place of Type I and Type II water)* |
| Remote Troubleshooting and Support | Phadia LabCommunity |
Phadia LabCommunity | Phadia LabCommunity | Phadia LabCommunity | Phadia LabCommunity |
| Ambient Operating Temperature | 18–32 °C |
18–32 °C | 18–32 °C | ImmunoCap 18–32 °C EliA 18-29 °C |
ImmunoCap 18–32 °C EliA 18-29 °C |
| Electrical Requirements | 110-240V 50/60Hz |
100 V, 120 V, 220 V, 230 V, 240 V ±10% 50/60 Hz |
200 V, 220 V, 230 V, 240 V±10% 50/60 Hz |
200 V, 208 V, 230 V, 240 V±10% 50/60 Hz |
200 V, 208 V, 230 V, 240 V±10% 50/60 Hz |
| Dimensions (HxWxD) | 55 x 80 x 60 cm (22 x 32 x 24 in) | 186x127x75 cm w/ table 73x50x30 in |
215x183x105 cm 85x72x42 in |
195x411x135 cm 77x162x53 in |
195x619x135 cm 77x244x53 in |
| Weight (empty) | 47 kg/ 104 lb. | 220 kg / 485 lb | 688 kg / 1516 lb | 2000 kg / 4409 lb | 3342 kg / 7367 lb |
*Clinical laboratory reagent water (CLRW) may be used in place of Type II Laboratory Grade water; Approved Guideline-Fourth Edition.
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
It maximizes your productivity with options to run both EliA™ and ImmunoCAP™ tests in parallel on a single lab instrument. Run any combination of tests on each sample. Continuous random access allows you to add samples, reagents, and consumables during the run without disrupting your high-throughput workflows. With the Phadia 2500+ series, rapidly increasing demands won’t slow you down.
The Phadia 2500+ series comprises three configuration options, giving you the flexibility to order your instrument to fit your testing needs whether for ImmunoCAP™ only (Phadia 2500), EliA™ only (Phadia 2500EE), or a combination of both (Phadia 2500E).
*The Phadia 2500+ series comprises the Phadia 2500, Phadia 2500E, and Phadia 2500EE instruments.
**Cluster setup allows several Phadia instruments networked together to be operated with one Phadia Prime computer.
Throughput up to 480 ImmunoCAP test results/hour or up to 300 EliA test results/hour.
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.