Contact Us
EliA assays for rheumatoid arthritis (RA)
Improve the confidence of RA diagnoses with EliA™ rheumatoid arthritis assays in your lab.
A close-up of a shoe and ankle in mid-step with an overlay graphic of the anatomical bone structures.
Why you should add EliA assays for RA to your lab.
RA is the most common form of inflammatory arthritis (IA) and early management can help slow or prevent irreversible joint destruction.3 Laboratories can help close the care gap by offering the full profile of tests to help improve diagnostic outcomes. Delivering comprehensive testing solutions for healthcare providers helps ensure diagnostic answers faster.
Steps to an accurate diagnosis 1-5
clipboard-icon
Primary care visit: Patient history and symptoms
chevron icon pointing right
Pre-test probability
test-tube-icon
RA specific serology testing (e.g. CCP, RF IgM, RF IgA)
chevron icon pointing right
Post-test probability to narrow differential diagnosis
magnifying glass icon
If appropriate, referral from primary care to specialist (rheumatology) for further evaluation
chevron icon pointing right
Further evaluation
checkmark icon
Accurate diagnosis
Patient history and symptoms
Pretest probability
RA specific serology testing (e.g., CCP, RF IgM, RF IgA)
Post-test probability to narrow differential
If appropriate, referral to rheumatology for further evaluation
Further evaluation
Accurate diagnosis
What assays are part of the EliA rheumatoid arthritis portfolio?
The EliA™ rheumatoid arthritis portfolio contains assays that detect ACPA (anti-citrullinated peptide antibodies) using CCP (cyclic citrullinated peptide) antigens as well as isotype-specific assays for rheumatoid factors (RF) IgM and IgA, all of which are recommended for a well-informed patient diagnosis. 3,6-9
When CCP, RF IgM and IgA are combined they reach nearly 100% posivite predictive value (PPV) for RA diagnosis. 5
Rheumatoid arthrisits probability in patients is best measured with IgM, IgA, and CCP testing. Increased positivity in all 3 tests equals increased risk.
Let's break that down
EliA CCP assay 
CCP is a clinically specific marker to support in the diagnosis of RA, and its specificity for RA reduces false positive results.7
Alert icon
In the US, 75% of initial RA testing in primary care does not include CCP. 11
Use of CCP assays helps increase the postive predictive value for testing. 5
A close-up of a bent knee with an overlay graphic of the anatomical bone structures.
EliA RF IgM and RF IgA assays
Combining EliA™ RF IgM and EliA™ RF IgA in laboratory analysis is important in helping identify and quickly manage RA.
4% of non-RA patients are positive for both RF IgM and IgA.12
These are isotype-specific tests, which provide added value compared to total RF assays based on agglutination (e.g. nephelometric or turbidimetric RF assays). 12,13
Combining assays to increase RA diagnostic confidence 
Testing with EliA CCP, EliA RF IgM and EliA RF IgA assays in parallel allows you to identify patients with positivity to two or more analytes. This significantly increases the positive likelihood ratio (LR+) and the positive predictive value (PPV) of test results, as well as diagnostic confidence. 5,14,15
 Become a trusted source for RA diagnostics today.
Contact us to learn more.
Send
EliA rheumatoid arthritis laboratory testing algorithm
Applying a standardized testing profile with EliA assays for RA allow for convenient and efficient testing for primary care and specialists alike. The following testing algorithm is based on relevant guidelines and selected references.5,6,14,16 It aims to guide the integration of EliA tests in routine laboratory work and the interpretation of assay results to support diagnoses and planning of further suitable follow-up evaluation.
Gain the benefits of Phadia Laboratory Systems
EliA rheumatoid arthritis assays are run on Phadia™ Laboratory Systems, including the Phadia™ 250 instrument.
Phadia 250 System runs tests for rheumatoid arthritis (RA)
 Let the Phadia 250 instrument be the backbone in maximizing your lab's workflow.
clock icon
Balance of specificity and sensitivity
Antigens used in EliA™ assays are manufactured including innovative recombinant protein technology, which optimizes accessibility and purity, aiding clinicians in making impactful and appropriate decisions.17
High quality assays, aiding in antibody identification
Even for difficult-to-detect antibodies high quality EliA™ antigens can help identify important antibodies associated with RA.5
Less false positives
The high specificity of EliA assays can help laboratories to reduce false positives. This supports clinicians in making accurate diagnoses and prescribing appropriate therapies.17, 18
Unlock your lab’s potential with EliA autoimmune assays and Phadia Laboratory Systems today.
References
  1. Chauhan K, et al. Rheumatoid Arthritis. In: StatPearls [Internet]. Treasure Island (FL): StatPearls Publishing; 2023. Available from: https://www.ncbi.nlm.nih.gov/books/NBK441999/
  2. Baker J, et al. Diagnosis and differential diagnosis of rheumatoid arthritis. UpToDate; 2024. Available from: https://www.uptodate.com/contents/diagnosis-and-differential-diagnosis-of-rheumatoid-arthritis
  3. Littlejohn EA, et al. Early Diagnosis and Treatment of Rheumatoid Arthritis. Prim Care. 2018;45(2):237-255.
  4. Jonsson T, et al. Elevation of only one rheumatoid factor isotype is not associated with increased prevalence of rheumatoid arthritis: a population based study. Scand J Rheumatol. 2000;29(3):190-191.
  5. Jaskowski TD, et al. Relationship Between Rheumatoid Factor Isotypes and IgG Anti-Cyclic Citrullinated Peptide Antibodies. J Rheumatol. 2010;37(8):1582-1588.
  6. Aletaha D, et al. 2010 Rheumatoid arthritis classification criteria: an American College of Rheumatology/European League Against Rheumatism collaborative initiative. Arthritis Rheum. 2010;62(9):2569-2581.
  7. van Venrooij WJ, et al. Anti-CCP antibodies: the past, the present and the future. Nat Rev Rheumatol. 2011;7(7):391-398.
  8. de Brito Rocha S, et al. Clinical and pathophysiologic relevance of autoantibodies in rheumatoid arthritis. Adv Rheumatol. 2019;59:2.
  9. Bizzaro N, et al. Antibodies to citrullinated peptides: a significant step forward in the early diagnosis of rheumatoid arthritis. Clin Chem Lab Med. 2007;45(2):150-157.
  10. Taylor P, et al. A Systematic Review of Serum Biomarkers Anti-Cyclic Citrullinated Peptide and Rheumatoid Factor as Tests for Rheumatoid Arthritis. Autoimmune Dis. 2011;2011:815038.
  11. Joy V, et al. Leveraging Real World Data to Identify Addressable Diagnostic Care Gaps in Rheumatoid Arthritis, In: International Congress on Autoimmunity; 2022.
  12. Jónsson T, et al. Combined elevation of IgM and IgA rheumatoid factor has high diagnostic specificity for rheumatoid arthritis. Rheumatol Int. 1998;18(3):119-122.
  13. Bas S, et al. Comparative study of different enzyme immunoassays for measurement of IgM and IgA rheumatoid factors. Ann Rheum Dis. 2002;61(6):505-510.
  14. Sieghart D, et al. Determination of Autoantibody Isotypes Increases the Sensitivity of Serodiagnostics in Rheumatoid Arthritis. Front Immunol. 2018;9:876.
  15. Internal Study. Data on File
  16. Steiner G, et al. Classification of rheumatoid arthritis: is it time to revise the criteria? RMD Open. 2024;10(2):e003851
  17. Schmitt J, Papisch W. Recombinant autoantigens. Autoimmun Rev. 2002;1(1-2):79-88.
  18. Jackson BR, et al. The dangers of false-positive and false-negative test results: false-positive results as a function of pretest probability. Clin Lab Med. 2008;28(2):305-319.