Talk with Us
Helios Automated IFA System
Load your slide and confidently walk away with the Helios systems’ organized and automated IFA diagnostics.
   As the official US distributor of Helios® Automated IFA Systems, Thermo Fisher Scientific is committed to helping you minimize downtime and strengthen your output through exceptional customer service.
Specifications
Size: 30 in x 25 in x 22.5 in (tabletop)
Sample capacity: Up to 190 barcoded samples
Slide capacity: Up to 20 slides
Software capabilities: FDA-cleared pattern recognition for ANA HEp-2, c-ANCA, p-ANCA, dsDNA (Crithidia luciliae)
Sample tubes for sample rack: Height: 50-110mm, Diameter: 10-17mm
IFA test capacity: Up to four different tests per run
Testing capabilities: AESKUSLIDES® ANA HEp-2, AESKUSLIDES® ANCA E, AESKUSLIDES® ANCA F, AESKUSLIDES® nDNA (Crithidia luciliae)
Interested in adding the Helios system to your laboratory or have questions?
Laboratory advantages of Helios Automated IFA systems
Impactful results for patients, optimized IFA testing workflow for you.
Give your pipette a break with IFA automation
Load up to 190 samples per run and confidently walk away as Helios systems take care of every step of the IFA diagnostic methodology, including dilution, incubation, washing, and even pipetting mounting media before imaging.
Saving time with automated microscopy and IFA test result processing
With on-board automatic image capturing, positive pattern recognition, and suggested endpoint titer, there’s no need to move samples to a new instrument to be read or sort through negative results when it comes time to review.
Bringing peace of mind to a sensitive and subjective methodology
No matter the microclimate of the lab, the HTC modual of the Helios systems maintain the optimal internal temperature and humidity during processing, ensuring high-quality results for healthcare providers and patients.
Give your pipette a break with IFA automation
Load up to 190 samples per run and confidently walk away as Helios systems take care of every step of the IFA diagnostic methodology, including dilution, incubation, washing, and even pipetting mounting media before imaging.
Saving time with automated microscopy and IFA test result processing
With on-board automatic image capturing, positive pattern recognition, and suggested endpoint titer, there’s no need to move samples to a new instrument to be read or sort through negative results when it comes time to review.
Bringing peace of mind to a sensitive and subjective methodology
No matter the microclimate of the lab, the HTC modual of the Helios systems maintain the optimal internal temperature and humidity during processing, ensuring high-quality results for healthcare providers and patients.
Take the uncertainty and tediousness out of IFA testing.
Learn how Helios systems can elevate your lab.
Let's talk
Pair Helios Automated IFA Systems with Phadia Laboratory Systems to streamline your autoimmune workflow.
Gain longer walkaway times and faster turnaround by incorporating Helios systems’ IFA test results with EliA™ autoimmune assays processed on Phadia™ Laboratory Systems.
Together, flexible Helios systems and scalable Phadia systems help to optimize your autoimmune resulting and patient outcomes by providing quality results faster.
Phadia 250 Instrument
Reliability meets versatility on the Phadia™ 250 system, which runs EliA tests and ImmunoCAP™ tests.
Learn more
Phadia 2500+ Series*
The Phadia™ 2500+ series meets the demand for high throughput.
Learn more
*The Phadia 2500+ comprises the Phadia™ 2500, Phadia™ 2500E, and Phadia™ 2500EE instruments.
Scalable testing for laboratories of all sizes.
Complementing IFA testing and helping to drive autoimmune automation by incorporating solid-phase assays for over 30 of the most common autoimmune disease markers. Explore specific disease state offerings:
EliA Autoimmunity Solutions
Automatic immunofluorescence pattern recognition for four substrates, processed on an FDA-cleared instrument.
Helios systems not only offer automated immunofluorescence test processing but are also FDA-cleared to recognize patterns from positive ANA, ANCA, and dsDNA IFA tests.1,2
ANCA-E
ANCA-F
HEp-2
Crithidia luciliae
Adopt the efficiency of Helios systems and unlock autoimmune automation and optimized workflow today.
References
  1. Evans R. AESKU receives FDA clearance Helios automated IFA system [internet]. Medical Design & Outsourcing; 2016 Aug 1 [cited 2024 June 18]. Available from: https://www.medicaldesignandoutsourcing.com/fda-clearance-helios-auto-ifa-system/.
  2. 510(k) Substantial Equivalence Determination Decision Memorandum [internet]. US Food and Drug Administration [cited 2024 June 18]. Available from: https://www.accessdata.fda.gov/cdrh_docs/reviews/K153117.pdf.