Whole Allergen
d74 Euroglyphus maynei
d74 Euroglyphus maynei Scientific Information
Type:
Whole Allergen
Display Name:
Euroglyphus maynei
Allergen code:
d74
Family:
Pyroglyphidae
Species:
Euroglyphus maynei
Route of Exposure:
Inhalation (2)
Source Material:
Mite feces
Latin Name:
Euroglyphus maynei
Other Names:
Mayne's house dust mite (1)
Summary
Euroglyphus maynei is one of the Pyroglyphinae family of dust mites, known commonly as Mayne's house dust mite.  E. maynei thrives in humid conditions within the home, notably in bedding, carpets and furniture and is found in dust in around 30–40% of homes. Sensitization to house dust mites (HDMs) represents a major cause of respiratory allergy worldwide in children, adolescents, and adults.
Allergen
Nature
E. maynei is a common species of HDM that thrives at relative humidity of >50%. HDM adults develop from eggs within 3–4 weeks under humid conditions at 25–30°C, and females produce 40–80 eggs during their 4–6-week lifespan (3).
Adult mites are found predominantly in bedding, carpets and furniture (4); a study of Polish homes found E. maynei to be the most abundant species of mite in in bedroom floors (175.0 specimens per gram) and upholstered furniture in the living room (200 E. maynei per gram) (5).
In the homes of patients with rhinitis and asthma, mites were found in 466 of 500 (93.2%) dust samples from 125 houses, with a total of 7,710 mite specimens isolated (6). Of these, 7,404 specimens were mites of the suborder Astigmata and 63.2% of those were E. maynei (6).
Taxonomy 
Taxonomic tree of Euroglyphus maynei (7)
Domain
Eukaryota
Kingdom
Metazoa
Phylum
Arthropoda
Subphylum
Chelicerata
Class
Arachnida
Subclass
Acari
Order
Astigmata
Family
Pyroglyphidae
Genus
Euroglyphus
Tissue
Mite feces are the most relevant source of allergens (8).
Epidemiology
Worldwide distribution 
Approximately 20% of the population in industrialized countries are sensitized to HDM allergens (9).
Environmental Characteristics
Living environment
Mites require warm, dark, humid conditions (10). E. maynei often co-inhabit with Dermatophagoides species of HDM (11).
Worldwide distribution 
E. maynei can be found worldwide, in humid regions (3).
Route of Exposure
Main 
Inhalation (2).
Secondary
Ingestion and contact (10).
Clinical Relevance
Mites are the sources of potent allergens that sensitize and induce IgE-mediated allergic reactions (12). HDMs are a major cause of atopic sensitization with symptoms induced by direct contact (conjunctivitis, eczema), inhalation (rhinitis, asthma, eczema), and ingestion (urticaria, anaphylaxis) (10).
Sensitization to house dust mites (HDMs) is a major cause of respiratory allergy worldwide in children, adolescents, and adults (2).
Prevention and Therapy
Prevention strategies
Several dust mite avoidance measures have been proposed, including mattress and pillow encasings, high efficiency particulate air filtration vacuum cleaners, air purification, acaricides, humidity control, and physical removal of mite reservoirs. The most common strategies that have been used to control domestic allergen exposure are physical barriers such as covers for pillows, duvet, and mattresses (2). 
Molecular Aspects
Allergenic molecules
Fifteen different allergens have been identified in E. maynei extracts; all of which have demonstrated moderate or strong IgE binding to at least one serum from 32 E. maynei-sensitized patients (13).
Table adapted from Allergome.org (14)
Allergen
Type
Mass (kDa)
Eur m 1
Cysteine protease
36.3
Eur m 2
NPC2 family
15.7
Eur m 3
Trypsin
28
Eur m 4
α-amylase
60
Eur m 10
Tropomyosin
n/a
Eur m 14
Apolipophorin
191.3
Eur m 29
Isomerase
35.6
Eur m 30
Ferritin
20.5
Eur m 31
Actin binding
17.5
Eur m 33
α-tubulin
50
Eur m 36
Unknown
17
Eur m 38
Bacterial lytic enzyme
16
Cross-reactivity
E. maynei produces an allergen that cross reacts with Der P1 and shows some cross-reactivity with Dermatophagoides pteronyssinus and Dermatophagoides farinae (Colloff, 2009). Antigens from E. maynei are also cross-reactive with some antigens from the ectoparasitic scabies mite, Sarcoptes scabiei (11). Cross-reactivity due to tropomyosin and haemocyanin among shrimp, cockroach, and house dust has been suggested (15).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer 
Last reviewed:May 2022
References
  1. Uniprot.org. Taxonomy - Euroglyphus maynei (Mayne's house dust mite) 2021 [cited 2022 04.01.22]. Available from: https://www.uniprot.org/taxonomy/6958.
  2. Zuiani C, Custovic A. Update on House Dust Mite Allergen Avoidance Measures for Asthma. Curr Allergy Asthma Rep. 2020;20(9):50.
  3. Gill NK, Dhaliwal AK. Seasonal Variation of Allergenic Acarofauna From the Homes of Allergic Rhinitis and Asthmatic Patients. Journal of Medical Entomology. 2017;55(2):262-8.
  4. Aggarwal P, Senthilkumaran S. Dust Mite Allergy.  StatPearls. Treasure Island (FL): StatPearls Publishing
  5. Copyright © 2021, StatPearls Publishing LLC.; 2021.
  6. Hajduga-Staśko B, Pawełczyk O, Solarz K. Comparison of the domestic mites abundance in dwellings on selected urban and rural areas of the Zawiercie district (south-west Poland). Ann Parasitol. 2020;66(3):319-29.
  7. Gill NK, Dhaliwal AK. Seasonal Variation of Allergenic Acarofauna From the Homes of Allergic Rhinitis and Asthmatic Patients. J Med Entomol. 2018;55(2):262-8.
  8. Interpro. Euroglyphus maynei 2021 [cited 2022 04.01.22]. Available from: https://www.ebi.ac.uk/interpro/taxonomy/uniprot/6958/.
  9. Erban T, Rybanska D, Harant K, Hortova B, Hubert J. Feces Derived Allergens of Tyrophagus putrescentiae Reared on Dried Dog Food and Evidence of the Strong Nutritional Interaction between the Mite and Bacillus cereus Producing Protease Bacillolysins and Exo-chitinases. Frontiers in Physiology. 2016;7.
  10. Chang YC, Hsiao YM, Hung SC, Chen YW, Ou CC, Chang WT, et al. Alleviation of Dermatophagoides microceras-induced allergy by an immunomodulatory protein, FIP-fve, from Flammulina velutipes in mice. Biosci Biotechnol Biochem. 2015;79(1):88-96.
  11. Miller JD. The Role of Dust Mites in Allergy. Clinical Reviews in Allergy & Immunology. 2019;57(3):312-29.
  12. Rider S, Morgan M, Arlian L. Allergen homologs in the Euroglyphus maynei draft genome. PLOS ONE. 2017;12:e0183535.
  13. van Hage-Hamsten M, Johansson SGO. Storage mites. Experimental & Applied Acarology. 1992;16(1):117-28.
  14. Arlian LG, Rapp CM, Fernandez-Caldas E. Allergenicity of Euroglyphus maynei and its cross-reactivity with Dermatophagoides species. J Allergy Clin Immunol. 1993;91(5):1051-8.
  15. Allergome.org. EUROGLYPHUS MAYNEI 2021 [cited 2022 04.01.21]. Available from: http://www.allergome.org/script/search_step2.php.
  16. Lin IH, Tsai MC, Chen JP, Fu LS. Allergic children with extremely high total IgE but no allergen identified in the initial screening panel. J Microbiol Immunol Infect. 2021;54(3):474-81.