Type:
Whole Allergen
Whole Allergen
Whole Allergen
Ferret epithelium
e217
Mustelidae
Mustela putorius
Ferret, Household Ferret, Polecat
Common in the Old World, Ferrets have been used for centuries to hunt Rats, Mice, and Rabbits. They are ferocious, and may attack animals much larger than themselves. Domestic Ferrets are found in many colours, including albino, brown, and black. They are Weasel-shaped, and, like all mustelids, have well-developed anal scent glands. Polecats help to control rodent populations in the wild. They have also been hunted for their fur, which is considered valuable, though not as valuable as that of other mustelids such as Mink or Ermine.
IgE mediated reactions
Asthma, allergic rhinitis, allergic conjunctivitis and contact dermatitis (1).
A 41-year-old male experienced a near fatal asthma episode after washing his Ferret. He had experienced pruritis and erythema of the skin where he had been in contact with the animal (1)
Specific serum IgE from a patient with Ferret allergy was shown to bind to 4 protein bands (103, 81, 28.8 and 14.8 kD) in the male and female urine but none in the hair. IgG-depleted serum bound to 2 additional bands (213 and 41.2 kD) in the urine and to 2 bands (81 and 10.1 kD) in the hair (1). No allergens have been characterised yet.
In a report on Ferret allergy, the authors note that in a Mink allergic subject, specific IgG to Ferret interfered with the specific IgE measurement; it is hypothesized that the Mink-allergic subject's serum contained similar competing antibodies (1). Mink is a mammal of the same family as Ferret.
Last Reviewed- April 2022
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.