Table of Contents
Component
e229 Can f 4
e229 Can f 4 Scientific Information
Type:
Component
Name; WHO/IUIS:
Can f 4
Biological function:
Lipocalin
Allergen code:
e229
Molecular Weight:
18 kDa
Route of Exposure:
Inhalation
Source Material:
Dog Dander
Summary
Canis familiaris allergen 4 (Can f 4) is the most abundantly detected allergen in dog fur and a major allergen component of dog hair and dander extracts. Can f 4 has at least two isoforms and is strongly affected by denaturation. These variations in molecular structure may contribute to the highly heterogeneous IgE reactivity (46–81%) seen in studies from different geographical regions. Sensitization to Can f 4 is significantly associated with asthma, and can serve as a marker for clinically relevant dog allergy. Specific IgE tests for Can f 4 became available in 2019 and, when tested in combination with other dog allergens, Can f 4 improves the reliability of dog allergy diagnosis.
Epidemiology
Worldwide distribution
Dog-allergic individuals are sensitive to a number of proteins found in dog hair and dander extracts (1). Can f 4 is considered a major allergen component of dog dander and is important for over 80% of people with dog allergies for IgE binding (2, 3). Can f 4 T-cell responses have been shown to be a TH2-deviated memory response in allergenic but not nonallergenic individuals (4).
An early assessment of recombinant dog allergens noted 60% (15/25) of dog dust-sensitized subjects exhibited IgE reactivity to an “18 kDa component”, later identified as Can f 4, in immunoblotting (5). In contrast, Mattsson et al. found that only 35% of dog-allergic individuals were sensitized to recombinant Can f 4 (rCan f 4) as examined by ImmunoCAP (6). In this study, the magnitude of IgE antibody binding to Can f 4 was comparable to that of Can f 1, greater than to Can f 2 and Can f 3, and lower than to Can f 5 (6).
Can f 4 has at least two isoforms, and the overall IgE reactivity of natural Can f 4 depends strongly on the integrity of the allergen’s conformation (3, 7). Geographic variation in the prevalence of sensitivity to Can f 4 ranges from 46% to 81%, suggesting a difference in reaction to the isoforms (7).
Environmental Characteristics
Source and tissue
Can f 4 is produced in the tongue and found principally in dog dander and fur (7, 8). Can f 4 is the most abundantly detected allergen in dog fur samples (7).
Clinical Relevance
Disease severity
Sensitization to lipocalins such as Can f 4 is significantly associated with asthma (9). Sensitization to Can f 4 and Can f 6 can serve as a marker for clinically relevant dog allergy (10). Kack et al. used nasal provocation tests (NPTs) to demonstrate a positive association between sensitization to dog allergen components and clinical symptoms of dog allergy in children aged 10–18 years (10). In this study, sensitization to the lipocalins Can f 4 and Can f 6, and to the serum albumin Can f 3, was significantly associated with a positive NPT result in the unadjusted logistic regression analysis (10). After adjusting for the effect of the other protein groups, only sensitization to lipocalins was shown to be associated with an increased risk for a positive NPT result (10). Additionally, the likelihood of a positive NPT result increased with sensitization to an increasing number of dog allergen components; multisensitization to all six investigated dog allergen components (Can f 1, Can f 2, Can f 3, Can f 4, Can f 5, and Can f 6) conferred the overall highest risk for a positive reaction (10).
Molecular Aspects
Allergenic Molecules
Humans mutually exchange innate immune molecules such as lipocalins with their domestic animals via skin shedding and secretions (2). Lipocalins comprise an important family of proteins that dominate the respiratory mammalian allergens, and usually carry lipids (or other hydrophobic/amphiphilic compounds) within a large calyx-like cavity formed by a characteristic molecular β-barrel fold (2). Ligand specificity of lipocalins is regulated by amino acid residues that line their binding pocket (2). Allergenic lipocalins have been especially identified in furry animals, and four of the six currently-identified dog allergens are lipocalins (Can f 1, Can f2, Can f 4 and Can f 6) (11).
Mattsson et al. were the first to purify, clone, and characterize Can f 4, an IgE-reactive, 18 kDa lipocalin-like protein that is distinct from Can f 1 and Can f 2 (6). The IgE binding capacity of immunoaffinity-purified natural Can f 4 is strongly affected by denaturation, which could be a factor accounting for conflicting results regarding IgE reactivity to Can f 4 in various studies (3). Mattsson et al. considered that isoform variability of Can f 4 may have contributed to heterogeneity observed during purification of the natural protein, even though no evidence of amino acid sequence variability was observed in DNA sequencing (6).
Cross-reactivity
Can f 4 displays 38-39% sequence identity to bovine and porcine odorant-binding proteins, and 25-29% identity to several known mammalian allergens including Equ c 1, Mus m 1, Rat n 1, and Fel d 4 (6). Despite belonging to the same protein family, Can f 4 showed only 24% and 26% amino acid sequence identity to Can f 1 and Can f 2, respectively (6).
Diagnostic Relevance
In vitro Diagnosis
The clinical utility of specific IgE test to Can f 4 has not been studied extensively since it became available for clinical use in 2019. However, it appears that assessment of Can f 4 in combination with other dog allergens improves the reliability of allergy tests in dog allergy (3). rCan f 4 displayed close biochemical and immunological similarity to purified natural Can f 4, and bound IgE antibodies from 35% of dog-allergic subjects (6). Can f 4 may be relevant as an independent sensitizer in dog allergy, however, taken together, the levels of IgE to five dog allergens (Can f 1, Can f 2, Can f 3, Can f 4, and Can f 5) amounted on average to 93% of that to dog dander (6). rCan f 4 brings the panel of available dog allergens closer to completion and will be important in component-resolved diagnostics in allergy to animal epithelial allergens (6). 
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Magnus Borres
Last reviewed: December 2020
References
  1. Ford AW, Alterman L, Kemeny DM. The allergens of dog. I. Identification using crossed radio-immunoelectrophoresis. Clin Exp Allergy. 1989;19(2):183-90.
  2. Jensen-Jarolim E, Pacios LF, Bianchini R, Hofstetter G, Roth-Walter F. Structural similarities of human and mammalian lipocalins, and their function in innate immunity and allergy. Allergy. 2016;71(3):286-94.
  3. Rytkönen-Nissinen M, Saarelainen S, Randell J, Häyrinen J, Kalkkinen N, Virtanen T. IgE Reactivity of the Dog Lipocalin Allergen Can f 4 and the Development of a Sandwich ELISA for Its Quantification. Allergy Asthma Immunol Res. 2015;7(4):384-92.
  4. Rönkä AL, Kinnunen TT, Goudet A, Rytkönen-Nissinen MA, Sairanen J, Kailaanmäki AH, et al. Characterization of human memory CD4(+) T-cell responses to the dog allergen Can f 4. J Allergy Clin Immunol. 2015;136(4):1047-54.e10.
  5. Saarelainen S, Taivainen A, Rytkönen-Nissinen M, Auriola S, Immonen A, Mäntyjärvi R, et al. Assessment of recombinant dog allergens Can f 1 and Can f 2 for the diagnosis of dog allergy. Clin Exp Allergy. 2004;34(10):1576-82.
  6. Mattsson L, Lundgren T, Olsson P, Sundberg M, Lidholm J. Molecular and immunological characterization of Can f 4: a dog dander allergen cross-reactive with a 23 kDa odorant-binding protein in cow dander. Clin Exp Allergy. 2010;40(8):1276-87.
  7. Wintersand A, Asplund K, Binnmyr J, Holmgren E, Nilsson OB, Gafvelin G, et al. Allergens in dog extracts: Implication for diagnosis and treatment. Allergy. 2019;74(8):1472-9.
  8. Hilger C, Kuehn A, Hentges F. Animal lipocalin allergens. Curr Allergy Asthma Rep. 2012;12(5):438-47.
  9. Schoos AM, Kattan JD, Gimenez G, Sampson HA. Sensitization phenotypes based on protein groups and associations to allergic diseases in children. J Allergy Clin Immunol. 2016;137(4):1277-80.
  10. Käck U, Asarnoj A, Grönlund H, Borres MP, van Hage M, Lilja G, et al. Molecular allergy diagnostics refine characterization of children sensitized to dog dander. J Allergy Clin Immunol. 2018;142(4):1113-20.e9.
  11. Konradsen JR, Fujisawa T, van Hage M, Hedlin G, Hilger C, Kleine-Tebbe J, et al. Allergy to furry animals: New insights, diagnostic approaches, and challenges. J Allergy Clin Immunol. 2015;135(3):616-25.