Table of Contents
Component
f353 Gly m 4
f353 Gly m 4 Scientific Information
Type:
Component
Name; WHO/IUIS:
Gly m 4
Biological function:
Plant defense protein, pathogenesis-related protein
Allergen code:
f353
Route of Exposure:
Ingestion
Source Material:
Soy extract
Other Names :
Bet v 1-homologous allergen, Group 1 Fagales-related protein, PR-10 protein, SAM22
Summary
Gly m 4 acts as the major soy allergen for patients allergic to birch pollen, accounting for pollen-related soybean allergy in adults. It is a pan-allergen, belonging to group 10 of pathogenesis-related proteins (PR-10) with a molecular weight of 17 kDa. Gly m 4 concentration varies widely among the different soybean products, depending strongly on the total quantity of soybean in the product, the extent of food processing, and heating. The differences in Gly m 4 content of different soy-containing products may account for the differences in symptom severity (mild to severe). Patients sensitized to Gly m 4 may present oral allergy syndrome (facial edema, itchy eyes, angioedema, nasal congestion, nasopharyngitis, painful swallowing or other buccal symptoms), gastrointestinal, or respiratory symptoms, and even anaphylaxis upon consumption of soy-containing products. The sensitization to Gly m 4 in birch-pollen allergic patients poses as a risk factor for the development of oral allergy syndrome (OAS) of severe type as well as systemic reactions to soy. Gly m 4 is cross-reactive with allergens of other legumes like Bet v 1 (Betulaceae pollen) as well as with other PR-10 proteins such as Aln g 1 (alder pollen), Ara h 8 (peanut), Cor a 1.01 (hazel pollen), Cor a 1.04 (hazelnut), Mal d 1 (apple), Pru p 1 (peach). Importantly, Gly m 4 has been recognized as a diagnostic marker for for severe allergic reactions to soy in especially birch pollen allergic patients. 
Epidemiology
Worldwide distribution
Soybean is one of the “big eight” foods, considered to be accountable for 90% of all food allergies (1). It forms a major allergen in the United States (US) and Canada (1) and is responsible for the most important food allergies in the US, Europe, and Japan (2, 3). Soy allergy can trigger anaphylaxis in severe cases, which may be life-threatening (4).
Gly m 4, a pan-allergen and Bet v 1- homolog, accords for pollen-related soybean allergy in adults (5), majorly accounting for immediate-type soybean allergy in birch pollen allergic patients after consumption of food products containing soy (6, 7). Nevertheless, the relevance of Gly m 4 as a clinically significant allergen is restricted to areas with birch pollen allergy (8).
In a study conducted on 91 Japanese children with soybean sensitization, Gly m 4 sensitization (related to pollen-food allergy) was found to be 17.5% (7 out of 40) in the symptomatic patients while it was 21.6% (11 out of 91) in the non-symptomatic patients. The patients with a positive oral food challenge (OFC) were considered as symptomatic (40 out of 91) while those with negative OFC were considered non-symptomatic (51 out of 91) (9).
A study on 30 European subjects (25 adults and 5 children) found Gly m 4-sIgE in 70% (21 out of 30) of soybean-allergic patients (10). Furthermore, in another study in Austria (birch pollen environment) on 20 patients with soy allergy (10-69 years), 95% (19 out of 20) patients had specific immunoglobulin (sIgE) to Gly m 4. Also, another study in Utrecht, the Netherlands on 35 soy allergic adults found Gly m 4-sIgE in 74% of individuals (11). While a study conducted in the Netherlands on 305 patients sensitized to one or more PR-10 proteins found Gly m 4 sensitization to be 38% (115 out of 305) (12).
Environmental Characteristics
Source and tissue
Gly m 4 is an allergenic soy protein (13), belonging to group 10 of pathogenesis-related proteins (PR-10) (8). Gly m 4, formerly known as starvation-associated- message 22 (SAM22), is produced in the soybeans under conditions of starvation (8, 14).
Fast protein liquid chromatography using chromatofocusing could be used for the purification of Gly m 4 from defatted soybean flakes (7). Recombinant Gly m 4 (rGly m 4) can be generated in Escherichia coli (9) and could be purified using anion exchange as well as reversed-phase chromatography (13). 
Risk factors
The sensitization to Gly m 4 in birch-pollen allergic patients poses as a risk factor for the development of oral allergy syndrome (OAS) of severe type as well as systemic reactions to soy (9, 15, 16).
Clinical Relevance
Disease severity
Gly m 4 is found to be associated with pollen-food allergy syndrome (PFAS) (12, 17) and is mainly accountable for severe OAS as well as moderate to severe generalized reactions in allergic individuals (5, 7, 16, 18). The severe symptoms include oral allergy syndrome (facial edema, itchy eyes, angioedema, nasal congestion, nasopharyngitis, painful swallowing or other buccal symptoms), gastrointestinal, or respiratory symptoms, and even anaphylaxis (5, 7, 19). The differences in Gly m 4 content of different soy-containing products may account for the differences in symptom severity (mild to severe) (20).Gly m 4 sensitivity is frequently found in birch pollen allergic patients. However, the sensitization to this soy allergen possibly suggests severe allergic reactions, unlike other Bet v 1 homolog allergens (14, 18).
Severe allergic reactions may develop following ingestion of mildly processed soy, such as soy drinks, in patients primarily sensitized to birch pollen (21). This was observed in a study on four Swedish children (10–14-year-old) allergic to birch pollen. All the four children manifested severe allergic reactions (ranging from allergic rhino-conjunctivitis to OAS and gastrointestinal symptoms) after consumption of soymilk during peak pollen season for birch (22).
Further, it has been observed that higher Gly m 4-sIgE is associated with soy-milk allergy and not with other soy products. This may be due to different ways of processing the soy products. A study in Utrecht, The Netherlands found significantly high sIgE for Gly m 4 in the soy-allergic patients who manifested anaphylactic reactions to soy milk. The study was conducted on 35 soy-allergic adults with 15 patients presenting mild symptoms while 20 patients presenting severe symptoms to soy food challenge (11).
In contrast, 92% of soybean allergic patients suffering from mild symptoms were found to be sensitized to Gly m 4 in a European study on 30 soybean-allergic participants (25 adults and 5 children). This study could not establish a strong association of Gly m 4-sIgE to the development of severe symptoms as compared to Gly m 5 or Gly m 6. However, they suggest that Gly m 4 could induce severe reactions in patients with high levels of IgE to Bet v 1 (10).
Cross-reactive molecules
Gly m 4 is a common cross-reactive protein in Europe due to frequent sensitization to birch (8). Thus, pollen-related allergy to soybean in adolescents and adults may be attributed to the IgE cross-reactivity between Bet v 1 (Betulaceae pollen) and its homolog, Gly m 4 (23). Usually, birch pollen allergic patients may develop a secondary allergy to soy, as a result of the cross-reactivity between birch pollen and soy (20). Further, this clinical cross-reactivity may manifest as severe symptoms (21, 23, 24).
A study reported facial, oropharyngeal, and/or systemic symptoms in 20 patients in less than 30 minutes of consuming a soy product. The birch pollen allergy was co-existent in 80% (16 out of 20) patients while 85% (17 out of 20) patients demonstrated sIgE to Gly m 4. Moreover, allergic symptoms were most commonly observed during peak pollen season for trees. Also, recognition of Gly m 4 in patients with high IgE levels to Bet v 1 was associated with the development of severe symptoms (7).
Clinical cross-reactivity has also been reported among patients allergic to peanuts, soybean, and other legumes (24). It may be due to cross-reactivity between Gly m 4 and Bet v 1 homolog, Ara h 8 in peanuts (15). A co-existing pattern of clinical birch, peanut and soy allergy was observed in a study in Gothenburg, Sweden conducted on 47 severe peanut-allergic adults. Sensitization to soy-IgE was observed in 72.3% (34 out of 47) of these adults. It was observed that IgE reactivities to Gly m 4 were correlated with sIgE to Ara h 8 as well as to birch (24).
This pattern of co-sensitization was further confirmed in a study conducted on 305 patients sensitized to one or more PR-10 proteins. Gly m 4 sensitization was found in 38% of patients. The co-sensitization prevalence of Bet v 1, Cor a 1.01 (hazel pollen), Cor a 1.04 (hazelnut), Mal d 1 (apple), Aln g 1 (alder pollen), Pru p 1 (peach), Ara h 8 (peanut), Api g 1 (celery) and Act d 8 (kiwi) was found to be 100%, 100%, 100%, 100%, 100%, 97%, 94%, 50% and 41%, respectively (12).
Furthermore, a retrospective cohort study in 1057 food-allergic patients in Vienna found cross-reactivity among soybean, peanut, hazelnut, apple, and peach allergens which was termed as “soybean-nuts-fruits” cross-reactive cluster. It was found in 132 patients, majorly due to cross-reactivity between Ara h 8 (peanut) and Cor a 1.0101 and Cor a 1.0401 (hazelnut), Gly m 4 (soybean), Mal d 1 (apple), and Pru p 1 (peach) (25). 
Molecular Aspects
Biochemistry
Gly m 4 belongs to the PR-10 superfamily of proteins including Bet v 1-like proteins (2, 5). The PR-10 proteins are cytosolic proteins having 154-163 amino acids with a molecular weight of (26) 17 kDa. They are mostly involved with defense mechanisms in plants. They share a common tertiary structure with a characteristic hydrophobic core which is the binding site for different ligands. It comprises of seven, anti-parallel arranged β- strands and 2 short α-helices forming a V-shape, surrounded by a long α-helix (27).
Gly m 4 weighs about 17 kDa (17). Structurally, it differs from other PR-10 proteins in the long α-helix which appears bent unlike the straight α3 helix in Bet v 1 (14).
Further, Gly m 4 is soluble in water and does not possess resistance to heat, gastric acid digestion, and proteases (4). It is also prone to degradation by fermentation (16). These properties account for the loss of the protein content during the processing of soy such as roasting, acid precipitation, or pasteurization (4). Further, it is also observed that Gly m 4 concentration varies widely among the different soybean products that depend strongly on the total quantity of soybean in the product, the extent of food processing, and heating (22).
Isoforms, epitopes, antibodies
As of 27th January 2021, only one isoallergen of Gly m 4 has been officially designated by the World Health Organization (WHO) and International Union of Immunological Societies (IUIS) Allergen Nomenclature Subcommittee, namely Gly m 4.0101 (9, 28).
Cross-reactivity
Gly m 4 is homologous to Bet v 1 (5), sharing 53% sequence identity to Bet v 1 (7). Extensive IgE cross-reactivity has been reported between Bet v 1 and Gly m 4 (22, 29, 30). It was demonstrated by a study on 4 soy-allergic patients where IgE to Gly m 4 in all the 4 patients was inhibited by Bet v 1 (14). Further, sIgE to Bet v 1 was found to be significantly correlated (p<0.001) with Gly m 4-sIgE in a study on 35 soy-allergic and 11 soy-tolerant adults in the Netherlands (11). The high degree of sequence similarity and highly similar tertiary structure accounts for the cross-reactivity that occurs between the amino acid sequences of Gly m 4 and Bet v 1 (2, 8).
A study on 57 birch pollen allergic patients identified the major epitope of Bet v 1 to be Bet v 1142-156. This major epitope was considered to play a role in cross-reactivity and was recognized by Gly m 4 in 44.4% of the 11 T cell clones (31). Similarly, another study on 203 Chinese patients with a clinical history of allergic rhinitis/asthma found a high correlation between specific IgE for Bet v 1 and Gly m 4 (p<0.0001) (32).
Gly m 4 has been extensively described to cross-react with other PR-10 proteins in food (17). A study identified amino acid sequence homology between Gly m 4 and other PR-10 proteins. It was found to be 44% for Act d 8 (kiwi), 35.4% for Api g 1 (celery), 70.2% for Ara h 8, 56.5% for Cor a 1.04 (hazelnut), 50.6% for Mal d 1, 52.5% for Pru p 1, 47.5% for Aln g 1 and 67.1% for Bet v 1 respectively (19). Further, it was also reported that IgE-binding to Gly m 4 was inhibited by Pru av 1 in 2 out of 4 soy-allergic patients (14).
Immuno-cross-reactivity between peanut and soybean may be partly attributed to similarities between their allergens, Ara h 8 and Gly m 4, in amino acid sequence, IgE epitope mapping, and molecular modeling (2).  
Further, the sequence similarity of 74%-84 % has been found between Gly m 4 and its homologous PR-10 proteins of yellow lupine, a flowering plant belonging to the legume family Fabaceae (14).
Diagnostic Relevance
Disease Severity
Sensitization to Gly m 4 can serve as a marker for severe allergic reactions to soy in especially birch pollen allergic patients (16, 18).
Interestingly, in a study conducted on 21 pollen-related soybean allergic patients and 93 pollen sensitized controls in Japan, sensitization to Gly m 4 in soybean allergic adult patients were found to be of more relevance than Gly m 5 or Gly m 6 (considered to be more relevant in children). Further, Gly m 4-sIgE levels could differentiate between pollen-related soybean allergy and pollen sensitization (p<0.001) (33). 
However, Gly m 4 sensitization may not be useful for determining the clinical expression of soybean allergy in sensitized individuals. This was observed in a study conducted on 74 soybean-sensitized Japanese individuals. Out of 74 individuals, 33 individuals were found symptomatic (severe symptoms in 14 and mild symptoms in 19) while 41 were non-symptomatic based on OFC. IgE reactivity to Gly m 4 was observed to be almost similar in symptomatic patients (21%) and non-symptomatic individuals (20%) (3). 
Exposure
The main route of exposure is through ingestion (9). 
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Christian Fischer
Last reviewed: February 2021
References
  1. Messina M, Venter C. Recent Surveys on Food Allergy Prevalence. Nutrition Today. 2020;55(1).
  2. Cabanillas B, Jappe U, Novak N. Allergy to Peanut, Soybean, and Other Legumes: Recent Advances in Allergen Characterization, Stability to Processing and IgE Cross-Reactivity. Mol Nutr Food Res. 2018;62(1):1700446.
  3. Ito K, Sjolander S, Sato S, Moverare R, Tanaka A, Soderstrom L, et al. IgE to Gly m 5 and Gly m 6 is associated with severe allergic reactions to soybean in Japanese children. J Allergy Clin Immunol. 2011;128(3):673-5.
  4. Ueberham E, Spiegel H, Havenith H, Rautenberger P, Lidzba N, Schillberg S, et al. Simplified Tracking of a Soy Allergen in Processed Food Using a Monoclonal Antibody-Based Sandwich ELISA Targeting the Soybean 2S Albumin Gly m 8. Journal of Agricultural and Food Chemistry. 2019;67(31):8660-7.
  5. Ito K. Grain and legume allergy. Chem Immunol Allergy. 2015;101:145-51.
  6. L'Hocine L, Boye JI. Allergenicity of soybean: new developments in identification of allergenic proteins, cross-reactivities and hypoallergenization technologies. Crit Rev Food Sci Nutr. 2007;47(2):127-43.
  7. Kleine-Tebbe J, Vogel L, Crowell DN, Haustein UF, Vieths S. Severe oral allergy syndrome and anaphylactic reactions caused by a Bet v 1- related PR-10 protein in soybean, SAM22. J Allergy Clin Immunol. 2002;110(5):797-804.
  8. Lu M, Jin Y, Cerny R, Ballmer-Weber B, Goodman RE. Combining 2-DE immunoblots and mass spectrometry to identify putative soybean (Glycine max) allergens. Food Chem Toxicol. 2018;116(Pt B):207-15.
  9. Maruyama N, Sato S, Cabanos C, Tanaka A, Ito K, Ebisawa M. Gly m 5/Gly m 8 fusion component as a potential novel candidate molecule for diagnosing soya bean allergy in Japanese children. Clin Exp Allergy. 2018;48(12):1726-34.
  10. Holzhauser T, Wackermann O, Ballmer-Weber BK, Bindslev-Jensen C, Scibilia J, Perono-Garoffo L, et al. Soybean (Glycine max) allergy in Europe: Gly m 5 (beta-conglycinin) and Gly m 6 (glycinin) are potential diagnostic markers for severe allergic reactions to soy. J Allergy Clin Immunol. 2009;123(2):452-8.
  11. Klemans RJ, Knol EF, Michelsen-Huisman A, Pasmans SG, de Kruijf-Broekman W, Bruijnzeel-Koomen CA, et al. Components in soy allergy diagnostics: Gly m 2S albumin has the best diagnostic value in adults. Allergy. 2013;68(11):1396-402.
  12. Blankestijn MA, Knulst AC, Knol EF, Le T-M, Rockmann H, Otten HG, et al. Sensitization to PR-10 proteins is indicative of distinctive sensitization patterns in adults with a suspected food allergy. Clinical and Translational Allergy. 2017;7(1):42.
  13. Neudecker P, Lehmann K, Rösch P. Sequence-specific 1H, 13C and 15N resonance assignments of SAM22, an allergenic stress-induced protein from soy bean. J Biomol NMR. 2003;26(2):191-2.
  14. Berkner H, Neudecker P, Mittag D, Ballmer-Weber B, Schweimer K, Vieths S, et al. Cross-reactivity of pollen and food allergens: Soybean Gly m 4 is a member of the Bet v 1 superfamily and closely resembles yellow lupine proteins. Bioscience reports. 2009;29:183-92.
  15. Borres MP, Ebisawa M, Eigenmann PA. Use of allergen components begins a new era in pediatric allergology. Pediatr Allergy Immunol. 2011;22(5):454-61.
  16. Borres MP, Maruyama N, Sato S, Ebisawa M. Recent advances in component resolved diagnosis in food allergy. Allergology International. 2016;65(4):378-87.
  17. Matsuo A, Matsushita K, Fukuzumi A, Tokumasu N, Yano E, Zaima N, et al. Comparison of Various Soybean Allergen Levels in Genetically and Non-Genetically Modified Soybeans. Foods. 2020;9(4).
  18. Berneder M, Bublin M, Hoffmann-Sommergruber K, Hawranek T, Lang R. Allergen chip diagnosis for soy-allergic patients: Gly m 4 as a marker for severe food-allergic reactions to soy. Int Arch Allergy Immunol. 2013;161(3):229-33.
  19. Scala E, Abeni D, Cecchi L, Guerra EC, Locanto M, Pirrotta L, et al. Molecular Recognition Profiles and Clinical Patterns of PR-10 Sensitization in a Birch-Free Mediterranean Area. International Archives of Allergy and Immunology. 2017;173(3):138-46.
  20. De Swert LF, Gadisseur R, Sjölander S, Raes M, Leus J, Van Hoeyveld E. Secondary soy allergy in children with birch pollen allergy may cause both chronic and acute symptoms. Pediatr Allergy Immunol. 2012;23(2):117-23.
  21. Popescu F-D. Cross-reactivity between aeroallergens and food allergens. World J Methodol. 2015;5(2):31-50.
  22. Kosma P, Sjölander S, Landgren E, Borres MP, Hedlin G. Severe reactions after the intake of soy drink in birch pollen-allergic children sensitized to Gly m 4. Acta Paediatr. 2011;100(2):305-6.
  23. Ebisawa M, Brostedt P, Sjölander S, Sato S, Borres MP, Ito K. Gly m 2S albumin is a major allergen with a high diagnostic value in soybean-allergic children. J Allergy Clin Immunol. 2013;132(4):976-8.e1-5.
  24. Rentzos G, Lundberg V, Lundqvist C, Rodrigues R, van Odijk J, Lundell A-C, et al. Use of a basophil activation test as a complementary diagnostic tool in the diagnosis of severe peanut allergy in adults. Clinical and translational allergy. 2015;5:22-.
  25. Dezfouli SG, Mothes-Luksch N, Jensen AN, Untersmayr E, Kundi M, Jensen-Jarolim E. Linking cross-reactivity clusters of food and respiratory allergens in PAMD@ to asthma and duration of allergy. World Allergy Organization Journal. 2020;13(12):100483.
  26. Poole JA, Barriga K, Leung DY, Hoffman M, Eisenbarth GS, Rewers M, et al. Timing of initial exposure to cereal grains and the risk of wheat allergy. Pediatrics. 2006;117(6):2175-82.
  27. Aglas L, Soh WT, Kraiem A, Wenger M, Brandstetter H, Ferreira F. Ligand Binding of PR-10 Proteins with a Particular Focus on the Bet v 1 Allergen Family. Curr Allergy Asthma Rep. 2020;20(7):25.
  28. WHO/IUIS. Allergen Nomenclature: WHO/IUIS Allergen Nomenclature Sub-Committee; 2019 [cited 2021 Jan 27]. Available from: http://www.allergen.org/viewallergen.php?aid=345.
  29. Moraes AH, Asam C, Almeida FCL, Wallner M, Ferreira F, Valente AP. Structural basis for cross-reactivity and conformation fluctuation of the major beech pollen allergen Fag s 1. Scientific Reports. 2018;8(1):10512.
  30. Kern K, Havenith H, Delaroque N, Rautenberger P, Lehmann J, Fischer M, et al. The immunome of soy bean allergy: Comprehensive identification and characterization of epitopes. Clin Exp Allergy. 2019;49(2):239-51.
  31. Jahn-Schmid B, Radakovics A, Lüttkopf D, Scheurer S, Vieths S, Ebner C, et al. Bet v 1142-156 is the dominant T-cell epitope of the major birch pollen allergen and important for cross-reactivity with Bet v 1-related food allergens. J Allergy Clin Immunol. 2005;116(1):213-9.
  32. Hao G-D, Zheng Y-W, Wang Z-X, Kong X-A, Song Z-J, Lai X-X, et al. High correlation of specific IgE sensitization between birch pollen, soy and apple allergens indicates pollen-food allergy syndrome among birch pollen allergic patients in northern China. J Zhejiang Univ Sci B. 2016;17(5):399-404.
  33. Fukutomi Y, Sjölander S, Nakazawa T, Borres MP, Ishii T, Nakayama S, et al. Clinical relevance of IgE to recombinant Gly m 4 in the diagnosis of adult soybean allergy. J Allergy Clin Immunol. 2012;129(3):860-3.e3.