Whole Allergen
f203 Pistachio
f203 Pistachio Scientific Information
Type:
Whole Allergen
Display Name:
Pistachio
Allergen code:
f203
Family:
Anacardiaceae
Species:
Pistacia vera
Route of Exposure:
Ingestion
Latin Name:
Pistacia vera
Other Names:
Pistachio nut
WHO/ICD-11 code:
XM2B46
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Pistachio nut is the seed of the Pistacia trees belonging to the genus Pistacia. The P. vera is the highest, economically cultivated species owing to its edible nuts. It has been native to Central and West Asia and now diversified in Europe, Africa, America, and Australia. The green pistachio kernels are widely consumed as crude, roasted, salted, or as part of the large variety of food items because of their unique taste or aroma. The prevalence of its allergy is rapidly rising and has been across Asia, Europe, and the United States of America. Pistachio nuts have been found to produce allergic reactions like oral allergy syndrome or occasionally anaphylaxis on consumption. Five of its proteins, namely Pis v 1, Pis v 2, Pis v 3, Pis v 4, and Pis v 5, have been identified as having allergenic potential. Pistachio and cashew show a high level of co-sensitization or cross-reactivity since they are genetically closely related nuts. Individuals with cashew nut allergy have a higher risk of co-sensitization with pistachio. Besides cashew, it also shows cross-reactivity with mango seed and other tree nuts (walnut and pink peppercorn). 
Allergen
Nature
Pistachio nut is the seed of Pistacia trees. These trees are considered as one of the oldest flowering nut trees that grow from small to medium and are generally wind-pollinated. They are dioecious (separate male and female trees) (1) and have well-developed resin canals. The leaves are alternate, deciduous or persistent, compound to pinnate, membranaceous or leathery, with elliptical-to-ovate leaflets. The flowers are usually small and unisexual (2). The fruits are flattened, asymmetrical, semidry drupes that consist of a combined exocarp, fleshy mesocarp, and a hard endocarp. The thin off-white endocarp forms the shell, which encloses the seed or nut. It splits open upon ripening, thus revealing the edible embryo. The seed consists of thin skin with light-green flesh with a distinct flavor (1, 3).
Habitat
Pistachio trees are temperate deciduous trees that adjust themselves in dry, hot summers, and moderate winters and can well-tolerate alkaline and saline environmental conditions (1). They grow well in low and high altitudes with a maximum average slope of 25-30% (4).
Taxonomy
Pistachio belongs to the genus Pistacia, and to date, the exact number of species included in this genus is unclear. Pistacia is believed to consist of at least 12 species, namely P. aethiopica, P. atlantica (4 subspecies), P. chinensis (also known as P. phillippinensis, 2 subspecies), P. coccinea (also known as P. weinmannifolia), P. khinjuk, P. lentiscus, P. mexicana, P. saportae (a hybrid of P. lentiscus and P. terebinthus), P. simaruba (also known as Bursera simaruba), P. terebinthus (2 subspecies), P. texana, and P. vera. The P. vera is considered to have the highest economic value because of its edible nuts (1).
Taxonomic tree of Pistachio (5)  
Domain Eukaryota
Kingdom Plantae
Phylum Spermatophyta
Subphylum Angiospermae
Class Dicotyledonae 
Order Sapindales
Family Anacardiaceae
Genus Pistacia
Species Pistacia vera
Tissue
Pistachio nuts have been found to produce allergic reactions on consumption, and five of its proteins, namely Pis v 1, Pis v 2, Pis v 3, Pis v 4, and Pis v 5, have been identified as having allergenic potential (1).
Epidemiology
Worldwide distribution
The prevalence of pistachio nut allergy is rapidly rising due to an increase in its consumption globally. Several studies have reported its prevalence across Asia, Europe, and the United States of America (USA).
A retrospective study conducted in South Korea reported that pistachio nut allergy prevalence was found to be 0.8% in pediatric patients with a severe allergic reaction to food-induced anaphylaxis (6). In a study conducted in Israel, clinical allergy to pistachio was observed in 40% of tree nut allergic patients (age range: 3 to 24 years) (7). Furthermore, pistachio allergy was found among the top 10 food allergen in Turkish children (8). A large prevalence study conducted in the USA for peanut and tree nut allergies found the pistachio allergy in 9.8% of tree nut allergic individuals (9).
Pistachio nut allergy has become common in Europe. A study was conducted to examine the cross-sensitization profiles of tree nuts in birch endemic areas in Finland. The results reported that the prevalence of pistachio allergy in tree nut allergic subjects with and without birch sensitization was found to be 55% and 14%, respectively (10). In a prospective study conducted in Leicester (UK), among South Asian and European children, pistachio nut allergy was reported to be 25.7% in the South Asian and 6.9% in the European population (11). The Pronuts study, a prospective multicenter European study, found pistachio allergy in 24%, 47.6%, and 13.3% of children in London, Geneva, and Valencia, respectively (12).
Risk factors
Individuals with cashew nut allergy have reported having a higher risk of co-sensitization with pistachio since they are genetically closely related, showing high homology between allergens of both. Furthermore, in an IDEAL study, co-sensitization between pistachio and cashew nut was reported in 98% of cashew-sensitized individuals (1, 10, 13). It has been reported that allergic reactions to cashews and almonds were found to be more in individuals living in areas with high cultivation of pistachios (3).
Environmental Characteristics
Worldwide distribution
The green pistachio kernels are widely consumed as crude, roasted, salted, or as part of a large variety of food items, including chocolates, cakes, confectionaries, ice-creams, because of its unique taste or aroma. They are even used as folk medicines for some diseases (1, 3). Pistacia vera has been native to Central (Kazakhstan, Kyrgyzstan, Tajikistan, Turkmenistan, and Uzbekistan) and West (Afghanistan and Iran) Asia, and then diversified into the Mediterranean region of Europe, North Africa, and the Middle East. It is now cultivated in several countries, which include America (USA and Mexico), Eastern Africa (Madagascar and Mauritius), Northern Africa (Morocco, Tunisia), Western Africa (Cote d’Ivoire), Asia (China, Turkey, Jordan, Azerbaijan), Europe (Greece, Italy, Spain) and Australia (Oceania). As per reports in 2014, the primary global producers for Pistachio nuts were Iran (48.4%), followed by the USA (27.2%), Turkey (9.3%), and China (9.0%). Furthermore, the largest domestic consumption of these nuts was recorded in Turkey, followed by the European Union, China, and the USA (1).
Route of Exposure
Main
Oral ingestion of dry pistachio or food containing pistachios can trigger allergic reactions like type I hypersensitivity reactions or oral allergic syndrome with clinical manifestations, such as vomiting, stomach pain, urticaria, pruritus, erythema, eczema, nasal congestion, itchy throat, frequent coughing, wheezing, dyspnea, red/watery eyes, angioedema, swelling of lips, hypotension or even anaphylaxis (1, 3).
Clinical Relevance
Oral allergy symptoms and Anaphylaxis
In a study conducted on 60 multi-allergic pediatric patients, a double-blind, placebo-controlled food challenge was found to be positive in 100% of pistachio allergic patients. The challenge reported skin reactions, gastrointestinal reactions, and respiratory reactions in 95%, 38%, and 2%, respectively (14). In a prospective study conducted in Spain on 18 pediatric patients allergic to pistachios and cashew, urticaria and oral symptoms were reported in 38.9% of children, gastrointestinal manifestations in 33.3%, and skin reactions in 16.6% of children. Furthermore, 50% of patients developed moderate anaphylactic reactions, while none developed severe anaphylaxis (15).
Ando, Watanabe (16) reported a case of a 69-year-old Japanese woman who showed oral allergy syndrome with severe anaphylaxis after consuming pistachio nut. However, pistachio-specific Immunoglobulin E (IgE) antibody reaction was negative in this patient (16). While another case of pistachio-related anaphylaxis among the 15 proven food-allergic adolescents was observed in a study conducted in Turkey to evaluate the prevalence of IgE-mediated food allergy among adolescents (17).
Allergic rhinitis
Allergic rhinitis was reported in 20.7% of children with a history of pistachio allergy and in 9.5% of children with a positive oral food challenge (OFC)/anaphylaxis to pistachio in the last 12 months in a prospective cohort study involving 112 children with a history of pistachio and/or cashew nut allergy (18). 
Asthma
According to a study conducted by Ahn et al., 42.8% of pistachio-allergic patients reported asthma (19).
In a prospective study conducted on 112 children with a history of pistachio and/or cashew allergy, asthma was reported in 46.6% of children with a history of pistachio allergy. Furthermore, 38% of children showed a positive OFC/anaphylaxis to pistachio in the last 12 months (18).
Asthma was observed in 47.5% of children who were allergic to pistachio and cashew in a study conducted in France (20).
Atopic Dermatitis
In a study conducted by Ahn et al., 57.14% of pistachio allergic individuals reported a history of atopic eczema (19).
In a prospective study involving 112 children with a history of pistachio and/or cashew allergy, atopic dermatitis was reported in 22.5% of children with a history of pistachio allergy and 54.5% of children who had a positive OFC or anaphylaxis in the last 12 months due to pistachio (18).
In a study conducted in France on 51 cashew allergic children, pistachio allergy was reported in 40 patients. In these dual allergic children, atopic dermatitis was observed in 62.5% (20).
Other diseases
Seasonal rhino-conjunctivitis and food-dependent exercise-induced anaphylaxis have been reported due to the ingestion of pistachio in some cases (1).
In a study conducted in 51 cashew nut-allergic children in France, 40 children were co-allergic to pistachios and showed rhino-conjunctivitis in 37.5% of children (20).
Diagnostics Sensitization
Challenge tests
Oral food challenges (especially double-blind placebo-controlled) are the gold standard for diagnosing clinical reactivity to tree nuts. However, they are time-consuming and associated with the risk of provoking serious reactions (7, 18, 21). 
Molecular Aspects
Allergenic molecules
To date, five allergenic molecules have been identified and characterized and published officially by the World Health Organization and International Union of Immunological Studies (WHO/IUIS) Allergen Nomenclature Sub-Committee for pistachio nut (1, 22, 23).
Among the five pistachio nut allergenic proteins, Pis v 1 and Pis v 2, 3, 5 are the seed storage proteins that belong to the prolamin and cupin protein superfamily, whereas Pis v 4 is a plant defense protein belonging to the manganese superoxide dismutase protein family (1). The table below provides detailed information on each of the allergenic proteins.
Name of allergen Isoallergen Biochemical name Molecular weight (kDa)    Allergenicity
Pis v 1 Pis v 1.01 2S albumin 7
  • Major allergen.
  • 67.8% of pistachio allergic patients reacted immunologically, accounting for more than 50% IgE-reactivity.
  • 66% homologous to Ana o 3 of cashew
Pis v 2 Pis v 2.01 11S legumin like globulin 32
  • Major allergen
  • 50% of patients reactive in immunoblots
  • 46-48% sequence homology to Ana o 2 of cashew
Pis v 2.02
Pis v 3 Pis v 3.01
7S Vicilin
55
  • Minor allergen
  • 36.8% of patients reactive in immunoblots
  • 80% sequence identity with Ana o 1 of cashew
Pis v 4 Pis v 4.01 Manganese superoxide dismutase 25.7
  • Major/minor allergen
  • 59% of patients and 40% of patients’ sera were found reactive in immunoblots
Pis v 5 Pis v 5.01 11S legumin like globulin 36
  • Minor allergen
  •  35.7% of patients reactive in immunoblots
 kDa: kilodaltons, IgE: Immunoglobulin E
Biomarkers of severity
There are no allergen components for pistachio available with a quantitative method. However, the cashew Ana o 3 has proven useful when investigating pistachio allergy due to high homology with Pis v 1 compared to whole extract testing with pistachio (24).
Cross-reactivity
Pistachio, cashew, and mango belong to the same botanical family Anacardiaceae. Pistachio and cashew show a high level of co-sensitization or cross-reactivity since they are genetically closely related nuts. Two initial case reports of pistachio allergy have shown to possess specific IgE for cashew nut (25). The NUTCRACKER study, the IDEAL (Improvement of Diagnostic mEthods for ALlergy assessment) study and a retrospective study conducted in France reported a high prevalence (75-98%) of pistachio sensitization in cashew nut sensitized children (7, 20, 26). However, clinically relevant allergy was found to be 34% for pistachio nuts in 29 cashew sensitized children (13). The strong association between cashew and pistachio nut allergy was further confirmed by conducting a double-blind, placebo-controlled food challenge in 42 pistachio allergic patients. All of them were found positive against cashew in the food challenge (14). The amino acid sequence similarity between cashew and pistachio nut allergens: Ana o 1/Pis v 3 (7S vicilin), Ana o 2/Pis v 2 (11S legumin), and Ana o 3/Pis v 1 (2S albumin) was found to be 78%, 80% and 70% respectively (26). Further, a prospective study conducted in Spain demonstrated no cross-reactivity between mango and pistachio nut (15). However, earlier studies have shown that cross-reactivity may exist between pistachio and mango seed and not mango pulp. Hence, patients may still tolerate mango, as only pulp is to be consumed (25).
Usually, all tree nuts are avoided in peanut-allergic individuals. However, as per a study, pistachios were serologically cross-reactive with peanuts, but 51.6% of peanut-allergic individuals (n=258) were recommended to consume pistachios (27). Pistachios showed a probability (odds ratio) of 0.64 for a negative skin prick test in peanut-allergic individuals in a study (28), while in another study, weak cross-reactivity between allergens of both was demonstrated (29). Furthermore, in a study conducted by Dubiela et al., it was reported that Jug r 6, vicilin protein from walnut might serve as a marker for cross-reactivity between walnut and pistachio and corresponds to Pis v 3, a minor allergen of pistachio (30). Cross sensitization between pink peppercorn, cashew, and pistachio has been demonstrated in 76.2% individuals by skin prick tests (31), while cross-reactivity, partly mediated by 2S albumins, was confirmed in two cases by serum IgE analysis (32). A retrospective analysis study showed 52% co-sensitization between pistachios and coconut but was not significantly associated (33). Further, cross-reactivity between yellow mustard seeds and pistachio may be possible owing to the sharing of IgG and IgE epitopes with 11S globulins of pistachio (34).
In all, significant cross-sensitization and cross-reactivity of pistachios have been shown with cashews, pink peppercorn, and walnut; however, a weak association exists with mango, peanuts, or coconut.
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Magnus Borres
Last reviewed: December 2020
References
  1. Costa J, Silva I, Vicente AA, Oliveira M, Mafra I. Pistachio nut allergy: An updated overview. Crit Rev Food Sci Nutr. 2019;59(4):546-62.
  2. Al-Saghir M, Porter D. Taxonomic Revision of the Genus Pistacia L. (Anacardiaceae). American Journal of Plant Sciences. 2012;3:12-32.
  3. Esmaeili Nadimi A, Ahmadi Z, Falahati-Pour SK, Mohamadi M, Nazari A, Hassanshahi G, et al. Physicochemical properties and health benefits of pistachio nuts. Int J Vitam Nutr Res. 2020;90(5-6):564-74.
  4. Kozhoridze G, Orlovsky N, Orlovsky L, Blumberg DG, Golan-Goldhirsh A. Geographic distribution and migration pathways of Pistacia – present, past and future. Ecography. 2015;38(11):1141-54.
  5. Pistachia vera (pistachio) [Internet]. CAB International. 2019 [cited November 2, 2020].
  6. Jeong K, Lee SY, Ahn K, Kim J, Lee HR, Suh DI, et al. A multicenter study on anaphylaxis caused by peanut, tree nuts, and seeds in children and adolescents. Allergy. 2017;72(3):507-10.
  7. Elizur A, Appel MY, Nachshon L, Levy MB, Epstein-Rigbi N, Golobov K, et al. NUT Co Reactivity - ACquiring Knowledge for Elimination Recommendations (NUT CRACKER) study. Allergy. 2018;73(3):593-601.
  8. Kahveci M, Koken G, Şahiner Ü M, Soyer Ö, Şekerel BE. Immunoglobulin E-Mediated Food Allergies Differ in East Mediterranean Children Aged 0-2 Years. Int Arch Allergy Immunol. 2020;181(5):365-74.
  9. Sicherer SH, Muñoz-Furlong A, Godbold JH, Sampson HA. US prevalence of self-reported peanut, tree nut, and sesame allergy: 11-year follow-up. J Allergy Clin Immunol. 2010;125(6):1322-6.
  10. Uotila R, Kukkonen AK, Pelkonen AS, Mäkelä MJ. Cross-sensitization profiles of edible nuts in a birch-endemic area. Allergy. 2016;71(4):514-21.
  11. Luyt DK, Vaughan D, Oyewole E, Stiefel G. Ethnic differences in prevalence of cashew nut, pistachio nut and almond allergy. Pediatr Allergy Immunol. 2016;27(6):651-4.
  12. Brough HA, Caubet JC, Mazon A, Haddad D, Bergmann MM, Wassenberg J, et al. Defining challenge-proven coexistent nut and sesame seed allergy: A prospective multicenter European study. J Allergy Clin Immunol. 2020;145(4):1231-9.
  13. van der Valk JPM, Bouche RE, Gerth van Wijk R, de Groot H, Wichers HJ, Dubois AEJ, et al. Low percentage of clinically relevant pistachio nut and mango co-sensitisation in cashew nut sensitised children. Clin Transl Allergy. 2017;7:8.
  14. Andorf S, Borres MP, Block W, Tupa D, Bollyky JB, Sampath V, et al. Association of Clinical Reactivity with Sensitization to Allergen Components in Multifood-Allergic Children. J Allergy Clin Immunol Pract. 2017;5(5):1325-34.e4.
  15. Azcona OM, Romero L, Bartolome B, Balboa V, Vila L. Clinical cross-reactivity among mango, pistachio nut, and cashew nut in allergic children. J Allergy Clin Immunol Pract. 2020;8(9):3234-6.e2.
  16. Ando K, Watanabe D, Tamada Y, Matsumoto Y. Oral allergy syndrome with severe anaphylaxis induced by pistachio. Int J Dermatol. 2011;50(5):632-3.
  17. Kaya A, Erkoçoğlu M, Civelek E, Çakır B, Kocabaş CN. Prevalence of confirmed IgE-mediated food allergy among adolescents in Turkey. Pediatric Allergy and Immunology. 2013;24(5):456-62.
  18. Cetinkaya PG, Karaguzel D, Esenboğa S, Sahiner UM, Soyer O, Buyuktiryaki B, et al. Pistachio and cashew nut allergy in childhood: Predictive factors towards development of a decision tree. Asian Pac J Allergy Immunol. 2019.
  19. Ahn K, Bardina L, Grishina G, Beyer K, Sampson HA. Identification of two pistachio allergens, Pis v 1 and Pis v 2, belonging to the 2S albumin and 11S globulin family. Clin Exp Allergy. 2009;39(6):926-34.
  20. Saba L, Clerc-Urmès I, Delahaye C, Chevillot E, Jarlot-Chevaux S, Dumond P, et al. Predictive factors of allergy to pistachio in children allergic to cashew nut. Pediatr Allergy Immunol. 2020;31(5):506-14.
  21. Sicherer SH, Sampson HA. Food allergy: A review and update on epidemiology, pathogenesis, diagnosis, prevention, and management. J Allergy Clin Immunol. 2018;141(1):41-58.
  22. Allergen Nomenclature "Pistacia vera - All Allergens" [Internet]. WHO/IUIS Allergen Nomenclature Sub-Committee. 2019 [cited November 2, 2020].
  23. Willison LN, Sathe SK, Roux KH. Production and analysis of recombinant tree nut allergens. Methods. 2014;66(1):34-43.
  24. Savvatianos S, Konstantinopoulos AP, Borgå Å, Stavroulakis G, Lidholm J, Borres MP, et al. Sensitization to cashew nut 2S albumin, Ana o 3, is highly predictive of cashew and pistachio allergy in Greek children. J Allergy Clin Immunol. 2015;136(1):192-4.
  25. Fernandez C, Fiandor A, Martinex-Garate A, Quesada JM. Allergy to pistachio: crossre activity between pistachio nut and other Anacardiaceae. Clinical & Experimental Allergy. 1995;25(12):1254-9.
  26. Bastiaan-Net S, Reitsma M, Cordewener JHG, van der Valk JPM, America T, Dubois AEJ, et al. IgE Cross-Reactivity of Cashew Nut Allergens. Int Arch Allergy Immunol. 2019;178(1):19-32.
  27. Mustafa SS, Vadamalai K, Bingemann T, Mortezavi M, Aranez V, Ramsey A. Real-world tree nut consumption in peanut-allergic individuals. Ann Allergy Asthma Immunol. 2020;124(3):277-82.
  28. Randhawa I, Morphew T, Marsteller NL. Correlation of negative skin-prick test results for tree nuts and successful tree nut challenges among children with peanut allergy. Allergy and asthma proceedings. 2018;39(6):456-60.
  29. Pitre M, L'Hocine L, Achouri A, Blaquière M, Des Roches A. Immunoglobulin E-Binding Pattern of Canadian Peanut Allergic Children and Cross-Reactivity with Almond, Hazelnut and Pistachio. Biomolecules. 2020;10(8).
  30. Dubiela P, Kabasser S, Smargiasso N, Geiselhart S, Bublin M, Hafner C, et al. Jug r 6 is the allergenic vicilin present in walnut responsible for IgE cross-reactivities to other tree nuts and seeds. Sci Rep. 2018;8(1):11366.
  31. Too JJY, Shek LP, Rajakulendran M. Cross-reactivity of pink peppercorn in cashew and pistachio allergic individuals. Asia Pac Allergy. 2019;9(3):e25.
  32. Fong AT, Du Toit G, Versteeg SA, van Ree R. Pink peppercorn: A cross-reactive risk for cashew- and pistachio-allergic patients. J Allergy Clin Immunol Pract. 2019;7(2):724-5.e1.
  33. Polk BI, Dinakarpandian D, Nanda M, Barnes C, Dinakar C. Association of tree nut and coconut sensitizations. Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology. 2016;117(4):412-6.
  34. Sirvent S, Akotenou M, Cuesta-Herranz J, Vereda A, Rodríguez R, Villalba M, et al. The 11S globulin Sin a 2 from yellow mustard seeds shows IgE cross-reactivity with homologous counterparts from tree nuts and peanut. Clin Transl Allergy. 2012;2(1):23.