Whole Allergen
f4 Wheat
f4 Wheat Scientific Information
Type:
Whole Allergen
Display Name:
Wheat
Allergen code:
f4
Family:
Poaceae (Gramineae)
Species:
Triticum aestivum
Route of Exposure:
Ingestion
Source Material:
Untreated planting seeds
Latin Name:
Triticum aestivum
Other Names:
Common wheat, Bread wheat, Dinkel wheat
WHO/ICD-11 code:
XM3XH8
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Wheat (Triticum aestivum) is the third most important crop across the globe and grows in various climatic conditions. Wheat grain is high in nutritional content and is processed into several food items. Allergic reactions to wheat are mainly attributed to its protein content. The wheat proteins are classified as glutens (gliadins and glutenins) and non-glutens (α-amylase inhibitors, lipid transfer proteins and avenin-like proteins). The prevalence of wheat allergy (WA) differs across geographies and among different age groups. It is considered as the third most common allergy, after cow’s milk and egg in countries such as, Japan, Korea, Finland, Germany, and the United States. Wheat can induce both immunoglobulin (Ig) E-mediated as well as non-IgE-mediated food allergies. The IgE-mediated allergies include food allergy manifested as angioedema, nausea, abdominal pain, anaphylaxis including wheat dependent exercise-induced anaphylaxis (WDEIA), respiratory allergy (Baker’s asthma) as well as skin allergy (contact urticaria). The non-IgE-mediated ones include enterocolitis syndrome, eosinophilic esophagitis and non-celiac gluten sensitivity. Wheat allergy can be induced by oral ingestion, or by inhalation of wheat flour as well as through skin contact. There are several allergens (~28) identified from wheat, of which major allergens are found from the gluten family, in addition to some of the non-gluten proteins. Sensitization to Tri a 17, 19, 20, 26, and 37 in IgE-mediated wheat-allergic patients may indicate the presence of severe allergic reactions, while especially Tri a 19 have been reported as markers for WDEIA. Further, Tri a 27 and Tri a 28 have been found to be the most important wheat allergens in patients with Baker’s asthma. It is recommended that individuals allergic to wheat should strictly avoid the consumption of wheat and its products. Further, cross-reactivity has been shown between cereals of the same family i.e. between wheat, barley and rye. Being sensitized to wheat is common, mainly due to cross-reactivity to grass. Up to 60% of grass-allergic subjects are sensitized to wheat but can tolerate it. A positive wheat test always needs to be further investigated in order to know if the test results are clinically relevant or due to pollen cross-reactivity. 
Allergen
Nature
Triticum aestivum (T. aestivum), also known as wheat is a grass plant that belongs to the family Poaceae (1). It is considered as one of the first food crops that is domesticated and cultivated (2). T. aestivum is a high-yielding variety, that is self-pollinating, and grows worldwide in various climatic conditions (3, 4). It is a hexaploidy type of species that is also referred to as common or bread or soft wheat (5).
The seed of wheat is a type of dry indehiscent fruit (does not split open once mature), that is smooth and rounded at the dorsal surface, whereas with a deep ridge on the ventral surface (5, 6). The T. aestivum variety has a larger grain size in comparison to other varieties of wheat (5). The grain mainly constitutes of an outer layer and an inner layer, with the outer layer comprising of pericarp (4 to 5% of grain weight) and seed coat (1%), whereas the inner layer has an aleurone tissue (6 to 9%), starchy endosperm (80 to 85%) and germ (3%) (7). Interestingly, there are characteristic changes observed in the color of the wheat grain as it matures. A fresh fertilized grain is creamish-white in color, that turns into green color when the grain is of maximum size and finally has a golden-yellow color, when it is completely matured (6).
Wheat is large in nutritional content and is considered as a highly palatable grain. It is processed into many foods, such as breads, pasta, bulgur (dried cracked wheat), pizza, couscous, and beer (3). About 95% of all the wheat cultivated globally comprises of common bread wheat, whereas the rest of the 5% is durum wheat (T. durum), used for pasta and couscous production (5, 7). 
Habitat
Wheat crop is widely adapted to grow in varied temperature zones as well as environment, across the globe. It is an adjustable crop that can grow well, both in warm and humid climate as well as cold and dry regions (8). The temperature condition can vary from 3℃ to 32℃ with an ideal temperature being 25℃. Furthermore, the wheat plant can grow on varied altitudes, ranging from sea level to ~3000 m above sea level (2).
Taxonomy
Triticum aestivum is an allohexaploid variety of wheat that comprises of six sets of chromosomes, with two sets from each of T. monococcum (einkorn wheat), T. dicoccum (emmer wheat) and T. spelta (spelt wheat). Further, T. durum is tetraploid species of wheat, commonly used for preparation of pasta and couscous dish (5).
The taxonomic tree of T. aestivum obtained as of February 1, 2021 (9) is as follows:      
Taxonomic tree of Wheat (9)  
Domain Eukaryota
Kingdom Plantae
Phylum Spermatophyta
Class Monocotyledonae
Order Cyperales
Family Poaceae
Genus Triticum
Species Triticum aestivum
Tissue
Allergic reactions to wheat are mainly attributed to its protein content (7-22% of total weight)(1, 7). The classification of wheat proteins has been described in the below figure (10, 11):
Epidemiology
Worldwide distribution
Wheat allergy (WA) occurs in many ways (food allergy in children, WDEIA, Baker’s asthma or skin allergy) and its prevalence differs across the geographies and among different age groups. It is considered as the third most common allergy, after cow’s milk and egg in countries such as, Japan, Korea, Finland, Germany, and the United States (US) (in preschool children) (15, 18, 19).
The prevalence of WA among 1612 children with food allergies was found to be 1%  in a study conducted in US (20). Further, the prevalence of IgE-mediated WA in young children (age: <14 years) was analyzed in 4 studies involved in a systematic review conducted on 36 studies. It was found to be between 0.2% and 0.5%, based on an oral food challenge (OFC) conducted among young children in Denmark, Germany and the United Kingdom (UK) (21). Another systematic review involving 25 WA studies in Europe reported an estimated pooled prevalence of 0.3% across all age groups, based on OFC or history of WA (22). Additionally, in a study conducted in Finland among 108 children (age: 0.6-17.3 years) with suspected WA, who underwent OFC, 52.8% of children reacted positively (30 with immediate symptoms and 27 with delayed symptoms) (23). Also, in a birth cohort study involving 827 children in UK, WA was confirmed in 0.48% of children, based on OFC (16).
A systematic review of 66 studies reported the prevalence of WA in Asia-Pacific region (Iran, Japan, Korea, Malaysia, Singapore, Pakistan, Australia). According to the results, the prevalence was reported between 0.16% and 0.37% via studies conducted in general population and between 10.4% and 26.1% via studies conducted among allergic population (19). Further, a retrospective study conducted in Korea among 1353 children and adolescents (0-18 years) with immediate reactions to food reported 1661 cases of food-allergic reactions in a span of one year. Wheat was reported to be the cause of immediate food-allergic reactions in 7.9% cases (18). Additionally, a prevalence study conducted in India among 4680 allergic patients (all age groups) with sensitization to different foods, WA was reported in 21.8% (24).
Severe reactions in wheat-allergic patients could also lead to anaphylaxis (wheat induced anaphylaxis [WIA]). A study analyzed the risk of near-fatal anaphylaxis due to wheat in 93 US children who underwent OFC after consuming low doses of wheat. It was found that wheat could be an independent risk factor for anaphylaxis, which was reported to be in 42% of patients (25). Interestingly, WDEIA is one of the most severe forms of WA in adults and various studies have shown w-5-gliadin sensitization in most of the WDEIA patients (12). A study conducted on 167 cases of food-dependent exercise-induced anaphylaxis in Japanese patients found that the causative agent in >50% of the cases was wheat (26). Further, a recent prospective study conducted among 33 Thai-allergic patients found WDEIA to be more prevalent than WA (WDEIA: 69.7% vs. WA: 30.3%) (27).
It has also been shown that skin sensitization due to exposure to wheat proteins could lead to WA. This was observed in 2111 Japanese adults (age:1-93 years) in a survey (April 2012 to October 2014), who were sensitized to wheat due to the use of special facial soap containing hydrolyzed wheat proteins. These adults, who were previously non-allergic to wheat, developed allergic reactions due to wheat consumption after such cutaneous sensitization (17).
Wheat allergy is also manifested as IgE-mediated respiratory allergy, clinically presented as occupational asthma (Baker’s asthma) and rhinitis (15). The prevalence of occupational asthma and/or rhinitis due to wheat hypersensitivity was found to be 25.1%  in Spain during a study conducted among workers from cereal industries (28).
Risk factors
Genetic predisposition is one of the risk factors for the development of food allergies, including WA in patients. Further, sensitization to egg, cow’s milk, and grasses are also reported to be associated with the risk for WA (15).
Alcohol and intake of non-steroidal anti-inflammatory drugs (NSAIDs) may be considered as cofactors for  WDEIA reactions, that may precipitate it, even in the absence of exercise (3).
Pediatric issues
The IgE-mediated WA is common in children (29). The prevalence of this allergy is reported to be higher in children as compared to adults in general. Furthermore, it is also documented that the majority of children outgrow their allergies by the time they reach 12-14 years (3, 30). With respect to the clinical manifestations of IgE-mediated food allergies, the gastrointestinal (GI) symptoms have been frequently observed in young children, which reduces with age, as compared to the skin symptoms. On the contrary, severe systemic reactions, like anaphylaxis is more noted in adults or teens than in children (31). 
Environmental Characteristics
Worldwide distribution
Wheat is considered as a food staple in North Africa, Europe and the Western part of Asia (Curtis 2002). It is found as the third most important crop across the globe, after maize and rice. Furthermore, wheat is extensively grown in countries, such as China, India, Australia, Argentina, Turkey, Canada and US, constituting of 79% of the total global production of wheat from these countries. China has been reported to be the world’s largest producer of wheat, which is followed by countries, such as India, Kazakhstan and Russia. (32).
Route of Exposure
Main
Oral ingestion of wheat is the major route of exposure leading to IgE-mediated food-allergic reactions within 2 hours of its intake. In addition to this, WDEIA has also been reported to be precipitated as a result of physical exercise, following oral ingestion of wheat-based products (3, 15, 33).
Furthermore, inhalation or handling of wheat flour could lead to Baker’s asthma or rhinitis in sensitized individuals (15, 33).
Another important route of exposure to WA includes skin contact with hydrolyzed wheat proteins, which could manifest as contact urticaria (15, 33).
Clinical Relevance
Wheat can induce both IgE-mediated as well as non-IgE-mediated food allergies. The IgE-mediated allergies include food allergy, such as skin allergy (angioedema, contact urticaria), nausea, abdominal pain, bronchial obstruction, anaphylaxis, as well as WDEIA. The non-IgE-mediated ones include enterocolitis syndrome, eosinophilic esophagitis and NCGS (1, 3, 7, 12). 
Food Allergy and Anaphylaxis
Wheat allergy occurs predominantly in children with a familial predisposition of atopy. It is usually IgE mediated and the clinical manifestation occurs within 1-2 hours of its intake (29). 
In a US-based study, 39 out of 93 children undergoing oral wheat challenge were found positive. The allergic reactions observed during oral wheat challenge were classified as cutaneous (urticaria, angioedema, erythema, or AD), respiratory (wheezing, dyspnea, tachypnea, cough, sneezing, rhinitis, or throat tightness), GI (diarrhea, vomiting, or abdominal pain), cardiovascular (hypotension), neurological (syncope) and multi-organ system. Further, the OFC with wheat resulted in multi-organ systemic reactions in 28 children (25).
In a prospective analysis performed on 50 children with WA, 88% reported GI symptoms (62% with vomiting, 44% with diarrhea and 8% with abdominal pain). The other symptoms observed were cutaneous reactions (urticaria, erythema) in the range of 40-50%, bronchospasm in around 10% and anaphylaxis in the range of 10-15%. It was found that 32% of children developed WA, when they were in infant stage. Interestingly, it was observed that WA in these children resolved with age and by the age of 18 years, 76% children had outgrown their allergy. The 12 children with persistent WA showed more prominent anaphylaxis (about 50%) and bronchospasm (40-45%) as compared to skin or GI reactions (34).
A retrospective study conducted on 156 patients with a history of WA found 95 individuals to be having a history of food allergy due to wheat ingestion. The WA was confirmed in 21 individuals by food challenge test. Furthermore, the most commonly observed symptom in these 21 patients was urticaria (95.2%), along with other symptoms like pruritis, angioedema, erythema, diarrhea, vomiting, rhinitis and conjunctivitis (35).
In a study conducted in Finland among 108 wheat-allergic children (age: 0.6-17.3 years) subjected to OFC with wheat, a positive reaction was observed in 57 children (immediate reactions in 30 and delayed in 27), while 51 children responded negative to OFC. The most frequent symptoms observed were cutaneous (urticaria or erythema; 44%), followed by GI (26%), and respiratory symptoms (16%) (23).
Furthermore, in a cross-sectional survey conducted among 100 children with WA (IgE-mediated), 49% of children presented with only skin reactions, while 51% of children presented with anaphylaxis due to wheat (36).
Wheat-dependent exercise-induced anaphylaxis (WDEIA)
The WDEIA is an occasional, but potential severe allergy caused due to wheat consumption combined with accompanying factors, such as physical exercise, aspirin, alcohol etc. It usually affects adolescents and young adults. This severe allergy can be evoked by physical exercise within 1 to 4 hours after wheat consumption (33, 37). The occurrence of this severe reaction is rare, however, it has reportedly been more found in countries, such as Europe and Japan (37). It is generally manifested as pruritis, urticaria, angioedema as well as severe allergic reactions, such as intense sweating, abdominal colic, bronchial obstruction, syncope, and even systemic reactions, like anaphylaxis (3, 15, 33, 37).
In a retrospective study conducted on 156 patients with a history of WA, 48 individuals were found to be having a history of allergy, due to wheat ingestion accompanied by exercise. However, on confirmation with exercise challenge on tread mill in addition to OFC, 10 individuals were found to have WDEIA. Furthermore, among the 10 confirmed WDEIA patients, all the patients were reported to have urticaria, with angioedema found only in one of them (35).
Further, it was reported that the wheat allergens found responsible for WDEIA were w-5-gliadin and HMW-glutenins. Besides these, other allergens which may also lead to WDEIA were α/β/g-gliadins along with LMW-glutenins (37).
A multicentric, retrospective study conducted on 132 adults with w-5-gliadin allergy (also known as WDEIA, severe anaphylaxis after wheat ingestion) in UK found wheat to be the causative agent in 82% of patients. Additionally, WDEIA was induced in 80% of patients due to exercise, while 25%, 9% and 5% were found due to alcohol, NSAIDs and heat, respectively (38).
Respiratory and skin allergy
Bakers asthma or rhinitis have been considered as the most common occupational diseases, occurring due to inhalation of wheat allergens among bakers or individuals working in mills/chocolate factories. Further, contact urticaria due to skin contact with wheat flour is the most common skin reaction, mostly noted in bakers as well as individuals working in mills/flour factories (7).
In a retrospective study conducted among 156 patients with a history of WA, 13 individuals reported to have the allergy due to wheat flour inhalation. However, on confirmation with nasal food challenge, 11 individuals (mostly bakers) were found to present with inhalational allergy to wheat. Furthermore it was reported that all the 11 patients presented with rhinitis, while conjunctivitis and asthma was reported in 54.5% and 18.2% of patients, respectively (35).
A study was conducted on 2111 Japanese patients with WA due to cutaneous sensitization via hydrolyzed wheat-proteins in a facial soap. It was found that most of the individuals presented with skin symptoms (urticaria, skin redness and itching; 71%) and eyelid swelling (40%) while using the soap. However, this sensitization resulted in food allergy which was observed as allergic reactions after wheat consumption. The allergic symptoms recorded after wheat ingestion from 899 patients were eyelid swelling (77%), urticaria (60%), dyspnea (43%), anaphylactic shock (25%), rhinitis (13%) and vomiting (11%)  (17).
Non-IgE-mediated immune disorders
The non-IgE-mediated immune disorders (eosinophilic GI disease and esophagitis) occurring due to wheat sensitivity has been observed in a prospective study conducted on 50 wheat-allergic children. According to the results, the eosinophilic GI disease and eosinophilic esophagitis was reported to be 12% (6 of 50) and 10% (5 of 50) of patients, respectively (34).
Atopic disorders (Allergic rhinitis, asthma and atopic dermatitis)
In a prospective analysis performed on 50 children with WA, atopic disorders, such as AR, AD and asthma was reported in 64%, 78% and 48% of patients, respectively (34).
In another study conducted in Finland among 108 suspected wheat-allergic children, 57 (30 with immediate symptoms and 27 with delayed symptoms) were found positive on oral wheat challenge. Atopic dermatitis, AR and asthma was observed in 84.2%, 35.1%  and 19.2 of OFC-positive children, respectively (23).
Further, in a cross-sectional survey conducted among 100 children with WA, 47% of children presented with AD, 44% with AR and 14% with asthma (36).
Other diseases
Non-celiac gluten sensitivity (NCGS)
The NCGS or gluten sensitivity mainly occurs due to the consumption of gluten protein. This sensitivity is presented with GI reactions as well as extra-intestinal symptoms after consumption of gluten-containing foods, in patients who are not the candidates of celiac disease or WA disease (39). It is generally difficult to differentiate NCGS from celiac diseases or WA, owing to similar clinical manifestations, similar triggers as well as similar avoidance strategy (7).
The pathomechanism predicted for NCGS is attributed to the intolerance of wheat carbohydrates rather than wheat proteins. The wheat fructans or α-trypsin inhibitors may be considered as culprits for the same (12).
Prevention and Therapy
Prevention strategies
Avoidance
Individuals allergic to wheat should strictly avoid consumption of wheat and its products. Besides, occupational/domestic exposure to wheat allergens, in the form of wheat flour or dust should also be avoided. Furthermore, patients with WDEIA should be careful while performing strenuous physical activity, either alone, or during hot/humid condition or during pollination period (3, 10).
Molecular Aspects
Allergenic molecules
The World Health Organization (WHO) and International Union of Immunological Societies’ (IUIS) Allergen Nomenclature Subcommittee have identified and officially published 28 allergenic components for wheat. Among these, 18 are exposed through ingestion, whereas 10 are exposed through inhalation (13). The table below provides detailed information on each of the allergenic protein identified by WHO/IUIS as of February 1, 2021:
Allergen
Biochemical Name
Molecular weight (kDa)
Allergenicity
Tri a 12
Prolin
14
  • Route of exposure (ER): ingestion and inhalation (15).
  • Significant in baker’s allergy, whereas non-significant in food allergy (10).
  • Low heat stability (10).
Tri a 14
Non-specific lipid transfer protein 1
9
  • Minor allergen (1).
  • ER: ingestion and inhalation (15).
  • Important in food allergy, Baker’s asthma, and WDEIA (10, 15).
  • High heat stability (10).
Tri a 15
Monomeric alpha-amylase inhibitor 
13.2 (40)
  • ER: Inhalation (15).
  • Recognized by about 60% of infants/children with IgE-mediated food allergy (10).
  • High heat stability (10).
Tri a 17
β-amylase
56
  • ER: Ingestion (13).
  • Correlated with severe reactions to wheat ingestion, like WIA (41).
Tri a 18
Agglutinin isolectin 1
21.2 (40)
  • ER: Ingestion (15).
  • Can be recognized by Baker’s asthmatic patients (10).
Tri a 19
w-5-gliadin, seed storage protein
65
  • Major allergen (1).
  • ER: Ingestion (15).
  • Recognized by 50-70% of patients with WA (10).
  • High heat stability (10).
Tri a 20
g-gliadin
35-38
  • ER: Ingestion (15).
  • Recognized by 50-70% of patients with WA (10).
Tri a 21
α/β-gliadin
32.6 (40)
  • Major allergen (1).
  • ER: Ingestion (15).
  • Recognized by >50% of patients with WA (10).
  • Low heat stability (10).
Tri a 25
Thioredoxin
13.3 (40)
  • ER: Ingestion (15).
Tri a 26
High MW glutenin
88
  • Major allergen (1).
  • ER: Ingestion (15).
  • Recognized by 50-70% of patients with WA  (10).
  • Mainly detected in patients with severe reactions (10).
Tri a 27
Thiol reductase homologue
27
  • ER: Inhalation (15).
  • Recognized by patients with Baker’s disease, and may not be significant in food allergy (10).
Tri a 28
Dimeric alpha-amylase inhibitor
13
  • ER: Ingestion and Inhalation  (15).
  • Recognized by 50-70% of patients with WA (10).
  • Significant in Baker’s allergy to wheat (42).
  • High heat stability (10).
Tri a 29
Tetrameric
alpha-amylase inhibitor 
13
  • ER: Inhalation (15).
  • Recognized by 37% of patients with WA (10).
  • Significant in Baker’s allergy to wheat (42).
  • High heat stability (10).
Tri a 30
Tetrameric
alpha-amylase inhibitor 
16
  • ER: Ingestion and Inhalation (15).
  • Significant in Baker’s allergy to wheat (42).
  • High heat stability (10).
Tri a 31
Triosephosphate-isomerase
26.8 (40)
  • ER: Inhalation (15).
  • Rarely seen in Baker’s asthma (10).
Tri a 32
1-cys-peroxiredoxin
23.9 (40)
  • ER: Inhalation (15).
  • Significant in Baker’s allergy to wheat (42).
Tri a 33
Serpin (Trypsin inhibitor)
43.3 (40)
  • ER: Ingestion and Inhalation (15).
  • Recognized by patients who had Baker’s asthma (10).
Tri a 34
Glyceraldehyde-3-phosphate-dehydrogenase
36.5 (40)
  • ER: Inhalation (15).
  • Rarely seen in Baker’s asthma (10).
Tri a 35
Dehydrin
11.5
  • ER: Inhalation (15).
  • Significant in Baker’s allergy to wheat (10).
Tri a 36
Low MW glutenin - GluB3-23
40
  • Major allergen (1).
  • ER: Ingestion (15).
  • Recognized by 60-80% of patients with WA (10).
Tri a 37
α-purothionin
12
  • ER: Ingestion (15).
  • Recognized by 16% of patients with WA (10).
  • Not sensitized in patients with Baker’s asthma (10).
  • High heat stability (10).
Tri a 39
Serine protease
inhibitor-like protein
9.9 (40)
  • ER: Inhalation (13).
  • Significant in Baker’s allergy to wheat (42).
Tri a 40
Chloroform/methanol-soluble 17 protein 
15.96
  • ER: Inhalation (13).
Tri a 41
Mitochondrial ubiquitin ligase activator of NFKB 1
6.9(40)
  • ER: Inhalation (13).
Tri a 42
Hypothetical protein from cDNA
8,2(40)
  • ER: Inhalation (13).
Tri a 43
Hypothetical protein from cDNA
11.6(40)
  • ER: Inhalation (13).
Tri a 44
Endosperm transfer cell specific PR60 precursor
11.9(40)
  • ER: Inhalation (13).
Tri a 45
Elongation factor 1 (EIF1)
9.7(40)
  • ER: Inhalation (13).
cDNA: Complementary deoxy ribonucleic acid; ER: Route of Exposure; kDa: Kilodalton; MW: Molecular weight; NFKB: Nuclear factor kappa B subunit; NR: Not reported; WA: Wheat allergy; WIA: Wheat induced anaphylaxis
Biomarkers of severity
Sensitization to Tri a 17, 19, 20, 26, and 37 in IgE-mediated wheat-allergic patients may indicate a risk of severe allergic reactions, if exposed to wheat proteins (10, 41). Furthermore, Tri a 19, 21, 26 and 36 have been reported as markers for WDEIA (1).
Sensitization to Tri a 19 (ω-5 gliadin) in wheat-allergic infants/children not only indicates severe reactions in OFC, but may also indicate the persistence of WA in adulthood (10, 43, 44). It has also been found to be significantly correlated with WIA and WDEIA. Studies have found that sensitization to Tri a 19 in patients with a history of clinical reactivity to wheat could be an indication to avoid OFC in individuals, as they could be at risk of developing anaphylaxis (43). Performing a wheat challenge in patient sensitized to Tri a 19 implies a high risk of reacting to wheat (45).
Tri a 27 and Tri a 28 have been found to be the most important wheat allergens in patients with Baker’s asthma (42).
Cross-reactivity
Individuals allergic to wheat are often found to have cross-reactive allergy to other cereals as well, including barley and rye, and rarely to oats (10). This cross-reactivity may be taxonomically related as wheat, barley, rye, and oats belongs to the same family (46). The possible cross-reactivity among wheat, barley and rye may be attributed to the presence of homologous prolamin proteins, such as g-70 and g-35 secalins in rye, g-3 hordein in barley and w-5 gliadin in wheat (31, 47). This cross-reactivity was shown in a study conducted on 23 patients with WDEIA, who had IgE sensitization as well as positive SPT to w-5 gliadin (wheat protein). The g-70 secalins (rye) was found IgE-reactive in 91%, g-35 secalin (rye) in 83% and g-3 hordein (barley) in 91% of sera of adult patients. Further, the cross-reactivity was also shown by positive SPT in 15 w-5 gliadin-positive WDEIA patients. The positive reactions observed with g-35 secalins, g-70 secalins, and g-3 hordein was 20%, 67%  and 47% respectively (48). Additionally, proteins such as α-purothionins from these 3 cereals have found to exhibit >80% of sequence identity (31).
Clinical cross-reactivity among wheat and barley is also shown in a study conducted on 20 barley-allergic patients in Korea. Clinical WA was found in 75% of barley-allergic patients along with significant correlation (p<0.001) between IgE levels of wheat and barley (49). Besides, in a study conducted on 10 wheat-allergic individuals, cross-reactivity of wheat with barley, oats and Job’s tears was evaluated. The results reported a cross-reactivity rate of 60%, 33.3% and 20% with barley, oats and Job’s tears, respectively based on the clinical reactivity or SPT (50). Owing to this cross-reactivity, barley and rye along with wheat are proposed to be avoided in the 6-food elimination diet (avoiding all gluten containing grains) in patients with gluten sensitivity (47).
In a study conducted among patients with food allergy (including wheat), high levels of IgE antibodies, particularly to rice was displayed in these patients (51). In addition to this, maize LTP was found to possess 59-79% of sequence identity with wheat LTP (52). However, both rice and corn have been often looked upon as an alternative for the wheat-allergic patients. A study conducted on 18 wheat-allergic children showed negative outcomes when challenged with rice and corn (46).
Patients with grass pollen allergy may have wheat sensitization too, although this has been shown not to be of clinical relevance. This was reported by a study that was conducted on 63 children with doctor-diagnosed WA, sensitized to wheat and avoiding wheat in diet. Only 32 children were confirmed wheat-allergic based on OFC. Sensitization to grass was found in 73% of these children with significantly more sensitization in wheat allergic (n=32) as compared to non-allergic children (n=31). It was obvious from this study that many subjects were diagnosed by physicians to have WA but was shown not to react to wheat upon challenge. Thus, it was reported that cross-reactivity among grass pollens and wheat could give rise to false-positive sensitization results (44). Similarly, in another study conducted among a birth cohort of 827 children in UK, high levels of cross-sensitization was found between grass and wheat, however were clinically irrelevant (16).    
Further, in another study conducted on 72 grass pollen-allergic patients who were able to consume wheat, 60% showed IgE sensitivity to wheat. Further, specific IgE towards wheat-specific allergen components (Tri a 14 and Tri a 19) as well timothy grass-pollen allergens (Phl p 1, 2, 4, 5, 6, 7, 11 and 12) were evaluated. It was shown that sensitization to Phl p 12 was found to be significantly (p<0.05) more in patients sensitized to wheat. However, all the children were able to consume wheat without any significant allergic reactions. Physicians should handle with care a positive IgE test to wheat in a grass-allergic patient. If the patient is consuming wheat without problem, the test result is most likely due to cross reaction between wheat and grass. If high levels to the wheat test are found, a possible WA should be further investigated (53).
Cross-reactivity between wheat and kiwi fruit in patients with Baker’s asthma has been noted which may be due to thiol proteases from wheat and carhohydrate determinants from kiwi (1).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Magnus Borres
Last reviewed: February 2021
References
  1. Kleine-Tebbe Jea. Molecular Allergy Diagnostics Innovation for a Better Patient Management. 2017:Chapter 17.
  2. Curtis B. Wheat in the world. Rome: Food and Agricultural Organization of the United Nations; 2002. Available from: http://www.fao.org/3/y4011e04.htm#bm04.
  3. Cianferoni A. Wheat allergy: diagnosis and management. Journal of asthma and allergy. 2016;9:13.
  4. Impe D, Reitz J, Köpnick C, Rolletschek H, Börner A, Senula A, et al. Assessment of pollen viability for wheat. Frontiers in plant science. 2020;10:1588.
  5. Martín-Gómez JJ, Rewicz A, Goriewa-Duba K, Wiwart M, Tocino Á, Cervantes E. Morphological Description and Classification of Wheat Kernels Based on Geometric Models. 2019;9(7):399.
  6. Kirby EJ. Botany of the wheat plant. Rome: Food and Agriculture Organization of the United Nations; 2002. Available from: http://www.fao.org/3/y4011e05.htm#bm05.
  7. Wieser H, Koehler P, Scherf KA. The Two Faces of Wheat. Front Nutr. 2020;7:517313.
  8. Acevedo E, Silva P, Silva H. Wheat growth and physiology. Rome: Food and Agricultural Organization of the United Nations; 2002. Available from: http://www.fao.org/3/y4011e06.htm#bm06.
  9. CABI. Triticum aestivum (wheat): CAB International; 2019 [updated November 20, 2019; cited 2021 February 1, 2021]. Available from: https://www.cabi.org/isc/datasheet/55204#totaxonomicTree.
  10. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, et al. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016;27:1-250.
  11. Wujun M, Zitong Y, Maoyun S, Yun Z, Shahidul I. Wheat gluten protein and its impacts on wheat processing quality. 2019;6(3):279-87.
  12. Juhasz A, Belova T, Florides CG, Maulis C, Fischer I, Gell G, et al. Genome mapping of seed-borne allergens and immunoresponsive proteins in wheat. Sci Adv. 2018;4(8):eaar8602.
  13. WHO/IUIS. Triticum aestivum (Wheat): WHO/IUIS Allergen Nomenclature Sub-Committee; 2020 [updated May 12, 2020; cited 2021 February 1, 2021]. Available from: http://www.allergen.org/search.php?allergenname=&allergensource=Triticum+aestivum&TaxSource=&TaxOrder=&foodallerg=all&bioname=.
  14. Garcia-Molina MD, Gimenez MJ, Sanchez-Leon S, Barro F. Gluten Free Wheat: Are We There? Nutrients. 2019;11(3).
  15. Ricci G, Andreozzi L, Cipriani F, Giannetti A, Gallucci M, Caffarelli C. Wheat Allergy in Children: A Comprehensive Update. Medicina (Kaunas). 2019;55(7).
  16. Venter C, Maslin K, Arshad SH, Patil V, Grundy J, Glasbey G, et al. Very low prevalence of IgE mediated wheat allergy and high levels of cross-sensitisation between grass and wheat in a UK birth cohort. Clinical and translational allergy. 2016;6:22.
  17. Yagami A, Aihara M, Ikezawa Z, Hide M, Kishikawa R, Morita E, et al. Outbreak of immediate-type hydrolyzed wheat protein allergy due to a facial soap in Japan. Journal of Allergy and Clinical Immunology. 2017;140(3):879-81.e7.
  18. Jeong K, Kim J, Ahn K-M, Lee S-Y, Min T, Pyun B, et al. Age-Based Causes and Clinical Characteristics of Immediate-Type Food Allergy in Korean Children. Allergy, Asthma & Immunology Research. 2017;9:423.
  19. Ashtari S, Pourhoseingholi MA, Rostami K, Aghdaei HA, Rostami-Nejad M, Busani L, et al. Prevalence of gluten-related disorders in Asia-Pacific region: a systematic review. Journal of gastrointestinal and liver diseases : JGLD. 2019;28(1):95-105.
  20. Poole JA, Barriga K, Leung DY, Hoffman M, Eisenbarth GS, Rewers M, et al. Timing of initial exposure to cereal grains and the risk of wheat allergy. Pediatrics. 2006;117(6):2175-82.
  21. Zuidmeer L, Goldhahn K, Rona RJ, Gislason D, Madsen C, Summers C, et al. The prevalence of plant food allergies: A systematic review. Journal of Allergy and Clinical Immunology. 2008;121(5):1210-8.e4.
  22. Nwaru BI, Hickstein L, Panesar SS, Roberts G, Muraro A, Sheikh A. Prevalence of common food allergies in Europe: a systematic review and meta-analysis. Allergy. 2014;69(8):992-1007.
  23. Mäkelä MJ, Eriksson C, Kotaniemi-Syrjänen A, Palosuo K, Marsh J, Borres M, et al. Wheat allergy in children – new tools for diagnostics. 2014;44(11):1420-30.
  24. Dey D, Ghosh N, Pandey N, Gupta Bhattacharya S. A hospital-based survey on food allergy in the population of Kolkata, India. International archives of allergy and immunology. 2014;164(3):218-21.
  25. Cianferoni A, Khullar K, Saltzman R, Fiedler J, Garrett JP, Naimi DR, et al. Oral food challenge to wheat: a near-fatal anaphylaxis and review of 93 food challenges in children. The World Allergy Organization journal. 2013;6(1):14.
  26. Harada S, Horikawa T, Icihashi M. [A study of food-dependent exercise-induced anaphylaxis by analyzing the Japanese cases reported in the literature]. Arerugi = [Allergy]. 2000;49(11):1066-73.
  27. Thongngarm T, Wongsa C, Pacharn P, Piboonpocanun S, Sompornrattanaphan M. Clinical Characteristics and Proposed Wheat-Cofactor Challenge Protocol with a High Diagnostic Yield in Adult-Onset IgE-Mediated Wheat Allergy. J Asthma Allergy. 2020;13:355-68.
  28. Armentia A, Martin-Santos J, Quintero A, Fernandez A, Barber D, Alonso E, et al. Bakers' asthma: prevalence and evaluation of immunotherapy with a wheat flour extract. Annals of allergy. 1990;65:265-72.
  29. El-Sayed Z, Shousha G. Wheat allergy %J The Egyptian Journal of Pediatric Allergy and Immunology. 2020;18(2):55-60.
  30. Keet CA, Matsui EC, Dhillon G, Lenehan P, Paterakis M, Wood RA. The natural history of wheat allergy. Ann Allergy Asthma Immunol. 2009;102(5):410-5.
  31. Czaja-Bulsa G, Bulsa M. What do we know now about IgE-mediated wheat allergy in children? Nutrients. 2017;9(1):35.
  32. Tshikunde NM, Mashilo J, Shimelis H, Odindo A. Agronomic and Physiological Traits, and Associated Quantitative Trait Loci (QTL) Affecting Yield Response in Wheat (Triticum aestivum L.): A Review. Front Plant Sci. 2019;10:1428.
  33. Calamelli E, Liotti L, Beghetti I, Piccinno V, Serra L, Bottau P. Component-resolved diagnosis in food allergies. Medicina. 2019;55(8):498.
  34. Czaja-Bulsa G, Bulsa M. The natural history of IgE mediated wheat allergy in children with dominant gastrointestinal symptoms. Allergy Asthma Clin Immunol. 2014;10(1):12.
  35. Christensen MJ, Eller E, Mortz CG, Bindslev-Jensen C. Patterns of suspected wheat-related allergy: a retrospective single-centre case note review in 156 patients. Clinical and translational allergy. 2014;4:39.
  36. Srisuwatchari W, Vichyanond P, Jirapongsananuruk O, Visitsunthorn N, Pacharn P. Characterization of children with IgE-mediated wheat allergy and risk factors that predict wheat anaphylaxis. Asian Pacific journal of allergy and immunology. 2020a.
  37. Scherf KA, Brockow K, Biedermann T, Koehler P, Wieser H. Wheat-dependent exercise-induced anaphylaxis. Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology. 2016;46(1):10-20.
  38. Kennard L, Thomas I, Rutkowski K, Azzu V, Yong PFK, Kasternow B, et al. A Multicenter Evaluation of Diagnosis and Management of Omega-5 Gliadin Allergy (Also Known as Wheat-Dependent Exercise-Induced Anaphylaxis) in 132 Adults. J Allergy Clin Immunol Pract. 2018;6(6):1892-7.
  39. Elli L, Tomba C, Branchi F, Roncoroni L, Lombardo V, Bardella MT, et al. Evidence for the Presence of Non-Celiac Gluten Sensitivity in Patients with Functional Gastrointestinal Symptoms: Results from a Multicenter Randomized Double-Blind Placebo-Controlled Gluten Challenge. Nutrients. 2016;8(2):84.
  40. Quirce S, Boyano-Martínez T, Díaz-Perales A. Clinical presentation, allergens, and management of wheat allergy. Expert review of clinical immunology. 2016;12(5):563-72.
  41. Hofer G, Wieser S, Bogdos MK, Gattinger P, Nakamura R, Ebisawa M, et al. Three-dimensional structure of the wheat β-amylase Tri a 17, a clinically relevant food allergen. Allergy. 2019;74(5):1009-13.
  42. Sander I, Rihs HP, Doekes G, Quirce S, Krop E, Rozynek P, et al. Component-resolved diagnosis of baker's allergy based on specific IgE to recombinant wheat flour proteins. The Journal of allergy and clinical immunology. 2015;135(6):1529-37.
  43. Ito K, Futamura M, Borres MP, Takaoka Y, Dahlstrom J, Sakamoto T, et al. IgE antibodies to omega-5 gliadin associate with immediate symptoms on oral wheat challenge in Japanese children. Allergy. 2008;63(11):1536-42.
  44. Nilsson N, Sjölander S, Baar A, Berthold M, Pahr S, Vrtala S, et al. Wheat allergy in children evaluated with challenge and IgE antibodies to wheat components. 2015;26(2):119-25.
  45. Ebisawa M, Shibata R, Sato S, Borres MP, Ito K. Clinical utility of IgE antibodies to omega-5 gliadin in the diagnosis of wheat allergy: a pediatric multicenter challenge study. International archives of allergy and immunology. 2012;158(1):71-6.
  46. Pourpak Z, Mesdaghi M, Mansouri M, Kazemnejad A, Toosi SB, Farhoudi A. Which cereal is a suitable substitute for wheat in children with wheat allergy? Pediatr Allergy Immunol. 2005;16(3):262-6.
  47. Kliewer K, Venter C, Cassin A, Abonia J, Aceves S, Bonis P, et al. Should wheat, barley, rye, and/or gluten be avoided in a 6-food elimination diet? Journal of Allergy and Clinical Immunology. 2015;137.
  48. Palosuo K, Alenius H, Varjonen E, Kalkkinen N, Reunala T. Rye gamma-70 and gamma-35 secalins and barley gamma-3 hordein cross-react with omega-5 gliadin, a major allergen in wheat-dependent, exercise-induced anaphylaxis. Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology. 2001;31(3):466-73.
  49. Lee E, Jeong K, Lee J, Jeon SA, Park B, Lee H, et al. Clinical and Laboratory Findings of Barley Allergy in Korean Children: a Single Hospital Based Retrospective Study. Journal of Korean medical science. 2020;35(3):e23.
  50. Srisuwatchari W, Piboonpocanun S, Wangthan U, Jirapongsananuruk O, Visitsunthorn N, Pacharn P. Clinical and in vitro cross-reactivity of cereal grains in children with IgE-mediated wheat allergy. Allergologia et immunopathologia. 2020b;48(6):589-96.
  51. Golias J, Humlova Z, Halada P, Habova V, Janatkova I, Tuckova L. Identification of rice proteins recognized by the IgE antibodies of patients with food allergies. J Agric Food Chem. 2013;61(37):8851-60.
  52. Pastorello EA, Farioli L, Pravettoni V, Ispano M, Scibola E, Trambaioli C, et al. The maize major allergen, which is responsible for food-induced allergic reactions, is a lipid transfer protein. The Journal of allergy and clinical immunology. 2000;106(4):744-51.
  53. Nilsson N, Nilsson C, Ekoff H, Wieser-Pahr S, Borres MP, Valenta R, et al. Grass-Allergic Children Frequently Show Asymptomatic Low-Level IgE Co-Sensitization and Cross-Reactivity to Wheat. International archives of allergy and immunology. 2018;177(2):135-44.