Type:
Whole Allergen
Whole Allergen
Whole Allergen
Douglas fir
t207
Pinaceae
Pseudotsuga taxifolia
Douglas fir tree, Oregon pine, yellow fir
Anecdotal evidence suggests that asthma, allergic rhinitis and allergic conjunctivitis, similar to reactions caused by other members of this family, are possible following exposure to pollen from this tree; however, no specific studies have been reported to date.
The Douglas fir is not an important allergen in urbanized communities – except those situated close to Douglas fir plantations, where the prevalence of atopic sensitization to pollen from this tree is expected to be high.
The tussock moth caterpillar, occurring in forested areas of Oregon, was associated with itching of the skin and eyes, nasal discharge, cough, and, at times, respiratory difficulty in 41 of 428 individuals working in a forest. (1)
No allergens from this plant have yet been characterized.
Extensive cross-reactivity between the different individual species of the genus could be expected. (2)
Last Reviewed- April 2022